Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190978765-190979266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT1
Entrez Gene ID
6772
Gene Name
signal transducer and activator of transcription 1
Gene Aliases
CANDF7, IMD31A, IMD31B, IMD31C, ISGF-3, STAT91
UniGene
Hs.642990
Gene Chromosome Location
Chr.2: 190969036-191014250 on Build GRCh38
Transcripts
NM_007315.3 NM_139266.2 XM_006712718.1 XM_017004783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6437308 c.1867G>C p.G623R Chr.2: 190978862-190978862 on Build GRCh38
6791654 c.1855C>T p.R619W Chr.2: 190978874-190978874 on Build GRCh38
4089730 c.1825G>A p.E609K Chr.2: 190978904-190978904 on Build GRCh38
1404122 c.1822C>T p.R608W Chr.2: 190978907-190978907 on Build GRCh38
3838164 c.1778A>T p.D593V Chr.2: 190978951-190978951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546180422 Chr.2: 190979111-190979111 on Build GRCh38
rs199812941 Chr.2: 190978899-190978899 on Build GRCh38
rs915052817 Chr.2: 190979008-190979008 on Build GRCh38
rs750574793 Chr.2: 190979001-190979001 on Build GRCh38
rs759270868 Chr.2: 190978818-190978818 on Build GRCh38
rs587776713 Chr.2: 190978968-190978969 on Build GRCh38
rs112649322 Chr.2: 190978789-190978789 on Build GRCh38
rs377602295 Chr.2: 190978824-190978824 on Build GRCh38
rs55956636 Chr.2: 190978825-190978825 on Build GRCh38
rs1037623370 Chr.2: 190979148-190979148 on Build GRCh38
rs762754243 Chr.2: 190978974-190978974 on Build GRCh38
rs567722352 Chr.2: 190978941-190978941 on Build GRCh38
rs368375652 Chr.2: 190978835-190978835 on Build GRCh38
rs780868200 Chr.2: 190979033-190979034 on Build GRCh38
rs922715383 Chr.2: 190979048-190979048 on Build GRCh38
rs376949116 Chr.2: 190979022-190979022 on Build GRCh38
rs910373622 Chr.2: 190979110-190979110 on Build GRCh38
rs768531104 Chr.2: 190978994-190978995 on Build GRCh38
rs376916250 Chr.2: 190978884-190978884 on Build GRCh38
rs745491762 Chr.2: 190978964-190978964 on Build GRCh38
rs754322162 Chr.2: 190978875-190978875 on Build GRCh38
rs905893914 Chr.2: 190979096-190979097 on Build GRCh38
rs749906520 Chr.2: 190978989-190978989 on Build GRCh38
rs375604257 Chr.2: 190979018-190979018 on Build GRCh38
rs532099636 Chr.2: 190978795-190978795 on Build GRCh38
rs758673338 Chr.2: 190979185-190979185 on Build GRCh38
rs761906729 Chr.2: 190978845-190978845 on Build GRCh38
rs976349556 Chr.2: 190979244-190979244 on Build GRCh38
rs746476886 Chr.2: 190978932-190978932 on Build GRCh38
rs181473662 Chr.2: 190979093-190979093 on Build GRCh38
rs137852678 Chr.2: 190978930-190978930 on Build GRCh38
rs1013538894 Chr.2: 190978902-190978902 on Build GRCh38
rs1007166025 Chr.2: 190979234-190979234 on Build GRCh38
rs190272459 Chr.2: 190978796-190978796 on Build GRCh38
rs776177724 Chr.2: 190978815-190978815 on Build GRCh38
rs34945497 Chr.2: 190979089-190979090 on Build GRCh38
rs2230101 Chr.2: 190978857-190978857 on Build GRCh38
rs144788879 Chr.2: 190978972-190978972 on Build GRCh38
rs796183827 Chr.2: 190978810-190978811 on Build GRCh38
rs1027186926 Chr.2: 190979165-190979165 on Build GRCh38
rs775409051 Chr.2: 190978821-190978821 on Build GRCh38
rs767740977 Chr.2: 190978849-190978849 on Build GRCh38
rs539295815 Chr.2: 190979146-190979146 on Build GRCh38
rs764915115 Chr.2: 190978819-190978819 on Build GRCh38
rs371147071 Chr.2: 190978983-190978983 on Build GRCh38
rs59582019 Chr.2: 190979036-190979036 on Build GRCh38
rs767475430 Chr.2: 190978996-190978996 on Build GRCh38
rs553747054 Chr.2: 190979098-190979098 on Build GRCh38
rs746699623 Chr.2: 190978992-190978993 on Build GRCh38
rs965027706 Chr.2: 190979243-190979243 on Build GRCh38
rs753230840 Chr.2: 190979023-190979023 on Build GRCh38
rs1018928447 Chr.2: 190979237-190979237 on Build GRCh38
rs778428911 Chr.2: 190978818-190978819 on Build GRCh38
rs897512492 Chr.2: 190978808-190978808 on Build GRCh38
rs564897518 Chr.2: 190978792-190978792 on Build GRCh38
rs758038647 Chr.2: 190978989-190978990 on Build GRCh38
rs754206062 Chr.2: 190979016-190979016 on Build GRCh38
rs377699474 Chr.2: 190979190-190979190 on Build GRCh38
rs554765236 Chr.2: 190979174-190979174 on Build GRCh38
rs112108750 Chr.2: 190978863-190978863 on Build GRCh38
rs761780153 Chr.2: 190978995-190978995 on Build GRCh38
rs1057253552 Chr.2: 190979124-190979124 on Build GRCh38
rs556189473 Chr.2: 190979039-190979039 on Build GRCh38
rs779830895 Chr.2: 190978990-190978991 on Build GRCh38
rs770315023 Chr.2: 190978945-190978945 on Build GRCh38
rs766636931 Chr.2: 190979011-190979011 on Build GRCh38
rs369060692 Chr.2: 190978873-190978873 on Build GRCh38
rs571315523 Chr.2: 190979053-190979053 on Build GRCh38
rs780801370 Chr.2: 190978947-190978947 on Build GRCh38
rs750571328 Chr.2: 190979144-190979144 on Build GRCh38
rs760805208 Chr.2: 190979005-190979005 on Build GRCh38
rs779585092 Chr.2: 190978890-190978890 on Build GRCh38
rs2280235 Chr.2: 190979104-190979104 on Build GRCh38
rs774306421 Chr.2: 190978985-190978985 on Build GRCh38

View Full Product Details