Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 81942882-81943385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL2
Entrez Gene ID
23266
Gene Name
adhesion G protein-coupled receptor L2
Gene Aliases
CIRL2, CL2, LEC1, LPHH1, LPHN2
UniGene
Hs.24212
Gene Chromosome Location
Chr.1: 81306155-81992738 on Build GRCh38
Transcripts
NM_001297704.1 NM_001297705.1 NM_001297706.1 NM_012302.3 XM_005270666.4 XM_005270668.2 XM_006710485.3 XM_006710488.2 XM_006710489.3 XM_017000782.1 XM_017000783.1 XM_017000784.1 XM_017000785.1 XM_017000786.1 XM_017000787.1 XM_017000788.1 XM_017000789.1 XM_017000790.1 XM_017000791.1 XM_017000792.1 XM_017000793.1 XM_017000794.1 XM_017000795.1 XM_017000796.1 XM_017000797.1 XM_017000798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5751415 c.467_468insT p.A158fs*12 Chr.1: 81943038-81943039 on Build GRCh38
5456666 c.677C>T p.T226M Chr.1: 81943248-81943248 on Build GRCh38
7147287 c.528C>G p.F176L Chr.1: 81943087-81943087 on Build GRCh38
6255177 c.638T>C p.V213A Chr.1: 81943209-81943209 on Build GRCh38
4698463 c.652G>T p.A218S Chr.1: 81943223-81943223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765365131 Chr.1: 81943336-81943336 on Build GRCh38
rs537681838 Chr.1: 81942923-81942923 on Build GRCh38
rs757586532 Chr.1: 81942956-81942961 on Build GRCh38
rs1002997342 Chr.1: 81942980-81942980 on Build GRCh38
rs200377136 Chr.1: 81943038-81943038 on Build GRCh38
rs773672775 Chr.1: 81943147-81943147 on Build GRCh38
rs1035652537 Chr.1: 81943017-81943017 on Build GRCh38
rs201538464 Chr.1: 81943146-81943146 on Build GRCh38
rs773817244 Chr.1: 81943271-81943271 on Build GRCh38
rs761033313 Chr.1: 81943051-81943051 on Build GRCh38
rs756262757 Chr.1: 81942933-81942933 on Build GRCh38
rs776948351 Chr.1: 81943171-81943171 on Build GRCh38
rs769181656 Chr.1: 81943025-81943025 on Build GRCh38
rs767655251 Chr.1: 81942918-81942918 on Build GRCh38
rs753951750 Chr.1: 81942941-81942941 on Build GRCh38
rs192401909 Chr.1: 81943244-81943244 on Build GRCh38
rs777738816 Chr.1: 81942950-81942950 on Build GRCh38
rs187930349 Chr.1: 81943050-81943050 on Build GRCh38
rs765294200 Chr.1: 81943195-81943195 on Build GRCh38
rs750601144 Chr.1: 81942930-81942930 on Build GRCh38
rs947062679 Chr.1: 81943156-81943156 on Build GRCh38
rs190614386 Chr.1: 81942957-81942957 on Build GRCh38
rs893280459 Chr.1: 81942915-81942915 on Build GRCh38
rs767336672 Chr.1: 81943223-81943223 on Build GRCh38
rs565066593 Chr.1: 81943249-81943249 on Build GRCh38
rs767511606 Chr.1: 81943342-81943342 on Build GRCh38
rs867891967 Chr.1: 81943245-81943245 on Build GRCh38
rs774724301 Chr.1: 81943130-81943130 on Build GRCh38
rs780721378 Chr.1: 81943004-81943004 on Build GRCh38
rs182862761 Chr.1: 81943023-81943023 on Build GRCh38
rs778539325 Chr.1: 81943250-81943250 on Build GRCh38
rs745459122 Chr.1: 81943010-81943010 on Build GRCh38
rs768154117 Chr.1: 81943126-81943126 on Build GRCh38
rs1051598019 Chr.1: 81943221-81943221 on Build GRCh38
rs750247592 Chr.1: 81943229-81943229 on Build GRCh38
rs767851795 Chr.1: 81943039-81943039 on Build GRCh38
rs1026922526 Chr.1: 81943152-81943152 on Build GRCh38
rs372512578 Chr.1: 81943267-81943267 on Build GRCh38
rs755815823 Chr.1: 81943112-81943112 on Build GRCh38
rs750832623 Chr.1: 81942966-81942967 on Build GRCh38
rs775689046 Chr.1: 81943340-81943340 on Build GRCh38
rs780344548 Chr.1: 81942940-81942940 on Build GRCh38
rs868376442 Chr.1: 81943347-81943347 on Build GRCh38
rs755853238 Chr.1: 81943233-81943233 on Build GRCh38
rs577293051 Chr.1: 81943158-81943158 on Build GRCh38
rs761088337 Chr.1: 81943154-81943154 on Build GRCh38
rs1041419067 Chr.1: 81943157-81943157 on Build GRCh38
rs762913159 Chr.1: 81943341-81943341 on Build GRCh38
rs561209754 Chr.1: 81943319-81943319 on Build GRCh38
rs765509598 Chr.1: 81942963-81942964 on Build GRCh38
rs748752495 Chr.1: 81943139-81943139 on Build GRCh38
rs755073321 Chr.1: 81943081-81943081 on Build GRCh38
rs865853749 Chr.1: 81943217-81943217 on Build GRCh38
rs777597517 Chr.1: 81943088-81943088 on Build GRCh38
rs771368684 Chr.1: 81943294-81943294 on Build GRCh38
rs557356494 Chr.1: 81942998-81942998 on Build GRCh38
rs144043060 Chr.1: 81943282-81943282 on Build GRCh38
rs976533411 Chr.1: 81943256-81943256 on Build GRCh38
rs781109858 Chr.1: 81943111-81943111 on Build GRCh38
rs144007597 Chr.1: 81942987-81942987 on Build GRCh38
rs759705946 Chr.1: 81943172-81943172 on Build GRCh38
rs146694157 Chr.1: 81943266-81943266 on Build GRCh38
rs866124798 Chr.1: 81943349-81943349 on Build GRCh38
rs747586277 Chr.1: 81943252-81943252 on Build GRCh38
rs369488419 Chr.1: 81943012-81943012 on Build GRCh38
rs755906651 Chr.1: 81943361-81943361 on Build GRCh38
rs750301010 Chr.1: 81943352-81943352 on Build GRCh38
rs754013841 Chr.1: 81943067-81943067 on Build GRCh38
rs748873382 Chr.1: 81943246-81943246 on Build GRCh38
rs1012139115 Chr.1: 81942917-81942917 on Build GRCh38
rs748818234 Chr.1: 81943118-81943118 on Build GRCh38
rs911093044 Chr.1: 81943198-81943198 on Build GRCh38
rs139062578 Chr.1: 81942908-81942908 on Build GRCh38
rs370761184 Chr.1: 81942942-81942942 on Build GRCh38
rs751460021 Chr.1: 81943093-81943093 on Build GRCh38
rs200603368 Chr.1: 81943297-81943297 on Build GRCh38
rs752716210 Chr.1: 81943213-81943213 on Build GRCh38
rs985685684 Chr.1: 81943166-81943166 on Build GRCh38
rs766684456 Chr.1: 81943060-81943060 on Build GRCh38
rs775041759 Chr.1: 81943016-81943016 on Build GRCh38
rs752679138 Chr.1: 81943106-81943106 on Build GRCh38
rs774872645 Chr.1: 81943030-81943030 on Build GRCh38
rs757278542 Chr.1: 81943219-81943219 on Build GRCh38

View Full Product Details