Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84495745-84496285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPF1
Entrez Gene ID
80135
Gene Name
ribosome production factor 1 homolog
Gene Aliases
BXDC5
UniGene
Hs.481202
Gene Chromosome Location
Chr.1: 84479237-84498350 on Build GRCh38
Transcripts
NM_025065.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738514 c.803C>T p.P268L Chr.1: 84495985-84495985 on Build GRCh38
6769061 c.780A>G p.G260G Chr.1: 84495962-84495962 on Build GRCh38
3419522 c.721G>A p.E241K Chr.1: 84495903-84495903 on Build GRCh38
5648834 c.823G>A p.G275R Chr.1: 84496005-84496005 on Build GRCh38
214286 c.701G>C p.R234T Chr.1: 84495883-84495883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886092144 Chr.1: 84496072-84496072 on Build GRCh38
rs768069827 Chr.1: 84495978-84495978 on Build GRCh38
rs770862630 Chr.1: 84496005-84496005 on Build GRCh38
rs763422975 Chr.1: 84495975-84495976 on Build GRCh38
rs751111118 Chr.1: 84495976-84495976 on Build GRCh38
rs553799862 Chr.1: 84495799-84495799 on Build GRCh38
rs1012457067 Chr.1: 84496157-84496157 on Build GRCh38
rs755237626 Chr.1: 84496202-84496202 on Build GRCh38
rs749561577 Chr.1: 84495988-84495988 on Build GRCh38
rs896822774 Chr.1: 84495777-84495777 on Build GRCh38
rs372051660 Chr.1: 84495934-84495934 on Build GRCh38
rs775118446 Chr.1: 84496022-84496022 on Build GRCh38
rs746221614 Chr.1: 84495882-84495882 on Build GRCh38
rs376300199 Chr.1: 84495957-84495957 on Build GRCh38
rs368861828 Chr.1: 84496220-84496220 on Build GRCh38
rs575850309 Chr.1: 84495924-84495924 on Build GRCh38
rs774733749 Chr.1: 84496047-84496049 on Build GRCh38
rs1022542187 Chr.1: 84496143-84496143 on Build GRCh38
rs756758387 Chr.1: 84496229-84496230 on Build GRCh38
rs758428600 Chr.1: 84496209-84496209 on Build GRCh38
rs565891899 Chr.1: 84496175-84496175 on Build GRCh38
rs199978767 Chr.1: 84496047-84496047 on Build GRCh38
rs540950254 Chr.1: 84495953-84495953 on Build GRCh38
rs375382930 Chr.1: 84495936-84495936 on Build GRCh38
rs1045337946 Chr.1: 84496232-84496232 on Build GRCh38
rs895300290 Chr.1: 84496124-84496124 on Build GRCh38
rs773973055 Chr.1: 84496150-84496150 on Build GRCh38
rs978066096 Chr.1: 84496153-84496153 on Build GRCh38
rs560629472 Chr.1: 84496205-84496205 on Build GRCh38
rs766622034 Chr.1: 84496014-84496015 on Build GRCh38
rs781733243 Chr.1: 84495877-84495877 on Build GRCh38
rs373661869 Chr.1: 84495998-84495998 on Build GRCh38
rs773292230 Chr.1: 84495860-84495862 on Build GRCh38
rs778812271 Chr.1: 84495922-84495922 on Build GRCh38
rs761366476 Chr.1: 84496037-84496037 on Build GRCh38
rs778168498 Chr.1: 84495847-84495847 on Build GRCh38
rs760054903 Chr.1: 84496093-84496094 on Build GRCh38
rs762734737 Chr.1: 84496024-84496024 on Build GRCh38
rs554976621 Chr.1: 84496017-84496017 on Build GRCh38
rs756545807 Chr.1: 84496059-84496059 on Build GRCh38
rs1029767464 Chr.1: 84496174-84496174 on Build GRCh38
rs941176137 Chr.1: 84496209-84496216 on Build GRCh38
rs767952587 Chr.1: 84496218-84496222 on Build GRCh38
rs774012681 Chr.1: 84496034-84496034 on Build GRCh38
rs75767109 Chr.1: 84495805-84495805 on Build GRCh38
rs369066394 Chr.1: 84496091-84496091 on Build GRCh38
rs756495833 Chr.1: 84495986-84495986 on Build GRCh38
rs769616392 Chr.1: 84496215-84496215 on Build GRCh38
rs149970308 Chr.1: 84496004-84496004 on Build GRCh38
rs573215493 Chr.1: 84495787-84495787 on Build GRCh38
rs139410871 Chr.1: 84495942-84495942 on Build GRCh38
rs748262403 Chr.1: 84496206-84496206 on Build GRCh38
rs529910681 Chr.1: 84496044-84496044 on Build GRCh38
rs753353389 Chr.1: 84496226-84496227 on Build GRCh38
rs72948448 Chr.1: 84496190-84496190 on Build GRCh38
rs752687482 Chr.1: 84495768-84495769 on Build GRCh38
rs1039321907 Chr.1: 84496030-84496030 on Build GRCh38
rs918477746 Chr.1: 84496227-84496227 on Build GRCh38
rs755148640 Chr.1: 84495989-84495989 on Build GRCh38
rs770838128 Chr.1: 84495939-84495939 on Build GRCh38
rs368932617 Chr.1: 84495943-84495943 on Build GRCh38
rs989889163 Chr.1: 84495912-84495912 on Build GRCh38
rs752156349 Chr.1: 84495831-84495831 on Build GRCh38
rs776514315 Chr.1: 84496012-84496012 on Build GRCh38
rs768385168 Chr.1: 84496237-84496237 on Build GRCh38
rs377031927 Chr.1: 84495837-84495837 on Build GRCh38
rs970890891 Chr.1: 84495795-84495795 on Build GRCh38
rs185909125 Chr.1: 84495908-84495908 on Build GRCh38
rs757809441 Chr.1: 84495872-84495872 on Build GRCh38
rs763702720 Chr.1: 84495964-84495964 on Build GRCh38
rs911089843 Chr.1: 84496203-84496203 on Build GRCh38
rs754136089 Chr.1: 84496107-84496107 on Build GRCh38
rs757888239 Chr.1: 84495834-84495834 on Build GRCh38
rs752158955 Chr.1: 84495855-84495855 on Build GRCh38
rs1011193116 Chr.1: 84496137-84496137 on Build GRCh38
rs748978580 Chr.1: 84496236-84496236 on Build GRCh38
rs763640936 Chr.1: 84496025-84496025 on Build GRCh38
rs769682938 Chr.1: 84496018-84496018 on Build GRCh38
rs771809205 Chr.1: 84496238-84496238 on Build GRCh38
rs1003269145 Chr.1: 84496076-84496076 on Build GRCh38
rs959266355 Chr.1: 84496151-84496151 on Build GRCh38
rs200701602 Chr.1: 84495985-84495985 on Build GRCh38
rs768828524 Chr.1: 84495932-84495932 on Build GRCh38
rs774098385 Chr.1: 84496239-84496239 on Build GRCh38
rs915132983 Chr.1: 84495827-84495827 on Build GRCh38
rs777811366 Chr.1: 84496210-84496210 on Build GRCh38
rs780282219 Chr.1: 84495913-84495913 on Build GRCh38
rs780259031 Chr.1: 84495987-84495987 on Build GRCh38
rs959447702 Chr.1: 84495789-84495789 on Build GRCh38
rs745848981 Chr.1: 84496211-84496211 on Build GRCh38
rs765001724 Chr.1: 84496244-84496247 on Build GRCh38
rs966724950 Chr.1: 84496147-84496147 on Build GRCh38
rs1033812960 Chr.1: 84496118-84496118 on Build GRCh38
rs770205933 Chr.1: 84495857-84495860 on Build GRCh38
rs985701882 Chr.1: 84496192-84496192 on Build GRCh38
rs762494204 Chr.1: 84495963-84495963 on Build GRCh38
rs568246604 Chr.1: 84495778-84495778 on Build GRCh38
rs748183638 Chr.1: 84495838-84495842 on Build GRCh38

View Full Product Details