Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30557184-30557689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL3ST1
Entrez Gene ID
9514
Gene Name
galactose-3-O-sulfotransferase 1
Gene Aliases
CST
UniGene
Hs.17958
Gene Chromosome Location
Chr.22: 30554635-30574588 on Build GRCh38
Transcripts
NM_001318103.1 NM_001318104.1 NM_001318105.1 NM_001318106.1 NM_001318107.1 NM_001318108.1 NM_001318109.1 NM_001318110.1 NM_001318111.1 NM_001318112.1 NM_001318113.1 NM_001318114.1 NM_001318115.1 NM_001318116.1 NM_004861.2 XM_011530518.2 XM_011530522.2 XM_011530524.1 XM_011530528.1 XM_017029096.1 XM_017029097.1 XM_017029098.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1177572 c.67A>T p.T23S Chr.22: 30557326-30557326 on Build GRCh38
5909886 c.129C>T p.S43S Chr.22: 30557264-30557264 on Build GRCh38
4847151 c.128C>G p.S43C Chr.22: 30557265-30557265 on Build GRCh38
5619562 c.100G>A p.V34M Chr.22: 30557293-30557293 on Build GRCh38
4573054 c.94T>C p.Y32H Chr.22: 30557299-30557299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2267161 Chr.22: 30557308-30557308 on Build GRCh38
rs752706235 Chr.22: 30557383-30557383 on Build GRCh38
rs112410786 Chr.22: 30557540-30557540 on Build GRCh38
rs1016336933 Chr.22: 30557616-30557617 on Build GRCh38
rs982383397 Chr.22: 30557488-30557488 on Build GRCh38
rs765213740 Chr.22: 30557318-30557318 on Build GRCh38
rs752866163 Chr.22: 30557333-30557333 on Build GRCh38
rs768132159 Chr.22: 30557371-30557371 on Build GRCh38
rs899162748 Chr.22: 30557621-30557621 on Build GRCh38
rs760984952 Chr.22: 30557375-30557375 on Build GRCh38
rs984406083 Chr.22: 30557591-30557591 on Build GRCh38
rs55674628 Chr.22: 30557293-30557293 on Build GRCh38
rs376326152 Chr.22: 30557262-30557262 on Build GRCh38
rs754376240 Chr.22: 30557312-30557312 on Build GRCh38
rs1022151928 Chr.22: 30557207-30557207 on Build GRCh38
rs199701190 Chr.22: 30557382-30557382 on Build GRCh38
rs757124639 Chr.22: 30557339-30557339 on Build GRCh38
rs42927 Chr.22: 30557637-30557637 on Build GRCh38
rs369562134 Chr.22: 30557304-30557304 on Build GRCh38
rs564953383 Chr.22: 30557599-30557599 on Build GRCh38
rs566183603 Chr.22: 30557513-30557513 on Build GRCh38
rs758281604 Chr.22: 30557330-30557330 on Build GRCh38
rs756053698 Chr.22: 30557424-30557424 on Build GRCh38
rs755039523 Chr.22: 30557317-30557317 on Build GRCh38
rs1019139223 Chr.22: 30557241-30557241 on Build GRCh38
rs961649507 Chr.22: 30557605-30557605 on Build GRCh38
rs753853924 Chr.22: 30557261-30557261 on Build GRCh38
rs200175330 Chr.22: 30557274-30557274 on Build GRCh38
rs545072220 Chr.22: 30557268-30557268 on Build GRCh38
rs138906121 Chr.22: 30557552-30557552 on Build GRCh38
rs147005072 Chr.22: 30557276-30557276 on Build GRCh38
rs900773355 Chr.22: 30557566-30557566 on Build GRCh38
rs751342433 Chr.22: 30557336-30557336 on Build GRCh38
rs754573941 Chr.22: 30557462-30557462 on Build GRCh38
rs368052166 Chr.22: 30557314-30557314 on Build GRCh38
rs910114682 Chr.22: 30557292-30557292 on Build GRCh38
rs923656152 Chr.22: 30557507-30557507 on Build GRCh38
rs764346414 Chr.22: 30557411-30557411 on Build GRCh38
rs373103780 Chr.22: 30557428-30557428 on Build GRCh38
rs547200013 Chr.22: 30557294-30557294 on Build GRCh38
rs763108641 Chr.22: 30557399-30557399 on Build GRCh38
rs202165827 Chr.22: 30557345-30557345 on Build GRCh38
rs114519146 Chr.22: 30557598-30557598 on Build GRCh38
rs750827035 Chr.22: 30557225-30557225 on Build GRCh38
rs781102799 Chr.22: 30557441-30557441 on Build GRCh38
rs773548824 Chr.22: 30557298-30557298 on Build GRCh38
rs973316334 Chr.22: 30557625-30557625 on Build GRCh38
rs552608750 Chr.22: 30557420-30557420 on Build GRCh38
rs776725191 Chr.22: 30557286-30557286 on Build GRCh38
rs532615501 Chr.22: 30557323-30557323 on Build GRCh38
rs999348961 Chr.22: 30557234-30557234 on Build GRCh38
rs74418371 Chr.22: 30557216-30557216 on Build GRCh38
rs148849932 Chr.22: 30557378-30557378 on Build GRCh38
rs766926134 Chr.22: 30557245-30557245 on Build GRCh38
rs997680188 Chr.22: 30557573-30557573 on Build GRCh38
rs745329677 Chr.22: 30557288-30557288 on Build GRCh38
rs770980558 Chr.22: 30557285-30557285 on Build GRCh38
rs575958807 Chr.22: 30557570-30557570 on Build GRCh38
rs756605361 Chr.22: 30557227-30557227 on Build GRCh38
rs777033091 Chr.22: 30557379-30557379 on Build GRCh38
rs769808879 Chr.22: 30557356-30557356 on Build GRCh38
rs138460948 Chr.22: 30557365-30557365 on Build GRCh38
rs4149490 Chr.22: 30557534-30557534 on Build GRCh38
rs150130261 Chr.22: 30557340-30557340 on Build GRCh38
rs778200200 Chr.22: 30557335-30557335 on Build GRCh38
rs200784913 Chr.22: 30557334-30557334 on Build GRCh38
rs942393122 Chr.22: 30557541-30557541 on Build GRCh38
rs773993854 Chr.22: 30557374-30557374 on Build GRCh38
rs375791748 Chr.22: 30557370-30557370 on Build GRCh38
rs4149491 Chr.22: 30557515-30557515 on Build GRCh38
rs34136895 Chr.22: 30557256-30557256 on Build GRCh38
rs370792583 Chr.22: 30557381-30557381 on Build GRCh38
rs763869576 Chr.22: 30557296-30557296 on Build GRCh38
rs776387935 Chr.22: 30557263-30557263 on Build GRCh38
rs555193878 Chr.22: 30557520-30557520 on Build GRCh38
rs769209210 Chr.22: 30557290-30557290 on Build GRCh38

View Full Product Details