Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23910619-23911117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKIRAS1
Entrez Gene ID
28512
Gene Name
NFKB inhibitor interacting Ras like 1
Gene Aliases
KBRAS1, kappaB-Ras1
UniGene
Hs.173202
Gene Chromosome Location
Chr.3: 23889536-23917046 on Build GRCh38
Transcripts
NM_020345.3 XM_005265076.3 XM_005265077.4 XM_005265078.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
212773 c.47G>T p.G16V Chr.3: 23910858-23910858 on Build GRCh38
4116605 c.39A>T p.L13F Chr.3: 23910866-23910866 on Build GRCh38
4116606 c.4G>T p.G2* Chr.3: 23910901-23910901 on Build GRCh38
353206 c.7A>C p.K3Q Chr.3: 23910898-23910898 on Build GRCh38
3592068 c.72C>T p.L24L Chr.3: 23910833-23910833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374408719 Chr.3: 23910927-23910927 on Build GRCh38
rs533663280 Chr.3: 23910990-23910990 on Build GRCh38
rs529115681 Chr.3: 23910744-23910744 on Build GRCh38
rs777729200 Chr.3: 23910836-23910836 on Build GRCh38
rs34993633 Chr.3: 23910803-23910803 on Build GRCh38
rs950367412 Chr.3: 23910862-23910862 on Build GRCh38
rs759865821 Chr.3: 23910938-23910938 on Build GRCh38
rs762202193 Chr.3: 23910894-23910894 on Build GRCh38
rs969833097 Chr.3: 23911021-23911021 on Build GRCh38
rs1054506498 Chr.3: 23910989-23910989 on Build GRCh38
rs949974095 Chr.3: 23910959-23910959 on Build GRCh38
rs370210022 Chr.3: 23910907-23910907 on Build GRCh38
rs946066022 Chr.3: 23911019-23911019 on Build GRCh38
rs751833204 Chr.3: 23910819-23910819 on Build GRCh38
rs891829525 Chr.3: 23910702-23910702 on Build GRCh38
rs766685636 Chr.3: 23910860-23910860 on Build GRCh38
rs149013494 Chr.3: 23910767-23910767 on Build GRCh38
rs756159428 Chr.3: 23911027-23911027 on Build GRCh38
rs143799784 Chr.3: 23911017-23911017 on Build GRCh38
rs550963927 Chr.3: 23910814-23910814 on Build GRCh38
rs781762033 Chr.3: 23910988-23910988 on Build GRCh38
rs1003072895 Chr.3: 23911024-23911024 on Build GRCh38
rs768685247 Chr.3: 23911082-23911082 on Build GRCh38
rs771808017 Chr.3: 23910760-23910760 on Build GRCh38
rs748980541 Chr.3: 23910763-23910764 on Build GRCh38
rs775103607 Chr.3: 23910849-23910849 on Build GRCh38
rs763021396 Chr.3: 23910910-23910910 on Build GRCh38
rs921325171 Chr.3: 23910857-23910857 on Build GRCh38
rs570099764 Chr.3: 23911004-23911004 on Build GRCh38
rs764449676 Chr.3: 23910784-23910784 on Build GRCh38
rs1027404609 Chr.3: 23910805-23910805 on Build GRCh38
rs777065820 Chr.3: 23910785-23910785 on Build GRCh38
rs372391274 Chr.3: 23910829-23910829 on Build GRCh38
rs567225782 Chr.3: 23911034-23911034 on Build GRCh38
rs376168712 Chr.3: 23910802-23910802 on Build GRCh38
rs757867719 Chr.3: 23910949-23910949 on Build GRCh38
rs745443884 Chr.3: 23910854-23910854 on Build GRCh38
rs751945454 Chr.3: 23910864-23910864 on Build GRCh38
rs1004118129 Chr.3: 23910982-23910982 on Build GRCh38
rs747117489 Chr.3: 23910952-23910952 on Build GRCh38
rs751780259 Chr.3: 23910933-23910933 on Build GRCh38
rs536088992 Chr.3: 23911050-23911050 on Build GRCh38
rs778820036 Chr.3: 23910797-23910797 on Build GRCh38
rs764682109 Chr.3: 23910838-23910838 on Build GRCh38
rs980407846 Chr.3: 23910974-23910974 on Build GRCh38
rs764740071 Chr.3: 23910792-23910792 on Build GRCh38
rs750333129 Chr.3: 23910945-23910945 on Build GRCh38
rs186174646 Chr.3: 23910688-23910688 on Build GRCh38
rs1054454081 Chr.3: 23910699-23910699 on Build GRCh38
rs557587289 Chr.3: 23911075-23911075 on Build GRCh38
rs768961278 Chr.3: 23910886-23910886 on Build GRCh38
rs775821349 Chr.3: 23910765-23910765 on Build GRCh38
rs746625383 Chr.3: 23910953-23910953 on Build GRCh38
rs1029870293 Chr.3: 23910736-23910736 on Build GRCh38
rs768701293 Chr.3: 23910957-23910958 on Build GRCh38
rs746678249 Chr.3: 23910872-23910872 on Build GRCh38
rs755912281 Chr.3: 23910828-23910828 on Build GRCh38
rs927177653 Chr.3: 23910975-23910975 on Build GRCh38
rs867242968 Chr.3: 23911060-23911060 on Build GRCh38
rs12107726 Chr.3: 23910751-23910751 on Build GRCh38
rs890987201 Chr.3: 23910922-23910922 on Build GRCh38
rs778980508 Chr.3: 23910944-23910944 on Build GRCh38
rs1044208056 Chr.3: 23910672-23910672 on Build GRCh38
rs879390062 Chr.3: 23910957-23910957 on Build GRCh38
rs770389054 Chr.3: 23910905-23910905 on Build GRCh38
rs747152078 Chr.3: 23910868-23910874 on Build GRCh38
rs375843221 Chr.3: 23910887-23910887 on Build GRCh38
rs1022696658 Chr.3: 23910675-23910675 on Build GRCh38
rs755596082 Chr.3: 23910940-23910940 on Build GRCh38
rs1010188313 Chr.3: 23910718-23910718 on Build GRCh38
rs757227193 Chr.3: 23910782-23910782 on Build GRCh38
rs762405480 Chr.3: 23910795-23910795 on Build GRCh38
rs765199591 Chr.3: 23910800-23910800 on Build GRCh38
rs200361050 Chr.3: 23910951-23910951 on Build GRCh38
rs936110832 Chr.3: 23910698-23910698 on Build GRCh38

View Full Product Details