Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 28322245-28322744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZI2
Entrez Gene ID
64343
Gene Name
5-azacytidine induced 2
Gene Aliases
AZ2, NAP1, TILP
UniGene
Hs.706676
Gene Chromosome Location
Chr.3: 28322353-28349129 on Build GRCh38
Transcripts
NM_001271650.1 NM_022461.4 XM_005265392.2 XM_011534009.2 XM_011533402.2

Gene Symbol
CMC1
Entrez Gene ID
152100
Gene Name
C-X9-C motif containing 1
Gene Aliases
C3orf68
UniGene
Hs.444724
Gene Chromosome Location
Chr.3: 28241588-28322874 on Build GRCh38
Transcripts
NM_001271650.1 NM_022461.4 XM_005265392.2 XM_011534009.2 XM_011533402.2

SNP Detail

SNP ID Chromosome Location
rs1044304703 Chr.3: 28322574-28322574 on Build GRCh38
rs944415588 Chr.3: 28322502-28322502 on Build GRCh38
rs1027913562 Chr.3: 28322651-28322651 on Build GRCh38
rs975585049 Chr.3: 28322607-28322607 on Build GRCh38
rs886759069 Chr.3: 28322435-28322435 on Build GRCh38
rs778958911 Chr.3: 28322424-28322424 on Build GRCh38
rs745703378 Chr.3: 28322433-28322433 on Build GRCh38
rs1004164076 Chr.3: 28322452-28322452 on Build GRCh38
rs758005171 Chr.3: 28322690-28322690 on Build GRCh38
rs973673252 Chr.3: 28322428-28322432 on Build GRCh38
rs962817517 Chr.3: 28322415-28322415 on Build GRCh38
rs921593200 Chr.3: 28322680-28322680 on Build GRCh38
rs865935286 Chr.3: 28322425-28322425 on Build GRCh38
rs1012285959 Chr.3: 28322638-28322638 on Build GRCh38
rs958815095 Chr.3: 28322503-28322503 on Build GRCh38
rs898598354 Chr.3: 28322648-28322648 on Build GRCh38
rs926215122 Chr.3: 28322580-28322580 on Build GRCh38
rs933754270 Chr.3: 28322635-28322635 on Build GRCh38
rs935579578 Chr.3: 28322609-28322609 on Build GRCh38
rs140850573 Chr.3: 28322542-28322542 on Build GRCh38
rs936225935 Chr.3: 28322587-28322587 on Build GRCh38
rs896735608 Chr.3: 28322620-28322620 on Build GRCh38
rs575747519 Chr.3: 28322501-28322501 on Build GRCh38
rs1050820904 Chr.3: 28322636-28322636 on Build GRCh38
rs1033440748 Chr.3: 28322478-28322478 on Build GRCh38
rs369361773 Chr.3: 28322719-28322719 on Build GRCh38
rs373174926 Chr.3: 28322487-28322487 on Build GRCh38
rs930881483 Chr.3: 28322440-28322440 on Build GRCh38
rs749940339 Chr.3: 28322611-28322611 on Build GRCh38
rs999756897 Chr.3: 28322295-28322295 on Build GRCh38
rs936177853 Chr.3: 28322322-28322322 on Build GRCh38
rs921430599 Chr.3: 28322629-28322629 on Build GRCh38
rs577141541 Chr.3: 28322669-28322670 on Build GRCh38
rs898389305 Chr.3: 28322463-28322463 on Build GRCh38
rs920422164 Chr.3: 28322435-28322436 on Build GRCh38
rs921706235 Chr.3: 28322533-28322533 on Build GRCh38
rs151008778 Chr.3: 28322519-28322519 on Build GRCh38
rs995580711 Chr.3: 28322650-28322650 on Build GRCh38
rs185027308 Chr.3: 28322716-28322716 on Build GRCh38
rs926171762 Chr.3: 28322308-28322308 on Build GRCh38
rs142702589 Chr.3: 28322457-28322457 on Build GRCh38
rs778223983 Chr.3: 28322549-28322549 on Build GRCh38
rs1035033672 Chr.3: 28322306-28322306 on Build GRCh38
rs1002451084 Chr.3: 28322468-28322468 on Build GRCh38
rs899585314 Chr.3: 28322339-28322339 on Build GRCh38
rs538570570 Chr.3: 28322529-28322529 on Build GRCh38
rs376021676 Chr.3: 28322531-28322531 on Build GRCh38
rs1050789788 Chr.3: 28322434-28322434 on Build GRCh38
rs370365467 Chr.3: 28322652-28322652 on Build GRCh38
rs1023622102 Chr.3: 28322641-28322641 on Build GRCh38
rs933692300 Chr.3: 28322358-28322358 on Build GRCh38
rs978059207 Chr.3: 28322288-28322288 on Build GRCh38
rs1018852531 Chr.3: 28322660-28322660 on Build GRCh38
rs375212007 Chr.3: 28322717-28322717 on Build GRCh38
rs1039869192 Chr.3: 28322328-28322328 on Build GRCh38
rs960661984 Chr.3: 28322321-28322321 on Build GRCh38

View Full Product Details