Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51395770-51396274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF1
Entrez Gene ID
9730
Gene Name
DDB1 and CUL4 associated factor 1
Gene Aliases
VPRBP
UniGene
Hs.716623
Gene Chromosome Location
Chr.3: 51395867-51505620 on Build GRCh38
Transcripts
NM_001171904.1 NM_014703.2 XM_005276753.4 XM_005276755.4 XM_011534273.2 XM_011534274.2 XM_011534275.2 XM_011534276.2 XM_011534277.2 XM_017007546.1 XM_017007547.1 XM_017007548.1 XM_017007549.1 XM_017007550.1 XM_017007551.1 NM_013286.4

Gene Symbol
RBM15B
Entrez Gene ID
29890
Gene Name
RNA binding motif protein 15B
Gene Aliases
HUMAGCGB, OTT3
UniGene
Hs.118738
Gene Chromosome Location
Chr.3: 51391268-51397908 on Build GRCh38
Transcripts
NM_001171904.1 NM_014703.2 XM_005276753.4 XM_005276755.4 XM_011534273.2 XM_011534274.2 XM_011534275.2 XM_011534276.2 XM_011534277.2 XM_017007546.1 XM_017007547.1 XM_017007548.1 XM_017007549.1 XM_017007550.1 XM_017007551.1 NM_013286.4

SNP Detail

SNP ID Chromosome Location
rs146912969 Chr.3: 51395823-51395823 on Build GRCh38
rs781893971 Chr.3: 51395839-51395839 on Build GRCh38
rs782627643 Chr.3: 51396136-51396137 on Build GRCh38
rs1056756354 Chr.3: 51395972-51395972 on Build GRCh38
rs782542146 Chr.3: 51395846-51395846 on Build GRCh38
rs112066805 Chr.3: 51395903-51395904 on Build GRCh38
rs74619531 Chr.3: 51395912-51395912 on Build GRCh38
rs782635423 Chr.3: 51395833-51395833 on Build GRCh38
rs782146326 Chr.3: 51396063-51396063 on Build GRCh38
rs781883187 Chr.3: 51396105-51396106 on Build GRCh38
rs537892459 Chr.3: 51396168-51396168 on Build GRCh38
rs139012689 Chr.3: 51395830-51395830 on Build GRCh38
rs759638316 Chr.3: 51395952-51395953 on Build GRCh38
rs143050198 Chr.3: 51395885-51395885 on Build GRCh38
rs573489619 Chr.3: 51395825-51395825 on Build GRCh38
rs141711597 Chr.3: 51396120-51396120 on Build GRCh38
rs148536321 Chr.3: 51395948-51395948 on Build GRCh38
rs549159717 Chr.3: 51396119-51396119 on Build GRCh38
rs908091715 Chr.3: 51395954-51395954 on Build GRCh38
rs963783200 Chr.3: 51395910-51395910 on Build GRCh38
rs1000624451 Chr.3: 51396109-51396109 on Build GRCh38
rs903964534 Chr.3: 51396106-51396106 on Build GRCh38
rs562498903 Chr.3: 51395982-51395982 on Build GRCh38
rs564582866 Chr.3: 51396134-51396135 on Build GRCh38
rs1049326336 Chr.3: 51395803-51395803 on Build GRCh38
rs782371024 Chr.3: 51395971-51395971 on Build GRCh38
rs782037059 Chr.3: 51396059-51396059 on Build GRCh38
rs1052225777 Chr.3: 51396138-51396138 on Build GRCh38
rs1019699003 Chr.3: 51395900-51395900 on Build GRCh38
rs538620378 Chr.3: 51396239-51396239 on Build GRCh38
rs892343561 Chr.3: 51396149-51396149 on Build GRCh38
rs930777087 Chr.3: 51395936-51395936 on Build GRCh38
rs571227665 Chr.3: 51396233-51396233 on Build GRCh38
rs560627077 Chr.3: 51395895-51395896 on Build GRCh38
rs782188195 Chr.3: 51395876-51395876 on Build GRCh38
rs533179936 Chr.3: 51396019-51396019 on Build GRCh38
rs201828759 Chr.3: 51396185-51396185 on Build GRCh38
rs896471208 Chr.3: 51395998-51395998 on Build GRCh38
rs782580811 Chr.3: 51395977-51395978 on Build GRCh38
rs577715459 Chr.3: 51395800-51395800 on Build GRCh38
rs551227918 Chr.3: 51396032-51396032 on Build GRCh38
rs782520710 Chr.3: 51395828-51395828 on Build GRCh38
rs566744019 Chr.3: 51396067-51396067 on Build GRCh38
rs578105038 Chr.3: 51395799-51395799 on Build GRCh38
rs761061627 Chr.3: 51395871-51395871 on Build GRCh38
rs782270577 Chr.3: 51395923-51395923 on Build GRCh38
rs78691708 Chr.3: 51396069-51396069 on Build GRCh38
rs1000678216 Chr.3: 51395841-51395841 on Build GRCh38
rs1019560971 Chr.3: 51396195-51396195 on Build GRCh38
rs150568658 Chr.3: 51396197-51396197 on Build GRCh38
rs996547212 Chr.3: 51396206-51396206 on Build GRCh38
rs940644900 Chr.3: 51395821-51395821 on Build GRCh38
rs765569203 Chr.3: 51396243-51396243 on Build GRCh38
rs1045537409 Chr.3: 51396092-51396092 on Build GRCh38
rs781822807 Chr.3: 51395816-51395816 on Build GRCh38
rs149643764 Chr.3: 51395801-51395801 on Build GRCh38

View Full Product Details