Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53817644-53818153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHDH
Entrez Gene ID
55349
Gene Name
choline dehydrogenase
Gene Aliases
-
UniGene
Hs.126688
Gene Chromosome Location
Chr.3: 53813130-53846393 on Build GRCh38
Transcripts
NM_018397.4 XM_006713250.3 XM_006713251.3 XM_006713252.3 XM_011533938.2 XM_011533939.2 XM_017006797.1 XM_017006798.1 XM_017006799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3365275 c.1739A>C p.K580T Chr.3: 53817823-53817823 on Build GRCh38
5355326 c.1672G>A p.A558T Chr.3: 53817890-53817890 on Build GRCh38
4442966 c.1567G>A p.D523N Chr.3: 53817995-53817995 on Build GRCh38
6576371 c.1542C>T p.C514C Chr.3: 53818020-53818020 on Build GRCh38
4467224 c.1477C>T p.Q493* Chr.3: 53818085-53818085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778521519 Chr.3: 53817830-53817830 on Build GRCh38
rs1034282100 Chr.3: 53818111-53818111 on Build GRCh38
rs779249534 Chr.3: 53818126-53818126 on Build GRCh38
rs757045949 Chr.3: 53817872-53817872 on Build GRCh38
rs756315152 Chr.3: 53817957-53817957 on Build GRCh38
rs557223892 Chr.3: 53817745-53817745 on Build GRCh38
rs942961896 Chr.3: 53817688-53817688 on Build GRCh38
rs767326265 Chr.3: 53818109-53818109 on Build GRCh38
rs867311053 Chr.3: 53817904-53817904 on Build GRCh38
rs1039940023 Chr.3: 53818017-53818017 on Build GRCh38
rs760627085 Chr.3: 53818117-53818117 on Build GRCh38
rs746797465 Chr.3: 53817732-53817732 on Build GRCh38
rs761184371 Chr.3: 53817984-53817984 on Build GRCh38
rs764998360 Chr.3: 53817910-53817910 on Build GRCh38
rs768360390 Chr.3: 53818079-53818079 on Build GRCh38
rs149941082 Chr.3: 53817782-53817782 on Build GRCh38
rs368634626 Chr.3: 53817991-53817991 on Build GRCh38
rs752255950 Chr.3: 53817835-53817835 on Build GRCh38
rs750407233 Chr.3: 53818115-53818115 on Build GRCh38
rs1054224326 Chr.3: 53817724-53817724 on Build GRCh38
rs542133535 Chr.3: 53817927-53817927 on Build GRCh38
rs748561278 Chr.3: 53817974-53817974 on Build GRCh38
rs746030795 Chr.3: 53817882-53817882 on Build GRCh38
rs758357120 Chr.3: 53817843-53817843 on Build GRCh38
rs781455103 Chr.3: 53817879-53817879 on Build GRCh38
rs146890343 Chr.3: 53818122-53818122 on Build GRCh38
rs372931744 Chr.3: 53818038-53818038 on Build GRCh38
rs971762177 Chr.3: 53817836-53817836 on Build GRCh38
rs764094831 Chr.3: 53817935-53817935 on Build GRCh38
rs1024887464 Chr.3: 53817842-53817842 on Build GRCh38
rs772773441 Chr.3: 53817938-53817938 on Build GRCh38
rs755131701 Chr.3: 53818123-53818123 on Build GRCh38
rs773678383 Chr.3: 53818087-53818087 on Build GRCh38
rs752245757 Chr.3: 53817918-53817918 on Build GRCh38
rs200221651 Chr.3: 53818106-53818106 on Build GRCh38
rs769815478 Chr.3: 53817776-53817776 on Build GRCh38
rs912808207 Chr.3: 53817669-53817669 on Build GRCh38
rs746818727 Chr.3: 53817866-53817866 on Build GRCh38
rs142509609 Chr.3: 53817972-53817972 on Build GRCh38
rs200668252 Chr.3: 53817902-53817902 on Build GRCh38
rs772448470 Chr.3: 53818043-53818043 on Build GRCh38
rs764597199 Chr.3: 53817795-53817795 on Build GRCh38
rs201249706 Chr.3: 53818054-53818054 on Build GRCh38
rs750179776 Chr.3: 53818024-53818024 on Build GRCh38
rs1047190029 Chr.3: 53817722-53817722 on Build GRCh38
rs574370935 Chr.3: 53818055-53818055 on Build GRCh38
rs753478907 Chr.3: 53817814-53817814 on Build GRCh38
rs763464372 Chr.3: 53817996-53817996 on Build GRCh38
rs958039338 Chr.3: 53817925-53817925 on Build GRCh38
rs764063975 Chr.3: 53817722-53817723 on Build GRCh38
rs759630876 Chr.3: 53817774-53817774 on Build GRCh38
rs763411472 Chr.3: 53817790-53817790 on Build GRCh38
rs757088122 Chr.3: 53818064-53818064 on Build GRCh38
rs746364665 Chr.3: 53817791-53817791 on Build GRCh38
rs78878070 Chr.3: 53817847-53817847 on Build GRCh38
rs1001749256 Chr.3: 53818064-53818064 on Build GRCh38
rs200159976 Chr.3: 53817983-53817983 on Build GRCh38
rs774758953 Chr.3: 53817897-53817897 on Build GRCh38
rs749427817 Chr.3: 53817891-53817891 on Build GRCh38
rs535087835 Chr.3: 53817695-53817695 on Build GRCh38
rs888547015 Chr.3: 53817726-53817726 on Build GRCh38
rs773230904 Chr.3: 53817973-53817973 on Build GRCh38
rs34336629 Chr.3: 53818026-53818026 on Build GRCh38
rs762533547 Chr.3: 53817995-53817995 on Build GRCh38
rs777919420 Chr.3: 53817851-53817851 on Build GRCh38
rs941451657 Chr.3: 53818069-53818069 on Build GRCh38
rs754007948 Chr.3: 53818033-53818033 on Build GRCh38
rs766119723 Chr.3: 53818118-53818118 on Build GRCh38
rs772633198 Chr.3: 53817977-53817977 on Build GRCh38
rs755617427 Chr.3: 53817936-53817936 on Build GRCh38
rs370861018 Chr.3: 53817890-53817890 on Build GRCh38
rs753559560 Chr.3: 53817962-53817962 on Build GRCh38
rs894463933 Chr.3: 53817787-53817787 on Build GRCh38
rs748272859 Chr.3: 53817841-53817841 on Build GRCh38
rs568422934 Chr.3: 53817730-53817730 on Build GRCh38
rs754678677 Chr.3: 53817829-53817829 on Build GRCh38
rs147713779 Chr.3: 53817699-53817699 on Build GRCh38
rs751678442 Chr.3: 53818037-53818037 on Build GRCh38
rs752228175 Chr.3: 53817821-53817821 on Build GRCh38
rs778414710 Chr.3: 53818052-53818052 on Build GRCh38
rs764981814 Chr.3: 53817990-53817990 on Build GRCh38
rs930144488 Chr.3: 53817718-53817718 on Build GRCh38
rs759040078 Chr.3: 53817908-53817908 on Build GRCh38
rs146668805 Chr.3: 53817963-53817963 on Build GRCh38
rs780809306 Chr.3: 53817939-53817939 on Build GRCh38
rs758769734 Chr.3: 53818023-53818023 on Build GRCh38
rs768771121 Chr.3: 53817966-53817966 on Build GRCh38
rs1017919562 Chr.3: 53817954-53817954 on Build GRCh38
rs111450925 Chr.3: 53817773-53817773 on Build GRCh38
rs751114246 Chr.3: 53817999-53817999 on Build GRCh38
rs773652681 Chr.3: 53817907-53817907 on Build GRCh38
rs951427789 Chr.3: 53817953-53817953 on Build GRCh38
rs577083114 Chr.3: 53817796-53817796 on Build GRCh38
rs759548864 Chr.3: 53817698-53817698 on Build GRCh38
rs201355737 Chr.3: 53817739-53817739 on Build GRCh38
rs200022373 Chr.3: 53818063-53818063 on Build GRCh38
rs1055888120 Chr.3: 53818088-53818088 on Build GRCh38
rs777557327 Chr.3: 53817956-53817956 on Build GRCh38
rs1055835048 Chr.3: 53817775-53817775 on Build GRCh38
rs762655955 Chr.3: 53818071-53818071 on Build GRCh38
rs758050811 Chr.3: 53817933-53817933 on Build GRCh38
rs143449485 Chr.3: 53817883-53817883 on Build GRCh38
rs766023057 Chr.3: 53817798-53817798 on Build GRCh38
rs755221735 Chr.3: 53818036-53818036 on Build GRCh38
rs767722910 Chr.3: 53817903-53817903 on Build GRCh38

View Full Product Details