Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 125225266-125225802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF148
Entrez Gene ID
7707
Gene Name
zinc finger protein 148
Gene Aliases
BERF-1, BFCOL1, HT-BETA, ZBP-89, ZFP148, pHZ-52
UniGene
Hs.592591
Gene Chromosome Location
Chr.3: 125225669-125375354 on Build GRCh38
Transcripts
NM_021964.2

SNP Detail

SNP ID Chromosome Location
rs145603585 Chr.3: 125225321-125225321 on Build GRCh38
rs143627237 Chr.3: 125225448-125225448 on Build GRCh38
rs117786707 Chr.3: 125225595-125225595 on Build GRCh38
rs1005542857 Chr.3: 125225719-125225719 on Build GRCh38
rs572888205 Chr.3: 125225675-125225675 on Build GRCh38
rs748238907 Chr.3: 125225691-125225692 on Build GRCh38
rs1011209442 Chr.3: 125225607-125225607 on Build GRCh38
rs139388408 Chr.3: 125225365-125225365 on Build GRCh38
rs1037337097 Chr.3: 125225429-125225429 on Build GRCh38
rs992623899 Chr.3: 125225735-125225735 on Build GRCh38
rs566142144 Chr.3: 125225386-125225386 on Build GRCh38
rs773821516 Chr.3: 125225483-125225483 on Build GRCh38
rs927104029 Chr.3: 125225348-125225348 on Build GRCh38
rs936078 Chr.3: 125225334-125225334 on Build GRCh38
rs892560363 Chr.3: 125225574-125225574 on Build GRCh38
rs879219957 Chr.3: 125225563-125225564 on Build GRCh38
rs73859149 Chr.3: 125225573-125225573 on Build GRCh38
rs1038773936 Chr.3: 125225630-125225630 on Build GRCh38
rs369317368 Chr.3: 125225640-125225640 on Build GRCh38
rs559121574 Chr.3: 125225605-125225605 on Build GRCh38
rs951013417 Chr.3: 125225522-125225522 on Build GRCh38
rs942167357 Chr.3: 125225564-125225564 on Build GRCh38
rs568278663 Chr.3: 125225377-125225377 on Build GRCh38
rs907597370 Chr.3: 125225749-125225749 on Build GRCh38
rs959931521 Chr.3: 125225355-125225355 on Build GRCh38
rs936079 Chr.3: 125225299-125225299 on Build GRCh38
rs550065841 Chr.3: 125225336-125225336 on Build GRCh38
rs7430070 Chr.3: 125225337-125225337 on Build GRCh38
rs991313770 Chr.3: 125225565-125225565 on Build GRCh38
rs557921987 Chr.3: 125225437-125225437 on Build GRCh38
rs1045573324 Chr.3: 125225574-125225574 on Build GRCh38
rs544824373 Chr.3: 125225769-125225769 on Build GRCh38
rs960122040 Chr.3: 125225654-125225654 on Build GRCh38
rs146784542 Chr.3: 125225579-125225579 on Build GRCh38
rs535471565 Chr.3: 125225361-125225361 on Build GRCh38
rs907649518 Chr.3: 125225371-125225371 on Build GRCh38
rs983499239 Chr.3: 125225547-125225547 on Build GRCh38
rs760228601 Chr.3: 125225598-125225598 on Build GRCh38
rs1030441896 Chr.3: 125225695-125225695 on Build GRCh38
rs577174674 Chr.3: 125225504-125225504 on Build GRCh38
rs1050728463 Chr.3: 125225482-125225482 on Build GRCh38
rs545865861 Chr.3: 125225572-125225572 on Build GRCh38
rs576027776 Chr.3: 125225389-125225389 on Build GRCh38
rs1031741019 Chr.3: 125225354-125225354 on Build GRCh38
rs951028620 Chr.3: 125225710-125225710 on Build GRCh38
rs976194343 Chr.3: 125225511-125225511 on Build GRCh38
rs1030557970 Chr.3: 125225516-125225516 on Build GRCh38

View Full Product Details