Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114679403-114679929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD1
Entrez Gene ID
270
Gene Name
adenosine monophosphate deaminase 1
Gene Aliases
MAD, MADA, MMDD
UniGene
Hs.89570
Gene Chromosome Location
Chr.1: 114673098-114695618 on Build GRCh38
Transcripts
NM_000036.2 NM_001172626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4899674 c.878C>T p.T293I Chr.1: 114679697-114679697 on Build GRCh38
4899675 c.779C>T p.T260I Chr.1: 114679697-114679697 on Build GRCh38
3369216 c.874T>C p.Y292H Chr.1: 114679701-114679701 on Build GRCh38
3369217 c.775T>C p.Y259H Chr.1: 114679701-114679701 on Build GRCh38
5960902 c.843C>T p.D281D Chr.1: 114679633-114679633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184999114 Chr.1: 114679489-114679489 on Build GRCh38
rs776813209 Chr.1: 114679553-114679553 on Build GRCh38
rs767813958 Chr.1: 114679756-114679756 on Build GRCh38
rs772361373 Chr.1: 114679538-114679538 on Build GRCh38
rs750114853 Chr.1: 114679707-114679707 on Build GRCh38
rs765931733 Chr.1: 114679711-114679711 on Build GRCh38
rs765247541 Chr.1: 114679579-114679579 on Build GRCh38
rs142477997 Chr.1: 114679599-114679599 on Build GRCh38
rs536037850 Chr.1: 114679675-114679675 on Build GRCh38
rs34526199 Chr.1: 114679616-114679616 on Build GRCh38
rs952520164 Chr.1: 114679560-114679560 on Build GRCh38
rs897826696 Chr.1: 114679769-114679769 on Build GRCh38
rs1054090898 Chr.1: 114679544-114679544 on Build GRCh38
rs779360535 Chr.1: 114679623-114679623 on Build GRCh38
rs748489138 Chr.1: 114679535-114679535 on Build GRCh38
rs543595554 Chr.1: 114679883-114679883 on Build GRCh38
rs771012215 Chr.1: 114679651-114679651 on Build GRCh38
rs935730402 Chr.1: 114679902-114679902 on Build GRCh38
rs377158588 Chr.1: 114679728-114679728 on Build GRCh38
rs189111132 Chr.1: 114679719-114679719 on Build GRCh38
rs771190375 Chr.1: 114679549-114679549 on Build GRCh38
rs547330091 Chr.1: 114679690-114679690 on Build GRCh38
rs973518689 Chr.1: 114679513-114679513 on Build GRCh38
rs927006880 Chr.1: 114679504-114679504 on Build GRCh38
rs890738273 Chr.1: 114679428-114679428 on Build GRCh38
rs139003085 Chr.1: 114679689-114679689 on Build GRCh38
rs145030449 Chr.1: 114679633-114679633 on Build GRCh38
rs115883987 Chr.1: 114679548-114679548 on Build GRCh38
rs780926201 Chr.1: 114679608-114679608 on Build GRCh38
rs1009163234 Chr.1: 114679723-114679723 on Build GRCh38
rs373551240 Chr.1: 114679746-114679746 on Build GRCh38
rs773478760 Chr.1: 114679542-114679542 on Build GRCh38
rs538687919 Chr.1: 114679609-114679609 on Build GRCh38
rs143235944 Chr.1: 114679602-114679602 on Build GRCh38
rs762806667 Chr.1: 114679591-114679591 on Build GRCh38
rs201724664 Chr.1: 114679466-114679466 on Build GRCh38
rs767132701 Chr.1: 114679704-114679704 on Build GRCh38
rs758792942 Chr.1: 114679736-114679736 on Build GRCh38
rs544323538 Chr.1: 114679523-114679523 on Build GRCh38
rs775559099 Chr.1: 114679688-114679688 on Build GRCh38
rs743041 Chr.1: 114679857-114679857 on Build GRCh38
rs367913980 Chr.1: 114679618-114679618 on Build GRCh38
rs531266075 Chr.1: 114679522-114679522 on Build GRCh38
rs751351697 Chr.1: 114679590-114679591 on Build GRCh38
rs577176058 Chr.1: 114679791-114679791 on Build GRCh38
rs554380360 Chr.1: 114679694-114679694 on Build GRCh38
rs752725976 Chr.1: 114679586-114679586 on Build GRCh38
rs737549 Chr.1: 114679486-114679486 on Build GRCh38
rs755615828 Chr.1: 114679710-114679710 on Build GRCh38
rs558478873 Chr.1: 114679781-114679781 on Build GRCh38
rs753014173 Chr.1: 114679761-114679762 on Build GRCh38
rs759660724 Chr.1: 114679665-114679665 on Build GRCh38
rs1010153766 Chr.1: 114679554-114679554 on Build GRCh38
rs764929472 Chr.1: 114679861-114679861 on Build GRCh38
rs917405654 Chr.1: 114679475-114679475 on Build GRCh38
rs949350359 Chr.1: 114679607-114679607 on Build GRCh38
rs371819369 Chr.1: 114679659-114679659 on Build GRCh38
rs1009293132 Chr.1: 114679478-114679478 on Build GRCh38
rs748720444 Chr.1: 114679632-114679632 on Build GRCh38
rs949421 Chr.1: 114679669-114679669 on Build GRCh38
rs990632988 Chr.1: 114679899-114679899 on Build GRCh38
rs571309576 Chr.1: 114679571-114679571 on Build GRCh38
rs886044350 Chr.1: 114679683-114679683 on Build GRCh38
rs764094155 Chr.1: 114679691-114679691 on Build GRCh38
rs575924901 Chr.1: 114679695-114679695 on Build GRCh38
rs971608320 Chr.1: 114679801-114679801 on Build GRCh38
rs192799036 Chr.1: 114679850-114679850 on Build GRCh38
rs777934972 Chr.1: 114679641-114679641 on Build GRCh38
rs761755 Chr.1: 114679813-114679813 on Build GRCh38
rs192753656 Chr.1: 114679445-114679445 on Build GRCh38
rs935656856 Chr.1: 114679537-114679537 on Build GRCh38
rs754882583 Chr.1: 114679669-114679671 on Build GRCh38
rs768332999 Chr.1: 114679692-114679692 on Build GRCh38
rs984064630 Chr.1: 114679437-114679437 on Build GRCh38
rs565839639 Chr.1: 114679642-114679642 on Build GRCh38
rs982156348 Chr.1: 114679612-114679612 on Build GRCh38
rs763848203 Chr.1: 114679595-114679595 on Build GRCh38

View Full Product Details