Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169094973-169095499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
UniGene
Hs.744090
Gene Chromosome Location
Chr.3: 169083499-169663781 on Build GRCh38
Transcripts
NM_001105077.3 NM_001105078.3 NM_001163999.1 NM_001164000.1 NM_001205194.1 NM_004991.3 NM_005241.3 XM_005247213.3 XM_005247214.3 XM_005247215.3 XM_005247219.2 XM_005247220.2 XM_005247221.2 XM_005247223.2 XM_005247224.3 XM_005247225.3 XM_005247226.3 XM_011512546.2 XM_017005874.1 XM_017005875.1 XM_017005876.1 XM_017005877.1 XM_017005878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
26577 c.2286-1G>A p.? Chr.3: 169095246-169095246 on Build GRCh38
1693769 c.2581G>A p.D861N Chr.3: 169095145-169095145 on Build GRCh38
5598488 c.2501C>T p.S834L Chr.3: 169095225-169095225 on Build GRCh38
1327736 c.2411A>G p.N804S Chr.3: 169095120-169095120 on Build GRCh38
6097029 c.2979T>A p.N993K Chr.3: 169095089-169095089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886662366 Chr.3: 169095035-169095035 on Build GRCh38
rs745390955 Chr.3: 169095280-169095280 on Build GRCh38
rs201205882 Chr.3: 169095158-169095158 on Build GRCh38
rs1012247480 Chr.3: 169095347-169095347 on Build GRCh38
rs780185513 Chr.3: 169095042-169095042 on Build GRCh38
rs920646089 Chr.3: 169095409-169095409 on Build GRCh38
rs777622718 Chr.3: 169095245-169095245 on Build GRCh38
rs199804681 Chr.3: 169095194-169095194 on Build GRCh38
rs747209504 Chr.3: 169095209-169095209 on Build GRCh38
rs182585574 Chr.3: 169095031-169095031 on Build GRCh38
rs749244302 Chr.3: 169095255-169095255 on Build GRCh38
rs61741716 Chr.3: 169095108-169095108 on Build GRCh38
rs371216345 Chr.3: 169095048-169095048 on Build GRCh38
rs369936185 Chr.3: 169095072-169095072 on Build GRCh38
rs771453465 Chr.3: 169095019-169095020 on Build GRCh38
rs754369058 Chr.3: 169095140-169095140 on Build GRCh38
rs1005317166 Chr.3: 169095315-169095315 on Build GRCh38
rs572891975 Chr.3: 169095385-169095385 on Build GRCh38
rs770417491 Chr.3: 169095066-169095066 on Build GRCh38
rs896582395 Chr.3: 169095344-169095344 on Build GRCh38
rs930659345 Chr.3: 169095410-169095410 on Build GRCh38
rs116736168 Chr.3: 169095083-169095083 on Build GRCh38
rs916393295 Chr.3: 169095332-169095332 on Build GRCh38
rs184545299 Chr.3: 169095349-169095349 on Build GRCh38
rs772293161 Chr.3: 169095034-169095034 on Build GRCh38
rs775317919 Chr.3: 169095284-169095284 on Build GRCh38
rs577645742 Chr.3: 169095306-169095306 on Build GRCh38
rs779045869 Chr.3: 169095032-169095032 on Build GRCh38
rs780278469 Chr.3: 169095185-169095185 on Build GRCh38
rs760399228 Chr.3: 169095289-169095289 on Build GRCh38
rs758434724 Chr.3: 169095164-169095164 on Build GRCh38
rs773077050 Chr.3: 169095056-169095056 on Build GRCh38
rs770916724 Chr.3: 169095274-169095274 on Build GRCh38
rs369652575 Chr.3: 169095283-169095283 on Build GRCh38
rs150253361 Chr.3: 169095020-169095020 on Build GRCh38
rs781490872 Chr.3: 169095295-169095295 on Build GRCh38
rs374713014 Chr.3: 169095053-169095053 on Build GRCh38
rs879115686 Chr.3: 169095022-169095022 on Build GRCh38
rs375683377 Chr.3: 169095143-169095143 on Build GRCh38
rs759048415 Chr.3: 169095073-169095073 on Build GRCh38
rs200552630 Chr.3: 169095178-169095178 on Build GRCh38
rs1034954143 Chr.3: 169095334-169095334 on Build GRCh38
rs559259065 Chr.3: 169095372-169095372 on Build GRCh38
rs200049869 Chr.3: 169095082-169095082 on Build GRCh38
rs769620459 Chr.3: 169095227-169095227 on Build GRCh38
rs774113812 Chr.3: 169095279-169095279 on Build GRCh38
rs563898299 Chr.3: 169095393-169095393 on Build GRCh38
rs141176563 Chr.3: 169095077-169095077 on Build GRCh38
rs762920930 Chr.3: 169095065-169095065 on Build GRCh38
rs1009085628 Chr.3: 169095041-169095041 on Build GRCh38
rs34224062 Chr.3: 169095154-169095154 on Build GRCh38
rs992035402 Chr.3: 169095301-169095301 on Build GRCh38
rs948845829 Chr.3: 169095371-169095371 on Build GRCh38
rs767306816 Chr.3: 169095076-169095076 on Build GRCh38

View Full Product Details