Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186046708-186047222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV5
Entrez Gene ID
2119
Gene Name
ETS variant 5
Gene Aliases
ERM
UniGene
Hs.43697
Gene Chromosome Location
Chr.3: 186046317-186109112 on Build GRCh38
Transcripts
NM_004454.2

SNP Detail

SNP ID Chromosome Location
rs999613500 Chr.3: 186046944-186046944 on Build GRCh38
rs777647675 Chr.3: 186046961-186046961 on Build GRCh38
rs540652113 Chr.3: 186046814-186046814 on Build GRCh38
rs373330948 Chr.3: 186046945-186046945 on Build GRCh38
rs564619897 Chr.3: 186046783-186046783 on Build GRCh38
rs746799982 Chr.3: 186046977-186046977 on Build GRCh38
rs1022003708 Chr.3: 186046858-186046858 on Build GRCh38
rs918722397 Chr.3: 186047018-186047018 on Build GRCh38
rs573241800 Chr.3: 186046809-186046809 on Build GRCh38
rs946102079 Chr.3: 186047121-186047124 on Build GRCh38
rs1001101333 Chr.3: 186047165-186047165 on Build GRCh38
rs1000750775 Chr.3: 186046831-186046831 on Build GRCh38
rs561934372 Chr.3: 186046887-186046887 on Build GRCh38
rs192057120 Chr.3: 186047130-186047130 on Build GRCh38
rs776696023 Chr.3: 186047174-186047174 on Build GRCh38
rs533361357 Chr.3: 186047091-186047091 on Build GRCh38
rs111652985 Chr.3: 186046780-186046780 on Build GRCh38
rs556850976 Chr.3: 186047092-186047092 on Build GRCh38
rs566598671 Chr.3: 186047197-186047197 on Build GRCh38
rs1028543768 Chr.3: 186046994-186046994 on Build GRCh38
rs959063529 Chr.3: 186046851-186046851 on Build GRCh38
rs1018643631 Chr.3: 186046959-186046959 on Build GRCh38
rs772088300 Chr.3: 186046986-186046986 on Build GRCh38
rs902437013 Chr.3: 186046820-186046820 on Build GRCh38
rs1010661334 Chr.3: 186046847-186046847 on Build GRCh38
rs1040999397 Chr.3: 186047064-186047064 on Build GRCh38
rs909928054 Chr.3: 186046741-186046741 on Build GRCh38
rs942695572 Chr.3: 186046738-186046738 on Build GRCh38
rs973209773 Chr.3: 186047145-186047145 on Build GRCh38
rs746724616 Chr.3: 186046739-186046739 on Build GRCh38
rs970443407 Chr.3: 186046937-186046937 on Build GRCh38
rs1037005631 Chr.3: 186046781-186046781 on Build GRCh38
rs1032671209 Chr.3: 186046833-186046833 on Build GRCh38
rs546500009 Chr.3: 186046896-186046896 on Build GRCh38
rs138737120 Chr.3: 186047049-186047049 on Build GRCh38
rs911450201 Chr.3: 186047056-186047056 on Build GRCh38
rs895017313 Chr.3: 186046823-186046823 on Build GRCh38
rs1024755316 Chr.3: 186046932-186046932 on Build GRCh38
rs772151795 Chr.3: 186046795-186046795 on Build GRCh38
rs902238506 Chr.3: 186046877-186046877 on Build GRCh38
rs9831679 Chr.3: 186047058-186047058 on Build GRCh38
rs939348153 Chr.3: 186047171-186047171 on Build GRCh38
rs909969294 Chr.3: 186047065-186047065 on Build GRCh38
rs928026355 Chr.3: 186047146-186047146 on Build GRCh38
rs1054975302 Chr.3: 186047184-186047184 on Build GRCh38
rs1032291260 Chr.3: 186047169-186047169 on Build GRCh38

View Full Product Details