Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154744497-154744994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRAT
Entrez Gene ID
9227
Gene Name
lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase)
Gene Aliases
LCA14
UniGene
Hs.658427
Gene Chromosome Location
Chr.4: 154740841-154753119 on Build GRCh38
Transcripts
NM_001301645.1 NM_004744.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4122970 c.259C>T p.R87C Chr.4: 154744585-154744585 on Build GRCh38
39674 c.441G>C p.K147N Chr.4: 154744767-154744767 on Build GRCh38
5821387 c.461A>G p.Y154C Chr.4: 154744787-154744787 on Build GRCh38
4122973 c.428G>A p.R143Q Chr.4: 154744754-154744754 on Build GRCh38
4122974 c.487C>T p.H163Y Chr.4: 154744813-154744813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765366741 Chr.4: 154744680-154744680 on Build GRCh38
rs201300725 Chr.4: 154744734-154744734 on Build GRCh38
rs915089130 Chr.4: 154744818-154744818 on Build GRCh38
rs959565413 Chr.4: 154744889-154744889 on Build GRCh38
rs202066076 Chr.4: 154744802-154744802 on Build GRCh38
rs766279892 Chr.4: 154744550-154744550 on Build GRCh38
rs751538809 Chr.4: 154744556-154744556 on Build GRCh38
rs551548180 Chr.4: 154744666-154744666 on Build GRCh38
rs777454624 Chr.4: 154744706-154744706 on Build GRCh38
rs1035206645 Chr.4: 154744624-154744624 on Build GRCh38
rs775488474 Chr.4: 154744906-154744906 on Build GRCh38
rs757305596 Chr.4: 154744547-154744547 on Build GRCh38
rs760890088 Chr.4: 154744538-154744538 on Build GRCh38
rs777314071 Chr.4: 154744597-154744597 on Build GRCh38
rs761717462 Chr.4: 154744723-154744724 on Build GRCh38
rs754762928 Chr.4: 154744557-154744557 on Build GRCh38
rs764224506 Chr.4: 154744539-154744539 on Build GRCh38
rs758030004 Chr.4: 154744826-154744826 on Build GRCh38
rs768065631 Chr.4: 154744524-154744524 on Build GRCh38
rs769051542 Chr.4: 154744846-154744846 on Build GRCh38
rs770608706 Chr.4: 154744626-154744626 on Build GRCh38
rs878853351 Chr.4: 154744799-154744799 on Build GRCh38
rs750516743 Chr.4: 154744681-154744681 on Build GRCh38
rs555728501 Chr.4: 154744689-154744689 on Build GRCh38
rs1029329083 Chr.4: 154744710-154744710 on Build GRCh38
rs780908255 Chr.4: 154744562-154744562 on Build GRCh38
rs770276396 Chr.4: 154744606-154744606 on Build GRCh38
rs770985855 Chr.4: 154744884-154744884 on Build GRCh38
rs1010347467 Chr.4: 154744813-154744813 on Build GRCh38
rs760517479 Chr.4: 154744796-154744796 on Build GRCh38
rs780578479 Chr.4: 154744830-154744830 on Build GRCh38
rs145215771 Chr.4: 154744860-154744860 on Build GRCh38
rs944643002 Chr.4: 154744711-154744711 on Build GRCh38
rs775854426 Chr.4: 154744664-154744664 on Build GRCh38
rs17031981 Chr.4: 154744668-154744668 on Build GRCh38
rs577066380 Chr.4: 154744894-154744894 on Build GRCh38
rs565728053 Chr.4: 154744579-154744579 on Build GRCh38
rs138750814 Chr.4: 154744928-154744928 on Build GRCh38
rs786205644 Chr.4: 154744559-154744568 on Build GRCh38
rs112943981 Chr.4: 154744561-154744561 on Build GRCh38
rs768389044 Chr.4: 154744584-154744584 on Build GRCh38
rs752489964 Chr.4: 154744696-154744696 on Build GRCh38
rs767475890 Chr.4: 154744693-154744693 on Build GRCh38
rs760475456 Chr.4: 154744907-154744907 on Build GRCh38
rs907805484 Chr.4: 154744690-154744690 on Build GRCh38
rs747900628 Chr.4: 154744570-154744570 on Build GRCh38
rs752620052 Chr.4: 154744791-154744791 on Build GRCh38
rs753677274 Chr.4: 154744803-154744803 on Build GRCh38
rs375365480 Chr.4: 154744845-154744845 on Build GRCh38
rs572457198 Chr.4: 154744737-154744737 on Build GRCh38
rs147855559 Chr.4: 154744724-154744724 on Build GRCh38
rs372344739 Chr.4: 154744740-154744740 on Build GRCh38
rs543629736 Chr.4: 154744938-154744938 on Build GRCh38
rs369482649 Chr.4: 154744908-154744908 on Build GRCh38
rs895443586 Chr.4: 154744542-154744542 on Build GRCh38
rs774998317 Chr.4: 154744640-154744640 on Build GRCh38
rs372762225 Chr.4: 154744878-154744878 on Build GRCh38
rs537325942 Chr.4: 154744685-154744685 on Build GRCh38
rs983456632 Chr.4: 154744684-154744684 on Build GRCh38
rs761849550 Chr.4: 154744671-154744671 on Build GRCh38
rs200607271 Chr.4: 154744812-154744812 on Build GRCh38
rs104893848 Chr.4: 154744851-154744851 on Build GRCh38
rs770007716 Chr.4: 154744775-154744775 on Build GRCh38
rs11575980 Chr.4: 154744677-154744677 on Build GRCh38
rs749900934 Chr.4: 154744823-154744823 on Build GRCh38
rs374704716 Chr.4: 154744809-154744809 on Build GRCh38
rs764092378 Chr.4: 154744910-154744910 on Build GRCh38
rs773237349 Chr.4: 154744783-154744783 on Build GRCh38
rs532889931 Chr.4: 154744645-154744645 on Build GRCh38
rs753911609 Chr.4: 154744546-154744546 on Build GRCh38
rs748964060 Chr.4: 154744717-154744717 on Build GRCh38
rs750287062 Chr.4: 154744918-154744919 on Build GRCh38
rs879058165 Chr.4: 154744758-154744758 on Build GRCh38
rs745372700 Chr.4: 154744727-154744727 on Build GRCh38
rs776906234 Chr.4: 154744753-154744753 on Build GRCh38
rs920685564 Chr.4: 154744642-154744642 on Build GRCh38
rs748544531 Chr.4: 154744852-154744852 on Build GRCh38
rs139819099 Chr.4: 154744729-154744729 on Build GRCh38
rs145355018 Chr.4: 154744843-154744843 on Build GRCh38
rs773474987 Chr.4: 154744876-154744876 on Build GRCh38
rs746431917 Chr.4: 154744641-154744641 on Build GRCh38
rs762080267 Chr.4: 154744770-154744770 on Build GRCh38
rs765063151 Chr.4: 154744780-154744781 on Build GRCh38
rs558725765 Chr.4: 154744620-154744620 on Build GRCh38
rs748811771 Chr.4: 154744600-154744600 on Build GRCh38
rs775838916 Chr.4: 154744531-154744531 on Build GRCh38
rs765003341 Chr.4: 154744735-154744735 on Build GRCh38
rs755913771 Chr.4: 154744704-154744704 on Build GRCh38
rs778504867 Chr.4: 154744725-154744725 on Build GRCh38

View Full Product Details