Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65569490-65570007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPWD1
Entrez Gene ID
23398
Gene Name
peptidylprolyl isomerase domain and WD repeat containing 1
Gene Aliases
-
UniGene
Hs.121432
Gene Chromosome Location
Chr.5: 65563236-65587549 on Build GRCh38
Transcripts
NM_001278926.1 NM_001278927.1 NM_001278929.1 NM_015342.3 XM_005248474.3 XM_006714583.3 XM_011543295.1 XM_017009286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
449965 c.389G>A p.R130H Chr.5: 65569721-65569721 on Build GRCh38
7146490 c.345C>T p.F115F Chr.5: 65569677-65569677 on Build GRCh38
1695837 c.337G>A p.V113I Chr.5: 65569669-65569669 on Build GRCh38
5868694 c.392G>A p.S131N Chr.5: 65569724-65569724 on Build GRCh38
6582121 c.401-7G>C p.? Chr.5: 65569871-65569871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3752673 Chr.5: 65569831-65569831 on Build GRCh38
rs755110359 Chr.5: 65569780-65569780 on Build GRCh38
rs377652689 Chr.5: 65569742-65569742 on Build GRCh38
rs755604351 Chr.5: 65569935-65569935 on Build GRCh38
rs777881593 Chr.5: 65569963-65569963 on Build GRCh38
rs756648807 Chr.5: 65569773-65569773 on Build GRCh38
rs768197268 Chr.5: 65569883-65569883 on Build GRCh38
rs918616167 Chr.5: 65569949-65569949 on Build GRCh38
rs373488081 Chr.5: 65569682-65569682 on Build GRCh38
rs149208 Chr.5: 65569578-65569578 on Build GRCh38
rs138014576 Chr.5: 65569775-65569775 on Build GRCh38
rs914484020 Chr.5: 65569518-65569519 on Build GRCh38
rs770759063 Chr.5: 65569621-65569621 on Build GRCh38
rs1049073144 Chr.5: 65569976-65569976 on Build GRCh38
rs370637902 Chr.5: 65569616-65569616 on Build GRCh38
rs915663824 Chr.5: 65569946-65569946 on Build GRCh38
rs781509397 Chr.5: 65569917-65569921 on Build GRCh38
rs13167170 Chr.5: 65569983-65569983 on Build GRCh38
rs759451714 Chr.5: 65569658-65569658 on Build GRCh38
rs149511641 Chr.5: 65569903-65569903 on Build GRCh38
rs149512 Chr.5: 65569519-65569519 on Build GRCh38
rs945727431 Chr.5: 65569519-65569519 on Build GRCh38
rs375281263 Chr.5: 65569588-65569588 on Build GRCh38
rs768946248 Chr.5: 65569610-65569610 on Build GRCh38
rs745695495 Chr.5: 65569848-65569848 on Build GRCh38
rs767105688 Chr.5: 65569890-65569890 on Build GRCh38
rs989228776 Chr.5: 65569517-65569517 on Build GRCh38
rs773780732 Chr.5: 65569884-65569884 on Build GRCh38
rs918697995 Chr.5: 65569584-65569584 on Build GRCh38
rs142403801 Chr.5: 65569660-65569660 on Build GRCh38
rs1022052269 Chr.5: 65569799-65569799 on Build GRCh38
rs748264149 Chr.5: 65569961-65569963 on Build GRCh38
rs757655272 Chr.5: 65569586-65569586 on Build GRCh38
rs778695962 Chr.5: 65569974-65569974 on Build GRCh38
rs761129597 Chr.5: 65569922-65569922 on Build GRCh38
rs111987815 Chr.5: 65569740-65569740 on Build GRCh38
rs1010696793 Chr.5: 65569776-65569776 on Build GRCh38
rs965437060 Chr.5: 65569670-65569670 on Build GRCh38
rs769671085 Chr.5: 65569855-65569855 on Build GRCh38
rs985928553 Chr.5: 65569825-65569825 on Build GRCh38
rs1055485907 Chr.5: 65569569-65569569 on Build GRCh38
rs150328237 Chr.5: 65569528-65569528 on Build GRCh38
rs1045296695 Chr.5: 65569636-65569636 on Build GRCh38
rs1021532694 Chr.5: 65569803-65569803 on Build GRCh38
rs756917074 Chr.5: 65569737-65569737 on Build GRCh38
rs778929335 Chr.5: 65569781-65569781 on Build GRCh38
rs13167526 Chr.5: 65569943-65569943 on Build GRCh38
rs780250369 Chr.5: 65569593-65569593 on Build GRCh38
rs944062002 Chr.5: 65569569-65569569 on Build GRCh38
rs953195846 Chr.5: 65569814-65569814 on Build GRCh38
rs774941741 Chr.5: 65569677-65569677 on Build GRCh38
rs199602684 Chr.5: 65569634-65569634 on Build GRCh38
rs763680805 Chr.5: 65569684-65569684 on Build GRCh38
rs766924542 Chr.5: 65569929-65569929 on Build GRCh38
rs754389800 Chr.5: 65569932-65569932 on Build GRCh38
rs763955094 Chr.5: 65569720-65569720 on Build GRCh38
rs144631160 Chr.5: 65569571-65569571 on Build GRCh38
rs547001816 Chr.5: 65569518-65569518 on Build GRCh38
rs748390907 Chr.5: 65569827-65569827 on Build GRCh38
rs375586974 Chr.5: 65569830-65569830 on Build GRCh38
rs573272686 Chr.5: 65569958-65569958 on Build GRCh38
rs200283546 Chr.5: 65569590-65569590 on Build GRCh38
rs758610685 Chr.5: 65569961-65569961 on Build GRCh38
rs938457384 Chr.5: 65569536-65569536 on Build GRCh38
rs377239494 Chr.5: 65569601-65569601 on Build GRCh38
rs761494524 Chr.5: 65569888-65569888 on Build GRCh38
rs986087586 Chr.5: 65569520-65569520 on Build GRCh38
rs996912801 Chr.5: 65569721-65569721 on Build GRCh38
rs769583440 Chr.5: 65569977-65569977 on Build GRCh38
rs775422373 Chr.5: 65569857-65569857 on Build GRCh38
rs750793354 Chr.5: 65569757-65569757 on Build GRCh38
rs762383588 Chr.5: 65569863-65569863 on Build GRCh38
rs1003227009 Chr.5: 65569712-65569712 on Build GRCh38
rs755275817 Chr.5: 65569778-65569778 on Build GRCh38

View Full Product Details