Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 77337086-77337616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE8B
Entrez Gene ID
8622
Gene Name
phosphodiesterase 8B
Gene Aliases
ADSD, PPNAD3
UniGene
Hs.584830
Gene Chromosome Location
Chr.5: 77180252-77428256 on Build GRCh38
Transcripts
NM_001029851.2 NM_001029852.2 NM_001029853.2 NM_001029854.2 NM_003719.3 XM_005248621.4 XM_005248623.4 XM_005248624.4 XM_006714725.3 XM_006714726.3 XM_011543699.2 XM_011543700.2 XM_011543701.2 XM_011543702.2 XM_011543703.2 XM_011543704.2 XM_017010003.1 XM_017010004.1 XM_017010005.1 XM_017010006.1 XM_017010007.1 XM_017010008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4909999 c.753G>C p.L251L Chr.5: 77337271-77337271 on Build GRCh38
3617985 c.772G>A p.E258K Chr.5: 77337290-77337290 on Build GRCh38
6171474 c.770G>C p.G257A Chr.5: 77337288-77337288 on Build GRCh38
6470514 c.781T>C p.S261P Chr.5: 77337299-77337299 on Build GRCh38
4558488 c.769G>A p.G257R Chr.5: 77337287-77337287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35571993 Chr.5: 77337551-77337552 on Build GRCh38
rs746448762 Chr.5: 77337379-77337379 on Build GRCh38
rs917037410 Chr.5: 77337507-77337507 on Build GRCh38
rs961552970 Chr.5: 77337283-77337283 on Build GRCh38
rs1038557916 Chr.5: 77337293-77337293 on Build GRCh38
rs562220220 Chr.5: 77337401-77337401 on Build GRCh38
rs771818778 Chr.5: 77337301-77337301 on Build GRCh38
rs373873289 Chr.5: 77337205-77337205 on Build GRCh38
rs781352838 Chr.5: 77337192-77337192 on Build GRCh38
rs150803443 Chr.5: 77337153-77337153 on Build GRCh38
rs776997027 Chr.5: 77337297-77337297 on Build GRCh38
rs374181853 Chr.5: 77337353-77337353 on Build GRCh38
rs562513868 Chr.5: 77337181-77337181 on Build GRCh38
rs895071743 Chr.5: 77337480-77337480 on Build GRCh38
rs770339436 Chr.5: 77337352-77337352 on Build GRCh38
rs35052848 Chr.5: 77337274-77337275 on Build GRCh38
rs937081108 Chr.5: 77337119-77337119 on Build GRCh38
rs552915542 Chr.5: 77337282-77337282 on Build GRCh38
rs958796439 Chr.5: 77337443-77337443 on Build GRCh38
rs778839979 Chr.5: 77337243-77337243 on Build GRCh38
rs941599243 Chr.5: 77337583-77337583 on Build GRCh38
rs772379820 Chr.5: 77337369-77337369 on Build GRCh38
rs1015782787 Chr.5: 77337237-77337237 on Build GRCh38
rs908770123 Chr.5: 77337545-77337545 on Build GRCh38
rs746700414 Chr.5: 77337315-77337315 on Build GRCh38
rs1032813982 Chr.5: 77337438-77337438 on Build GRCh38
rs553718728 Chr.5: 77337532-77337532 on Build GRCh38
rs991663358 Chr.5: 77337123-77337123 on Build GRCh38
rs753509997 Chr.5: 77337228-77337228 on Build GRCh38
rs891129172 Chr.5: 77337370-77337370 on Build GRCh38
rs532821027 Chr.5: 77337269-77337269 on Build GRCh38
rs538431695 Chr.5: 77337337-77337337 on Build GRCh38
rs1012539322 Chr.5: 77337516-77337516 on Build GRCh38
rs7703993 Chr.5: 77337527-77337527 on Build GRCh38
rs371801170 Chr.5: 77337296-77337296 on Build GRCh38
rs542444122 Chr.5: 77337278-77337278 on Build GRCh38
rs936517177 Chr.5: 77337462-77337462 on Build GRCh38
rs529791320 Chr.5: 77337127-77337127 on Build GRCh38
rs180811025 Chr.5: 77337128-77337128 on Build GRCh38
rs1012912724 Chr.5: 77337150-77337150 on Build GRCh38
rs757811933 Chr.5: 77337188-77337198 on Build GRCh38
rs943806225 Chr.5: 77337365-77337365 on Build GRCh38
rs982443318 Chr.5: 77337529-77337529 on Build GRCh38
rs576839427 Chr.5: 77337551-77337551 on Build GRCh38
rs746367438 Chr.5: 77337339-77337339 on Build GRCh38
rs544861686 Chr.5: 77337589-77337589 on Build GRCh38
rs1053693506 Chr.5: 77337124-77337124 on Build GRCh38
rs974753737 Chr.5: 77337286-77337286 on Build GRCh38
rs1022938823 Chr.5: 77337173-77337173 on Build GRCh38
rs771357345 Chr.5: 77337274-77337274 on Build GRCh38
rs559972687 Chr.5: 77337591-77337591 on Build GRCh38
rs569307874 Chr.5: 77337323-77337323 on Build GRCh38
rs748005965 Chr.5: 77337254-77337254 on Build GRCh38
rs1002934644 Chr.5: 77337194-77337194 on Build GRCh38
rs1002987103 Chr.5: 77337584-77337584 on Build GRCh38
rs7717046 Chr.5: 77337472-77337472 on Build GRCh38
rs554906833 Chr.5: 77337429-77337429 on Build GRCh38
rs376744097 Chr.5: 77337466-77337466 on Build GRCh38
rs375768747 Chr.5: 77337314-77337314 on Build GRCh38
rs967342937 Chr.5: 77337400-77337400 on Build GRCh38
rs912188014 Chr.5: 77337350-77337350 on Build GRCh38
rs1020198994 Chr.5: 77337388-77337388 on Build GRCh38
rs753658190 Chr.5: 77337193-77337193 on Build GRCh38

View Full Product Details