Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 97107096-97107621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIX1
Entrez Gene ID
167410
Gene Name
limb and CNS expressed 1
Gene Aliases
C5orf11, Lft
UniGene
Hs.656702
Gene Chromosome Location
Chr.5: 97091870-97142816 on Build GRCh38
Transcripts
NM_153234.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1695984 c.332G>A p.R111K Chr.5: 97107415-97107415 on Build GRCh38
6701553 c.271A>G p.R91G Chr.5: 97107476-97107476 on Build GRCh38
5533847 c.309C>T p.L103L Chr.5: 97107438-97107438 on Build GRCh38
1071510 c.387C>T p.S129S Chr.5: 97107360-97107360 on Build GRCh38
6412619 c.333G>T p.R111S Chr.5: 97107414-97107414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770393249 Chr.5: 97107349-97107349 on Build GRCh38
rs781545420 Chr.5: 97107345-97107345 on Build GRCh38
rs772520455 Chr.5: 97107314-97107314 on Build GRCh38
rs570310506 Chr.5: 97107163-97107163 on Build GRCh38
rs184658292 Chr.5: 97107138-97107138 on Build GRCh38
rs762887624 Chr.5: 97107474-97107474 on Build GRCh38
rs371309305 Chr.5: 97107472-97107472 on Build GRCh38
rs780656334 Chr.5: 97107459-97107459 on Build GRCh38
rs200256217 Chr.5: 97107351-97107351 on Build GRCh38
rs115415106 Chr.5: 97107581-97107581 on Build GRCh38
rs760373048 Chr.5: 97107407-97107407 on Build GRCh38
rs376000708 Chr.5: 97107471-97107471 on Build GRCh38
rs747657330 Chr.5: 97107159-97107159 on Build GRCh38
rs780689544 Chr.5: 97107151-97107151 on Build GRCh38
rs771783696 Chr.5: 97107398-97107398 on Build GRCh38
rs868347687 Chr.5: 97107447-97107447 on Build GRCh38
rs751572432 Chr.5: 97107425-97107425 on Build GRCh38
rs747603292 Chr.5: 97107482-97107482 on Build GRCh38
rs948995282 Chr.5: 97107360-97107360 on Build GRCh38
rs559100888 Chr.5: 97107274-97107274 on Build GRCh38
rs760557926 Chr.5: 97107517-97107517 on Build GRCh38
rs975069997 Chr.5: 97107241-97107241 on Build GRCh38
rs778566465 Chr.5: 97107479-97107479 on Build GRCh38
rs978828680 Chr.5: 97107285-97107285 on Build GRCh38
rs373855407 Chr.5: 97107381-97107381 on Build GRCh38
rs921274670 Chr.5: 97107251-97107251 on Build GRCh38
rs746261676 Chr.5: 97107304-97107304 on Build GRCh38
rs970804777 Chr.5: 97107182-97107182 on Build GRCh38
rs759474404 Chr.5: 97107548-97107548 on Build GRCh38
rs934752595 Chr.5: 97107348-97107348 on Build GRCh38
rs776448351 Chr.5: 97107530-97107530 on Build GRCh38
rs866170302 Chr.5: 97107523-97107523 on Build GRCh38
rs6886042 Chr.5: 97107212-97107212 on Build GRCh38
rs1045077481 Chr.5: 97107376-97107376 on Build GRCh38
rs756406584 Chr.5: 97107443-97107443 on Build GRCh38
rs750868633 Chr.5: 97107441-97107441 on Build GRCh38
rs766405862 Chr.5: 97107418-97107418 on Build GRCh38
rs536009189 Chr.5: 97107483-97107483 on Build GRCh38
rs751803383 Chr.5: 97107321-97107321 on Build GRCh38
rs772734107 Chr.5: 97107506-97107506 on Build GRCh38
rs1021088473 Chr.5: 97107177-97107178 on Build GRCh38
rs536987358 Chr.5: 97107343-97107343 on Build GRCh38
rs1018745687 Chr.5: 97107179-97107179 on Build GRCh38
rs181110692 Chr.5: 97107268-97107268 on Build GRCh38
rs189522985 Chr.5: 97107239-97107239 on Build GRCh38
rs974682641 Chr.5: 97107588-97107588 on Build GRCh38
rs764190384 Chr.5: 97107424-97107424 on Build GRCh38
rs895989640 Chr.5: 97107359-97107359 on Build GRCh38
rs781383793 Chr.5: 97107437-97107437 on Build GRCh38
rs142107983 Chr.5: 97107423-97107423 on Build GRCh38
rs773073424 Chr.5: 97107399-97107399 on Build GRCh38
rs316179 Chr.5: 97107253-97107253 on Build GRCh38
rs376614473 Chr.5: 97107473-97107473 on Build GRCh38
rs770656318 Chr.5: 97107529-97107529 on Build GRCh38
rs958219237 Chr.5: 97107152-97107152 on Build GRCh38
rs768256040 Chr.5: 97107371-97107371 on Build GRCh38
rs368015626 Chr.5: 97107385-97107385 on Build GRCh38
rs185798147 Chr.5: 97107419-97107419 on Build GRCh38
rs116832505 Chr.5: 97107131-97107131 on Build GRCh38
rs757487556 Chr.5: 97107434-97107434 on Build GRCh38

View Full Product Details