Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150796265-150796803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSK
Entrez Gene ID
1513
Gene Name
cathepsin K
Gene Aliases
CTS02, CTSO, CTSO1, CTSO2, PKND, PYCD
UniGene
Hs.632466
Gene Chromosome Location
Chr.1: 150796208-150808441 on Build GRCh38
Transcripts
NM_000396.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022048 c.990A>G p.*330W Chr.1: 150796799-150796799 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201836605 Chr.1: 150796351-150796351 on Build GRCh38
rs774969011 Chr.1: 150796740-150796740 on Build GRCh38
rs201937004 Chr.1: 150796457-150796457 on Build GRCh38
rs1018316048 Chr.1: 150796579-150796579 on Build GRCh38
rs199862335 Chr.1: 150796335-150796335 on Build GRCh38
rs200974564 Chr.1: 150796338-150796338 on Build GRCh38
rs184390283 Chr.1: 150796486-150796486 on Build GRCh38
rs1049965775 Chr.1: 150796718-150796718 on Build GRCh38
rs201606490 Chr.1: 150796309-150796309 on Build GRCh38
rs587681751 Chr.1: 150796679-150796679 on Build GRCh38
rs899909150 Chr.1: 150796596-150796596 on Build GRCh38
rs202102294 Chr.1: 150796658-150796658 on Build GRCh38
rs587691660 Chr.1: 150796661-150796661 on Build GRCh38
rs949729794 Chr.1: 150796404-150796404 on Build GRCh38
rs587770564 Chr.1: 150796672-150796672 on Build GRCh38
rs981452256 Chr.1: 150796491-150796494 on Build GRCh38
rs891099345 Chr.1: 150796522-150796522 on Build GRCh38
rs200244717 Chr.1: 150796593-150796593 on Build GRCh38
rs200704218 Chr.1: 150796721-150796721 on Build GRCh38
rs201778975 Chr.1: 150796626-150796626 on Build GRCh38
rs1040117244 Chr.1: 150796741-150796741 on Build GRCh38
rs201085105 Chr.1: 150796480-150796480 on Build GRCh38
rs200109342 Chr.1: 150796468-150796468 on Build GRCh38
rs200621633 Chr.1: 150796329-150796329 on Build GRCh38
rs189456337 Chr.1: 150796367-150796367 on Build GRCh38
rs929857346 Chr.1: 150796510-150796510 on Build GRCh38
rs920220863 Chr.1: 150796559-150796559 on Build GRCh38
rs202196963 Chr.1: 150796573-150796573 on Build GRCh38
rs1008236566 Chr.1: 150796531-150796531 on Build GRCh38
rs768142570 Chr.1: 150796756-150796756 on Build GRCh38
rs956881073 Chr.1: 150796748-150796748 on Build GRCh38
rs1802666 Chr.1: 150796356-150796356 on Build GRCh38
rs587764536 Chr.1: 150796344-150796344 on Build GRCh38
rs200285919 Chr.1: 150796662-150796662 on Build GRCh38
rs181056495 Chr.1: 150796396-150796396 on Build GRCh38
rs1000986315 Chr.1: 150796614-150796614 on Build GRCh38
rs143752854 Chr.1: 150796677-150796677 on Build GRCh38
rs201446377 Chr.1: 150796332-150796332 on Build GRCh38
rs1063586 Chr.1: 150796668-150796668 on Build GRCh38
rs937138638 Chr.1: 150796462-150796462 on Build GRCh38
rs199806511 Chr.1: 150796316-150796316 on Build GRCh38
rs199537516 Chr.1: 150796692-150796692 on Build GRCh38
rs1038590435 Chr.1: 150796296-150796296 on Build GRCh38
rs998783780 Chr.1: 150796448-150796448 on Build GRCh38
rs201185711 Chr.1: 150796757-150796757 on Build GRCh38
rs1038597004 Chr.1: 150796464-150796464 on Build GRCh38
rs140135152 Chr.1: 150796456-150796456 on Build GRCh38
rs1047029333 Chr.1: 150796512-150796512 on Build GRCh38
rs961337211 Chr.1: 150796519-150796519 on Build GRCh38
rs1008650521 Chr.1: 150796363-150796363 on Build GRCh38
rs888907609 Chr.1: 150796340-150796340 on Build GRCh38

View Full Product Details