Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41770442-41770946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRS3
Entrez Gene ID
10817
Gene Name
fibroblast growth factor receptor substrate 3
Gene Aliases
FRS2-beta, FRS2B, FRS2beta, SNT-2, SNT2
UniGene
Hs.194208
Gene Chromosome Location
Chr.6: 41770176-41779905 on Build GRCh38
Transcripts
NM_006653.4 XM_011514254.2 XM_011514255.2 XM_017010193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4899506 c.1318C>T p.Q440* Chr.6: 41770780-41770780 on Build GRCh38
395339 c.1451G>C p.R484P Chr.6: 41770647-41770647 on Build GRCh38
7368839 c.1315C>A p.P439T Chr.6: 41770783-41770783 on Build GRCh38
3078755 c.1222G>T p.G408W Chr.6: 41770876-41770876 on Build GRCh38
7368838 c.1178G>T p.R393L Chr.6: 41770920-41770920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779642151 Chr.6: 41770750-41770750 on Build GRCh38
rs1008109447 Chr.6: 41770608-41770608 on Build GRCh38
rs201021700 Chr.6: 41770730-41770730 on Build GRCh38
rs769430931 Chr.6: 41770649-41770649 on Build GRCh38
rs148388051 Chr.6: 41770708-41770708 on Build GRCh38
rs1028874495 Chr.6: 41770477-41770477 on Build GRCh38
rs765943962 Chr.6: 41770917-41770917 on Build GRCh38
rs561176382 Chr.6: 41770756-41770756 on Build GRCh38
rs200002633 Chr.6: 41770800-41770800 on Build GRCh38
rs776868521 Chr.6: 41770780-41770780 on Build GRCh38
rs113801307 Chr.6: 41770744-41770744 on Build GRCh38
rs45493499 Chr.6: 41770509-41770509 on Build GRCh38
rs141634590 Chr.6: 41770729-41770729 on Build GRCh38
rs774684327 Chr.6: 41770579-41770579 on Build GRCh38
rs376991946 Chr.6: 41770703-41770703 on Build GRCh38
rs756179328 Chr.6: 41770603-41770603 on Build GRCh38
rs201804746 Chr.6: 41770610-41770610 on Build GRCh38
rs890969310 Chr.6: 41770597-41770597 on Build GRCh38
rs761065569 Chr.6: 41770679-41770679 on Build GRCh38
rs754585539 Chr.6: 41770864-41770864 on Build GRCh38
rs780744813 Chr.6: 41770733-41770733 on Build GRCh38
rs757731877 Chr.6: 41770924-41770924 on Build GRCh38
rs369896862 Chr.6: 41770813-41770813 on Build GRCh38
rs567855440 Chr.6: 41770589-41770589 on Build GRCh38
rs763060887 Chr.6: 41770790-41770790 on Build GRCh38
rs757223319 Chr.6: 41770809-41770809 on Build GRCh38
rs776302253 Chr.6: 41770574-41770574 on Build GRCh38
rs369539341 Chr.6: 41770902-41770902 on Build GRCh38
rs771501954 Chr.6: 41770778-41770778 on Build GRCh38
rs755660918 Chr.6: 41770818-41770818 on Build GRCh38
rs751306785 Chr.6: 41770801-41770801 on Build GRCh38
rs746527722 Chr.6: 41770640-41770640 on Build GRCh38
rs745577374 Chr.6: 41770647-41770647 on Build GRCh38
rs919999753 Chr.6: 41770551-41770551 on Build GRCh38
rs142682061 Chr.6: 41770885-41770885 on Build GRCh38
rs35653109 Chr.6: 41770842-41770843 on Build GRCh38
rs201687292 Chr.6: 41770773-41770773 on Build GRCh38
rs148685960 Chr.6: 41770923-41770923 on Build GRCh38
rs991342352 Chr.6: 41770635-41770635 on Build GRCh38
rs752476650 Chr.6: 41770615-41770615 on Build GRCh38
rs150452181 Chr.6: 41770643-41770643 on Build GRCh38
rs758278191 Chr.6: 41770632-41770632 on Build GRCh38
rs754676624 Chr.6: 41770920-41770920 on Build GRCh38
rs267601032 Chr.6: 41770761-41770761 on Build GRCh38
rs766403918 Chr.6: 41770724-41770724 on Build GRCh38
rs760530626 Chr.6: 41770691-41770691 on Build GRCh38
rs767384110 Chr.6: 41770812-41770812 on Build GRCh38
rs373596042 Chr.6: 41770633-41770633 on Build GRCh38
rs138554790 Chr.6: 41770646-41770646 on Build GRCh38
rs768011239 Chr.6: 41770593-41770593 on Build GRCh38
rs767269665 Chr.6: 41770911-41770911 on Build GRCh38
rs375323570 Chr.6: 41770781-41770781 on Build GRCh38
rs997039207 Chr.6: 41770476-41770476 on Build GRCh38
rs749028496 Chr.6: 41770840-41770840 on Build GRCh38
rs372825044 Chr.6: 41770918-41770918 on Build GRCh38
rs201392291 Chr.6: 41770921-41770921 on Build GRCh38
rs747503068 Chr.6: 41770507-41770507 on Build GRCh38
rs1028068707 Chr.6: 41770782-41770782 on Build GRCh38
rs769449384 Chr.6: 41770749-41770749 on Build GRCh38
rs759845275 Chr.6: 41770702-41770702 on Build GRCh38
rs753966448 Chr.6: 41770698-41770698 on Build GRCh38
rs202004280 Chr.6: 41770660-41770660 on Build GRCh38
rs185330233 Chr.6: 41770867-41770867 on Build GRCh38
rs887006788 Chr.6: 41770717-41770717 on Build GRCh38
rs761314357 Chr.6: 41770777-41770777 on Build GRCh38
rs914148461 Chr.6: 41770538-41770538 on Build GRCh38
rs762993887 Chr.6: 41770547-41770547 on Build GRCh38
rs868023164 Chr.6: 41770541-41770541 on Build GRCh38
rs532167853 Chr.6: 41770871-41770871 on Build GRCh38
rs930086867 Chr.6: 41770562-41770562 on Build GRCh38
rs144170980 Chr.6: 41770916-41770916 on Build GRCh38
rs773439380 Chr.6: 41770774-41770774 on Build GRCh38
rs747282093 Chr.6: 41770872-41770872 on Build GRCh38
rs75934525 Chr.6: 41770492-41770492 on Build GRCh38
rs34062751 Chr.6: 41770884-41770885 on Build GRCh38
rs777234512 Chr.6: 41770878-41770878 on Build GRCh38
rs774209432 Chr.6: 41770909-41770909 on Build GRCh38
rs772489038 Chr.6: 41770670-41770670 on Build GRCh38
rs769993190 Chr.6: 41770502-41770502 on Build GRCh38
rs746342094 Chr.6: 41770879-41770879 on Build GRCh38
rs750411279 Chr.6: 41770915-41770915 on Build GRCh38
rs534138466 Chr.6: 41770576-41770576 on Build GRCh38
rs770041090 Chr.6: 41770882-41770882 on Build GRCh38
rs762117793 Chr.6: 41770669-41770669 on Build GRCh38
rs757127616 Chr.6: 41770732-41770732 on Build GRCh38
rs1046859323 Chr.6: 41770571-41770571 on Build GRCh38
rs773809864 Chr.6: 41770674-41770674 on Build GRCh38
rs574639227 Chr.6: 41770787-41770787 on Build GRCh38
rs200505680 Chr.6: 41770609-41770609 on Build GRCh38
rs781671519 Chr.6: 41770925-41770925 on Build GRCh38
rs57289703 Chr.6: 41770788-41770788 on Build GRCh38
rs775415683 Chr.6: 41770884-41770884 on Build GRCh38
rs779519233 Chr.6: 41770831-41770831 on Build GRCh38
rs778920144 Chr.6: 41770612-41770612 on Build GRCh38
rs936595074 Chr.6: 41770903-41770903 on Build GRCh38
rs745309580 Chr.6: 41770741-41770741 on Build GRCh38
rs1008382609 Chr.6: 41770797-41770797 on Build GRCh38
rs137862726 Chr.6: 41770877-41770877 on Build GRCh38

View Full Product Details