Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136924514-136925020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35D3
Entrez Gene ID
340146
Gene Name
solute carrier family 35 member D3
Gene Aliases
FRCL1
UniGene
Hs.369703
Gene Chromosome Location
Chr.6: 136922241-136925660 on Build GRCh38
Transcripts
NM_001008783.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5061433 c.1239A>G p.L413L Chr.6: 136924684-136924684 on Build GRCh38
739940 c.1100C>G p.P367R Chr.6: 136924545-136924545 on Build GRCh38
4521835 c.1132G>A p.E378K Chr.6: 136924577-136924577 on Build GRCh38
3621014 c.1246C>T p.P416S Chr.6: 136924691-136924691 on Build GRCh38
739939 c.1170C>A p.L390L Chr.6: 136924615-136924615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986458585 Chr.6: 136924794-136924794 on Build GRCh38
rs930802669 Chr.6: 136924902-136924902 on Build GRCh38
rs943936940 Chr.6: 136924939-136924939 on Build GRCh38
rs982870207 Chr.6: 136924877-136924877 on Build GRCh38
rs368466205 Chr.6: 136924695-136924695 on Build GRCh38
rs919679754 Chr.6: 136924857-136924857 on Build GRCh38
rs746987224 Chr.6: 136924679-136924679 on Build GRCh38
rs757230047 Chr.6: 136924589-136924589 on Build GRCh38
rs144509540 Chr.6: 136924647-136924647 on Build GRCh38
rs774008192 Chr.6: 136924770-136924770 on Build GRCh38
rs912189856 Chr.6: 136924796-136924796 on Build GRCh38
rs149997137 Chr.6: 136924621-136924621 on Build GRCh38
rs756330680 Chr.6: 136924572-136924572 on Build GRCh38
rs558466578 Chr.6: 136924839-136924839 on Build GRCh38
rs767981863 Chr.6: 136924557-136924557 on Build GRCh38
rs974986741 Chr.6: 136924879-136924879 on Build GRCh38
rs146545099 Chr.6: 136924676-136924676 on Build GRCh38
rs535021312 Chr.6: 136924767-136924767 on Build GRCh38
rs375806022 Chr.6: 136924714-136924714 on Build GRCh38
rs772818751 Chr.6: 136924722-136924722 on Build GRCh38
rs578174006 Chr.6: 136924919-136924919 on Build GRCh38
rs779924613 Chr.6: 136924678-136924678 on Build GRCh38
rs973785634 Chr.6: 136924832-136924832 on Build GRCh38
rs905825273 Chr.6: 136924977-136924977 on Build GRCh38
rs776097741 Chr.6: 136924657-136924657 on Build GRCh38
rs746817776 Chr.6: 136924604-136924604 on Build GRCh38
rs753707580 Chr.6: 136924555-136924555 on Build GRCh38
rs550790929 Chr.6: 136924655-136924655 on Build GRCh38
rs955055099 Chr.6: 136924783-136924783 on Build GRCh38
rs765284697 Chr.6: 136924675-136924675 on Build GRCh38
rs376307317 Chr.6: 136924842-136924842 on Build GRCh38
rs768525970 Chr.6: 136924615-136924615 on Build GRCh38
rs201048173 Chr.6: 136924588-136924588 on Build GRCh38
rs764185622 Chr.6: 136924573-136924573 on Build GRCh38
rs757332089 Chr.6: 136924550-136924550 on Build GRCh38
rs199789519 Chr.6: 136924662-136924662 on Build GRCh38
rs758383949 Chr.6: 136924596-136924596 on Build GRCh38
rs570202899 Chr.6: 136924659-136924659 on Build GRCh38
rs775936507 Chr.6: 136924738-136924739 on Build GRCh38
rs753943999 Chr.6: 136924667-136924667 on Build GRCh38
rs1020028786 Chr.6: 136924774-136924774 on Build GRCh38
rs754901014 Chr.6: 136924651-136924653 on Build GRCh38
rs760098762 Chr.6: 136924653-136924653 on Build GRCh38
rs757562966 Chr.6: 136924668-136924668 on Build GRCh38
rs766081092 Chr.6: 136924748-136924748 on Build GRCh38
rs765266011 Chr.6: 136924671-136924671 on Build GRCh38
rs780324658 Chr.6: 136924874-136924874 on Build GRCh38
rs114845543 Chr.6: 136924934-136924934 on Build GRCh38
rs201799622 Chr.6: 136924545-136924545 on Build GRCh38
rs551625874 Chr.6: 136924616-136924616 on Build GRCh38
rs62420739 Chr.6: 136924717-136924717 on Build GRCh38
rs140834292 Chr.6: 136924593-136924593 on Build GRCh38
rs769361386 Chr.6: 136924628-136924628 on Build GRCh38
rs965336508 Chr.6: 136924811-136924811 on Build GRCh38
rs200380943 Chr.6: 136924597-136924597 on Build GRCh38
rs745796092 Chr.6: 136924594-136924594 on Build GRCh38
rs1053773146 Chr.6: 136924986-136924986 on Build GRCh38
rs760188514 Chr.6: 136924555-136924555 on Build GRCh38
rs951057372 Chr.6: 136924869-136924869 on Build GRCh38
rs753163288 Chr.6: 136924562-136924562 on Build GRCh38
rs971374447 Chr.6: 136924771-136924771 on Build GRCh38
rs140043418 Chr.6: 136924696-136924696 on Build GRCh38
rs1045706111 Chr.6: 136924976-136924976 on Build GRCh38
rs867467868 Chr.6: 136924645-136924645 on Build GRCh38
rs907951365 Chr.6: 136924967-136924967 on Build GRCh38
rs775805967 Chr.6: 136924730-136924730 on Build GRCh38
rs920772250 Chr.6: 136924884-136924884 on Build GRCh38
rs770590109 Chr.6: 136924726-136924726 on Build GRCh38
rs572373721 Chr.6: 136924713-136924713 on Build GRCh38
rs753820094 Chr.6: 136924542-136924542 on Build GRCh38
rs1039610220 Chr.6: 136924538-136924538 on Build GRCh38
rs774941211 Chr.6: 136924554-136924554 on Build GRCh38
rs563533702 Chr.6: 136924975-136924975 on Build GRCh38
rs764825335 Chr.6: 136924547-136924547 on Build GRCh38
rs747743269 Chr.6: 136924704-136924704 on Build GRCh38
rs748770185 Chr.6: 136924723-136924723 on Build GRCh38

View Full Product Details