Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27153335-27153845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA-AS3
Entrez Gene ID
100133311
Gene Name
HOXA cluster antisense RNA 3
Gene Aliases
HOXA6as
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27140364-27155928 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 NM_006896.3

Gene Symbol
HOXA7
Entrez Gene ID
3204
Gene Name
homeobox A7
Gene Aliases
ANTP, HOX1, HOX1.1, HOX1A
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27153716-27156677 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 NM_006896.3

SNP Detail

SNP ID Chromosome Location
rs957333272 Chr.7: 27153517-27153517 on Build GRCh38
rs531988879 Chr.7: 27153545-27153545 on Build GRCh38
rs1038611604 Chr.7: 27153379-27153379 on Build GRCh38
rs1055870758 Chr.7: 27153646-27153646 on Build GRCh38
rs937294170 Chr.7: 27153784-27153784 on Build GRCh38
rs912884936 Chr.7: 27153549-27153549 on Build GRCh38
rs1008241500 Chr.7: 27153746-27153746 on Build GRCh38
rs774706408 Chr.7: 27153653-27153653 on Build GRCh38
rs966873932 Chr.7: 27153620-27153620 on Build GRCh38
rs1018876865 Chr.7: 27153426-27153426 on Build GRCh38
rs995628169 Chr.7: 27153390-27153390 on Build GRCh38
rs764616299 Chr.7: 27153810-27153810 on Build GRCh38
rs190750155 Chr.7: 27153794-27153794 on Build GRCh38
rs1004801447 Chr.7: 27153415-27153415 on Build GRCh38
rs887150789 Chr.7: 27153408-27153408 on Build GRCh38
rs1010951591 Chr.7: 27153558-27153558 on Build GRCh38
rs749354036 Chr.7: 27153547-27153547 on Build GRCh38
rs941028871 Chr.7: 27153705-27153705 on Build GRCh38
rs1047185036 Chr.7: 27153732-27153732 on Build GRCh38
rs895112554 Chr.7: 27153803-27153803 on Build GRCh38
rs554291478 Chr.7: 27153747-27153747 on Build GRCh38
rs989948404 Chr.7: 27153533-27153533 on Build GRCh38
rs981624098 Chr.7: 27153606-27153606 on Build GRCh38
rs985114571 Chr.7: 27153660-27153660 on Build GRCh38
rs534304376 Chr.7: 27153733-27153733 on Build GRCh38
rs1003482076 Chr.7: 27153503-27153503 on Build GRCh38
rs920135972 Chr.7: 27153661-27153661 on Build GRCh38
rs1026053021 Chr.7: 27153405-27153405 on Build GRCh38
rs1028725490 Chr.7: 27153508-27153508 on Build GRCh38
rs778020335 Chr.7: 27153507-27153507 on Build GRCh38
rs928903524 Chr.7: 27153625-27153625 on Build GRCh38
rs116895484 Chr.7: 27153697-27153697 on Build GRCh38
rs1040288225 Chr.7: 27153761-27153761 on Build GRCh38
rs994416428 Chr.7: 27153413-27153413 on Build GRCh38
rs771315711 Chr.7: 27153588-27153588 on Build GRCh38
rs763102098 Chr.7: 27153558-27153558 on Build GRCh38
rs756037706 Chr.7: 27153452-27153452 on Build GRCh38
rs775860691 Chr.7: 27153767-27153767 on Build GRCh38
rs185652202 Chr.7: 27153630-27153630 on Build GRCh38
rs1022809970 Chr.7: 27153579-27153579 on Build GRCh38
rs907148781 Chr.7: 27153442-27153442 on Build GRCh38
rs918346625 Chr.7: 27153718-27153721 on Build GRCh38
rs973816400 Chr.7: 27153715-27153715 on Build GRCh38
rs1045597422 Chr.7: 27153432-27153433 on Build GRCh38
rs75300473 Chr.7: 27153662-27153662 on Build GRCh38
rs375261420 Chr.7: 27153648-27153651 on Build GRCh38

View Full Product Details