Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109556764-109557265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBAG9
Entrez Gene ID
9166
Gene Name
estrogen receptor binding site associated, antigen, 9
Gene Aliases
EB9, PDAF
UniGene
Hs.409368
Gene Chromosome Location
Chr.8: 109539700-109565996 on Build GRCh38
Transcripts
NM_001278938.1 NM_004215.4 NM_198120.2 XM_017013960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4544984 c.367G>A p.G123R Chr.8: 109556980-109556980 on Build GRCh38
1094985 c.340G>T p.E114* Chr.8: 109556953-109556953 on Build GRCh38
5041267 c.425A>G p.Q142R Chr.8: 109557038-109557038 on Build GRCh38
461497 c.364G>C p.D122H Chr.8: 109556977-109556977 on Build GRCh38
3884726 c.348G>T p.L116F Chr.8: 109556961-109556961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756671301 Chr.8: 109556912-109556912 on Build GRCh38
rs965778655 Chr.8: 109557234-109557234 on Build GRCh38
rs375694130 Chr.8: 109557042-109557042 on Build GRCh38
rs372276656 Chr.8: 109556979-109556979 on Build GRCh38
rs758704614 Chr.8: 109556937-109556937 on Build GRCh38
rs563652595 Chr.8: 109556870-109556870 on Build GRCh38
rs185499732 Chr.8: 109556996-109556996 on Build GRCh38
rs767904213 Chr.8: 109557071-109557071 on Build GRCh38
rs182803837 Chr.8: 109557161-109557161 on Build GRCh38
rs569999297 Chr.8: 109556877-109556877 on Build GRCh38
rs764217741 Chr.8: 109557049-109557049 on Build GRCh38
rs777162647 Chr.8: 109556984-109556984 on Build GRCh38
rs896575811 Chr.8: 109557104-109557104 on Build GRCh38
rs577670753 Chr.8: 109557148-109557148 on Build GRCh38
rs534001160 Chr.8: 109557050-109557050 on Build GRCh38
rs938024160 Chr.8: 109557009-109557009 on Build GRCh38
rs775325297 Chr.8: 109557201-109557201 on Build GRCh38
rs1027601353 Chr.8: 109557173-109557173 on Build GRCh38
rs746651770 Chr.8: 109556987-109556987 on Build GRCh38
rs372226082 Chr.8: 109557079-109557079 on Build GRCh38
rs1044007074 Chr.8: 109556978-109556978 on Build GRCh38
rs775429216 Chr.8: 109557026-109557026 on Build GRCh38
rs945618853 Chr.8: 109556967-109556967 on Build GRCh38
rs1014006127 Chr.8: 109557110-109557110 on Build GRCh38
rs985849961 Chr.8: 109556858-109556858 on Build GRCh38
rs977851916 Chr.8: 109557094-109557094 on Build GRCh38
rs923371010 Chr.8: 109557107-109557107 on Build GRCh38
rs568472807 Chr.8: 109556794-109556794 on Build GRCh38
rs749793004 Chr.8: 109556959-109556959 on Build GRCh38
rs986181065 Chr.8: 109557137-109557137 on Build GRCh38
rs967384326 Chr.8: 109557091-109557091 on Build GRCh38
rs951670160 Chr.8: 109557196-109557196 on Build GRCh38
rs201074477 Chr.8: 109557014-109557014 on Build GRCh38
rs757607217 Chr.8: 109556897-109556897 on Build GRCh38
rs182265187 Chr.8: 109556895-109556895 on Build GRCh38
rs751744225 Chr.8: 109556894-109556894 on Build GRCh38
rs550433240 Chr.8: 109556804-109556804 on Build GRCh38
rs190164146 Chr.8: 109557155-109557155 on Build GRCh38
rs1017331620 Chr.8: 109557230-109557230 on Build GRCh38
rs914110273 Chr.8: 109556942-109556942 on Build GRCh38
rs745612010 Chr.8: 109557000-109557000 on Build GRCh38
rs994256722 Chr.8: 109556916-109556916 on Build GRCh38
rs1038266533 Chr.8: 109557235-109557235 on Build GRCh38
rs770610827 Chr.8: 109556992-109556992 on Build GRCh38
rs747158750 Chr.8: 109556994-109556994 on Build GRCh38
rs781683869 Chr.8: 109556903-109556903 on Build GRCh38
rs201700353 Chr.8: 109556921-109556921 on Build GRCh38
rs1024550470 Chr.8: 109557127-109557127 on Build GRCh38
rs756509881 Chr.8: 109557083-109557083 on Build GRCh38
rs780564381 Chr.8: 109556954-109556954 on Build GRCh38
rs770940185 Chr.8: 109557084-109557085 on Build GRCh38
rs951715525 Chr.8: 109556825-109556825 on Build GRCh38
rs778083347 Chr.8: 109556817-109556820 on Build GRCh38
rs750861557 Chr.8: 109556908-109556908 on Build GRCh38
rs866321465 Chr.8: 109556974-109556974 on Build GRCh38

View Full Product Details