Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 124556123-124556623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTSS1
Entrez Gene ID
9788
Gene Name
metastasis suppressor 1
Gene Aliases
MIM, MIMA, MIMB
UniGene
Hs.336994
Gene Chromosome Location
Chr.8: 124550770-124728507 on Build GRCh38
Transcripts
NM_001282971.1 NM_001282974.1 NM_014751.5 XM_005251111.1 XM_005251113.1 XM_005251118.1 XM_006716700.1 XM_006716701.1 XM_006716702.1 XM_006716703.1 XM_006716704.1 XM_006716705.1 XM_006716706.1 XM_011517403.1 XM_011517404.2 XM_011517407.1 XM_017014086.1 XM_017014087.1 XM_017014088.1 XM_017014089.1 XM_017014090.1 XM_017014091.1 XM_017014092.1 XM_017014093.1 XM_017014094.1 XM_017014095.1 XM_017014096.1 XM_017014097.1 XM_017014098.1 XM_017014099.1 XM_017014100.1 XM_017014101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6262607 c.1401C>A p.T467T Chr.8: 124556235-124556235 on Build GRCh38
3663552 c.1299A>T p.P433P Chr.8: 124556262-124556262 on Build GRCh38
486062 c.1352C>T p.A451V Chr.8: 124556284-124556284 on Build GRCh38
486061 c.1394C>T p.A465V Chr.8: 124556242-124556242 on Build GRCh38
7225546 c.1315G>A p.G439R Chr.8: 124556321-124556321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907545260 Chr.8: 124556174-124556174 on Build GRCh38
rs3829036 Chr.8: 124556382-124556382 on Build GRCh38
rs368901804 Chr.8: 124556538-124556538 on Build GRCh38
rs766709048 Chr.8: 124556389-124556389 on Build GRCh38
rs375921579 Chr.8: 124556447-124556447 on Build GRCh38
rs765084882 Chr.8: 124556215-124556215 on Build GRCh38
rs745964103 Chr.8: 124556189-124556189 on Build GRCh38
rs758854509 Chr.8: 124556422-124556422 on Build GRCh38
rs748523312 Chr.8: 124556270-124556270 on Build GRCh38
rs145702575 Chr.8: 124556261-124556261 on Build GRCh38
rs894467663 Chr.8: 124556519-124556519 on Build GRCh38
rs772902027 Chr.8: 124556377-124556377 on Build GRCh38
rs907532510 Chr.8: 124556361-124556361 on Build GRCh38
rs143535407 Chr.8: 124556354-124556354 on Build GRCh38
rs534193043 Chr.8: 124556462-124556462 on Build GRCh38
rs531462070 Chr.8: 124556145-124556145 on Build GRCh38
rs879523273 Chr.8: 124556288-124556288 on Build GRCh38
rs771635761 Chr.8: 124556168-124556181 on Build GRCh38
rs780688659 Chr.8: 124556347-124556347 on Build GRCh38
rs748461204 Chr.8: 124556338-124556338 on Build GRCh38
rs565115426 Chr.8: 124556152-124556152 on Build GRCh38
rs151067767 Chr.8: 124556245-124556245 on Build GRCh38
rs774073612 Chr.8: 124556304-124556304 on Build GRCh38
rs141157052 Chr.8: 124556271-124556271 on Build GRCh38
rs570817732 Chr.8: 124556321-124556321 on Build GRCh38
rs750455722 Chr.8: 124556255-124556255 on Build GRCh38
rs772974553 Chr.8: 124556206-124556206 on Build GRCh38
rs773010137 Chr.8: 124556191-124556191 on Build GRCh38
rs757493607 Chr.8: 124556175-124556175 on Build GRCh38
rs940406945 Chr.8: 124556192-124556192 on Build GRCh38
rs781093222 Chr.8: 124556188-124556188 on Build GRCh38
rs751829788 Chr.8: 124556167-124556167 on Build GRCh38
rs770644756 Chr.8: 124556211-124556211 on Build GRCh38
rs779375889 Chr.8: 124556265-124556265 on Build GRCh38
rs752374662 Chr.8: 124556364-124556364 on Build GRCh38
rs755561232 Chr.8: 124556262-124556262 on Build GRCh38
rs552509798 Chr.8: 124556312-124556312 on Build GRCh38
rs117595835 Chr.8: 124556190-124556190 on Build GRCh38
rs752480171 Chr.8: 124556153-124556153 on Build GRCh38
rs969525792 Chr.8: 124556328-124556328 on Build GRCh38
rs527705728 Chr.8: 124556201-124556201 on Build GRCh38
rs60522958 Chr.8: 124556520-124556520 on Build GRCh38
rs761725291 Chr.8: 124556305-124556305 on Build GRCh38
rs1049178471 Chr.8: 124556170-124556170 on Build GRCh38
rs757355652 Chr.8: 124556251-124556251 on Build GRCh38
rs753908447 Chr.8: 124556397-124556397 on Build GRCh38
rs145509745 Chr.8: 124556244-124556244 on Build GRCh38
rs994930141 Chr.8: 124556496-124556496 on Build GRCh38
rs377523813 Chr.8: 124556331-124556331 on Build GRCh38
rs1032764633 Chr.8: 124556586-124556586 on Build GRCh38
rs779624283 Chr.8: 124556333-124556333 on Build GRCh38
rs752674509 Chr.8: 124556217-124556217 on Build GRCh38
rs761480446 Chr.8: 124556368-124556368 on Build GRCh38
rs774914986 Chr.8: 124556352-124556352 on Build GRCh38
rs574306790 Chr.8: 124556481-124556481 on Build GRCh38
rs745341192 Chr.8: 124556281-124556281 on Build GRCh38
rs761461466 Chr.8: 124556153-124556153 on Build GRCh38
rs769181024 Chr.8: 124556203-124556203 on Build GRCh38
rs753299348 Chr.8: 124556409-124556409 on Build GRCh38
rs768566662 Chr.8: 124556357-124556357 on Build GRCh38
rs760575019 Chr.8: 124556221-124556222 on Build GRCh38
rs762715694 Chr.8: 124556220-124556220 on Build GRCh38
rs755705849 Chr.8: 124556431-124556431 on Build GRCh38
rs373836065 Chr.8: 124556437-124556437 on Build GRCh38
rs767626905 Chr.8: 124556307-124556307 on Build GRCh38
rs1056234324 Chr.8: 124556503-124556503 on Build GRCh38
rs920935858 Chr.8: 124556429-124556429 on Build GRCh38
rs529286591 Chr.8: 124556300-124556300 on Build GRCh38
rs764024228 Chr.8: 124556303-124556303 on Build GRCh38
rs776415609 Chr.8: 124556275-124556275 on Build GRCh38
rs760351952 Chr.8: 124556210-124556210 on Build GRCh38
rs777943064 Chr.8: 124556158-124556158 on Build GRCh38
rs568990349 Chr.8: 124556343-124556343 on Build GRCh38
rs372644783 Chr.8: 124556440-124556440 on Build GRCh38
rs775950756 Chr.8: 124556212-124556212 on Build GRCh38
rs566830744 Chr.8: 124556298-124556298 on Build GRCh38
rs1040139544 Chr.8: 124556220-124556225 on Build GRCh38
rs774339288 Chr.8: 124556358-124556358 on Build GRCh38
rs3116129 Chr.8: 124556563-124556563 on Build GRCh38
rs748791835 Chr.8: 124556439-124556439 on Build GRCh38
rs767393317 Chr.8: 124556370-124556370 on Build GRCh38
rs146742280 Chr.8: 124556332-124556332 on Build GRCh38
rs759147853 Chr.8: 124556213-124556213 on Build GRCh38
rs756281738 Chr.8: 124556257-124556257 on Build GRCh38
rs373748993 Chr.8: 124556191-124556191 on Build GRCh38
rs1051970702 Chr.8: 124556225-124556225 on Build GRCh38
rs140769162 Chr.8: 124556405-124556405 on Build GRCh38
rs967197210 Chr.8: 124556561-124556561 on Build GRCh38
rs556171116 Chr.8: 124556590-124556590 on Build GRCh38
rs759760129 Chr.8: 124556404-124556404 on Build GRCh38
rs773862533 Chr.8: 124556149-124556150 on Build GRCh38
rs371399134 Chr.8: 124556297-124556297 on Build GRCh38
rs149780096 Chr.8: 124556253-124556253 on Build GRCh38
rs545220580 Chr.8: 124556151-124556151 on Build GRCh38
rs753405889 Chr.8: 124556149-124556149 on Build GRCh38
rs780666652 Chr.8: 124556196-124556196 on Build GRCh38
rs1024153021 Chr.8: 124556536-124556536 on Build GRCh38
rs975697082 Chr.8: 124556568-124556568 on Build GRCh38
rs922953303 Chr.8: 124556601-124556601 on Build GRCh38
rs756122618 Chr.8: 124556322-124556322 on Build GRCh38
rs751876372 Chr.8: 124556425-124556425 on Build GRCh38
rs780673847 Chr.8: 124556260-124556260 on Build GRCh38
rs932139601 Chr.8: 124556182-124556182 on Build GRCh38
rs199822222 Chr.8: 124556423-124556423 on Build GRCh38
rs749665452 Chr.8: 124556202-124556202 on Build GRCh38
rs554181662 Chr.8: 124556472-124556472 on Build GRCh38
rs140348524 Chr.8: 124556351-124556351 on Build GRCh38
rs1013614498 Chr.8: 124556461-124556461 on Build GRCh38
rs745477417 Chr.8: 124556350-124556350 on Build GRCh38

View Full Product Details