Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 116699977-116700481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
UniGene
Hs.601562
Gene Chromosome Location
Chr.9: 116425225-117415039 on Build GRCh38
Transcripts
NM_014010.4 NM_001099679.1 NM_012210.3 XM_005251813.3 XM_011518398.2 XM_017014486.1

Gene Symbol
TRIM32
Entrez Gene ID
22954
Gene Name
tripartite motif containing 32
Gene Aliases
BBS11, HT2A, LGMD2H, TATIP
UniGene
Hs.591910
Gene Chromosome Location
Chr.9: 116687302-116701300 on Build GRCh38
Transcripts
NM_014010.4 NM_001099679.1 NM_012210.3 XM_005251813.3 XM_011518398.2 XM_017014486.1

SNP Detail

SNP ID Chromosome Location
rs556842802 Chr.9: 116700212-116700212 on Build GRCh38
rs1046718253 Chr.9: 116700110-116700110 on Build GRCh38
rs990730197 Chr.9: 116700424-116700424 on Build GRCh38
rs899533188 Chr.9: 116700273-116700273 on Build GRCh38
rs192837427 Chr.9: 116700234-116700234 on Build GRCh38
rs533288656 Chr.9: 116700435-116700435 on Build GRCh38
rs957394452 Chr.9: 116700275-116700275 on Build GRCh38
rs1042917467 Chr.9: 116700320-116700320 on Build GRCh38
rs116058338 Chr.9: 116700396-116700396 on Build GRCh38
rs541032542 Chr.9: 116700380-116700380 on Build GRCh38
rs568812188 Chr.9: 116700232-116700232 on Build GRCh38
rs917981790 Chr.9: 116700146-116700146 on Build GRCh38
rs16933835 Chr.9: 116700293-116700293 on Build GRCh38
rs577814903 Chr.9: 116700096-116700096 on Build GRCh38
rs930335506 Chr.9: 116700447-116700447 on Build GRCh38
rs1009722752 Chr.9: 116700030-116700030 on Build GRCh38
rs968677643 Chr.9: 116700072-116700072 on Build GRCh38
rs753259162 Chr.9: 116700187-116700187 on Build GRCh38
rs779667568 Chr.9: 116700452-116700452 on Build GRCh38
rs927913945 Chr.9: 116700120-116700121 on Build GRCh38
rs908462619 Chr.9: 116700188-116700188 on Build GRCh38
rs1019586421 Chr.9: 116700228-116700228 on Build GRCh38
rs927829780 Chr.9: 116700410-116700410 on Build GRCh38
rs988883568 Chr.9: 116700286-116700286 on Build GRCh38
rs750062920 Chr.9: 116700428-116700428 on Build GRCh38
rs534626362 Chr.9: 116700053-116700053 on Build GRCh38
rs768024209 Chr.9: 116700186-116700186 on Build GRCh38
rs898873787 Chr.9: 116700182-116700182 on Build GRCh38
rs937638936 Chr.9: 116700173-116700173 on Build GRCh38
rs1050857521 Chr.9: 116700289-116700289 on Build GRCh38
rs535411189 Chr.9: 116700013-116700014 on Build GRCh38
rs1009752257 Chr.9: 116700309-116700309 on Build GRCh38
rs966915576 Chr.9: 116700303-116700303 on Build GRCh38
rs759347153 Chr.9: 116700207-116700207 on Build GRCh38
rs957445349 Chr.9: 116700073-116700073 on Build GRCh38
rs996541786 Chr.9: 116700249-116700249 on Build GRCh38
rs1021226731 Chr.9: 116700031-116700031 on Build GRCh38
rs543534409 Chr.9: 116700183-116700183 on Build GRCh38
rs937938071 Chr.9: 116700411-116700411 on Build GRCh38
rs932295071 Chr.9: 116700287-116700287 on Build GRCh38
rs527953664 Chr.9: 116700332-116700332 on Build GRCh38
rs959389227 Chr.9: 116700371-116700371 on Build GRCh38
rs758004405 Chr.9: 116700429-116700429 on Build GRCh38
rs751662240 Chr.9: 116700101-116700101 on Build GRCh38
rs549857802 Chr.9: 116700441-116700441 on Build GRCh38
rs866692844 Chr.9: 116700075-116700075 on Build GRCh38
rs187467050 Chr.9: 116700076-116700076 on Build GRCh38
rs1033011324 Chr.9: 116700262-116700262 on Build GRCh38
rs1025025394 Chr.9: 116700393-116700393 on Build GRCh38
rs974784095 Chr.9: 116700035-116700035 on Build GRCh38
rs913732447 Chr.9: 116700090-116700090 on Build GRCh38
rs1001557859 Chr.9: 116700365-116700365 on Build GRCh38
rs1033921107 Chr.9: 116700369-116700369 on Build GRCh38
rs868637838 Chr.9: 116700089-116700089 on Build GRCh38
rs778038121 Chr.9: 116700372-116700372 on Build GRCh38
rs144172952 Chr.9: 116700216-116700216 on Build GRCh38
rs759997807 Chr.9: 116700174-116700174 on Build GRCh38
rs774289005 Chr.9: 116700196-116700196 on Build GRCh38
rs904108162 Chr.9: 116700329-116700329 on Build GRCh38

View Full Product Details