Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21845078-21845614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBTPS2
Entrez Gene ID
51360
Gene Name
membrane bound transcription factor peptidase, site 2
Gene Aliases
BRESEK, IFAP, KFSD, KFSDX, OLMSX, S2P
UniGene
Hs.443490
Gene Chromosome Location
Chr.X: 21839538-21885423 on Build GRCh38
Transcripts
NM_015884.3

SNP Detail

SNP ID Chromosome Location
rs3834687 Chr.X: 21845333-21845335 on Build GRCh38
rs122468177 Chr.X: 21845207-21845207 on Build GRCh38
rs758052304 Chr.X: 21845278-21845278 on Build GRCh38
rs750731000 Chr.X: 21845273-21845273 on Build GRCh38
rs748013480 Chr.X: 21845148-21845148 on Build GRCh38
rs1042400593 Chr.X: 21845416-21845416 on Build GRCh38
rs776688883 Chr.X: 21845127-21845127 on Build GRCh38
rs148895141 Chr.X: 21845354-21845354 on Build GRCh38
rs779941206 Chr.X: 21845220-21845220 on Build GRCh38
rs773969619 Chr.X: 21845312-21845313 on Build GRCh38
rs772245559 Chr.X: 21845524-21845524 on Build GRCh38
rs779572550 Chr.X: 21845334-21845336 on Build GRCh38
rs761369288 Chr.X: 21845313-21845336 on Build GRCh38
rs768048265 Chr.X: 21845121-21845121 on Build GRCh38
rs748147417 Chr.X: 21845315-21845315 on Build GRCh38
rs773100960 Chr.X: 21845165-21845165 on Build GRCh38
rs760781040 Chr.X: 21845371-21845371 on Build GRCh38
rs777721295 Chr.X: 21845326-21845326 on Build GRCh38
rs762761040 Chr.X: 21845286-21845303 on Build GRCh38
rs372340222 Chr.X: 21845402-21845402 on Build GRCh38
rs368198170 Chr.X: 21845363-21845363 on Build GRCh38
rs767007808 Chr.X: 21845328-21845330 on Build GRCh38
rs139868528 Chr.X: 21845196-21845196 on Build GRCh38
rs780952921 Chr.X: 21845391-21845391 on Build GRCh38
rs891193561 Chr.X: 21845480-21845480 on Build GRCh38
rs769567598 Chr.X: 21845321-21845321 on Build GRCh38
rs980525992 Chr.X: 21845132-21845132 on Build GRCh38
rs749021769 Chr.X: 21845330-21845330 on Build GRCh38
rs751265531 Chr.X: 21845417-21845417 on Build GRCh38
rs996600137 Chr.X: 21845409-21845409 on Build GRCh38
rs768337254 Chr.X: 21845285-21845286 on Build GRCh38
rs957684476 Chr.X: 21845502-21845502 on Build GRCh38
rs199974151 Chr.X: 21845236-21845236 on Build GRCh38
rs926217626 Chr.X: 21845421-21845421 on Build GRCh38
rs770841608 Chr.X: 21845353-21845353 on Build GRCh38
rs780537051 Chr.X: 21845303-21845303 on Build GRCh38
rs7063422 Chr.X: 21845285-21845285 on Build GRCh38
rs762823403 Chr.X: 21845491-21845491 on Build GRCh38
rs765667784 Chr.X: 21845401-21845401 on Build GRCh38
rs775161544 Chr.X: 21845271-21845271 on Build GRCh38
rs146730876 Chr.X: 21845253-21845253 on Build GRCh38
rs775712588 Chr.X: 21845205-21845205 on Build GRCh38
rs368866917 Chr.X: 21845206-21845206 on Build GRCh38
rs1023183663 Chr.X: 21845582-21845582 on Build GRCh38
rs764062816 Chr.X: 21845215-21845215 on Build GRCh38
rs759857679 Chr.X: 21845195-21845195 on Build GRCh38
rs983068417 Chr.X: 21845171-21845171 on Build GRCh38
rs762399077 Chr.X: 21845394-21845394 on Build GRCh38
rs768347321 Chr.X: 21845381-21845381 on Build GRCh38
rs376912578 Chr.X: 21845403-21845403 on Build GRCh38
rs368054850 Chr.X: 21845415-21845415 on Build GRCh38
rs951286372 Chr.X: 21845115-21845115 on Build GRCh38
rs899599973 Chr.X: 21845500-21845500 on Build GRCh38
rs769485185 Chr.X: 21845406-21845409 on Build GRCh38
rs375737330 Chr.X: 21845327-21845327 on Build GRCh38
rs749878965 Chr.X: 21845328-21845333 on Build GRCh38
rs746374071 Chr.X: 21845150-21845150 on Build GRCh38
rs7059508 Chr.X: 21845284-21845284 on Build GRCh38
rs7063399 Chr.X: 21845443-21845443 on Build GRCh38
rs754566056 Chr.X: 21845294-21845294 on Build GRCh38
rs1012245595 Chr.X: 21845556-21845556 on Build GRCh38
rs766857614 Chr.X: 21845340-21845345 on Build GRCh38
rs143335256 Chr.X: 21845237-21845237 on Build GRCh38

View Full Product Details