Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 43949110-43949620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDP
Entrez Gene ID
4693
Gene Name
NDP, norrin cystine knot growth factor
Gene Aliases
EVR2, FEVR, ND
UniGene
Hs.522615
Gene Chromosome Location
Chr.X: 43948776-43973675 on Build GRCh38
Transcripts
NM_000266.3

SNP Detail

SNP ID Chromosome Location
rs975929518 Chr.X: 43949249-43949249 on Build GRCh38
rs748106198 Chr.X: 43949279-43949279 on Build GRCh38
rs376833865 Chr.X: 43949374-43949374 on Build GRCh38
rs997377300 Chr.X: 43949538-43949538 on Build GRCh38
rs895271245 Chr.X: 43949307-43949307 on Build GRCh38
rs556578848 Chr.X: 43949318-43949318 on Build GRCh38
rs902736970 Chr.X: 43949343-43949343 on Build GRCh38
rs961255210 Chr.X: 43949501-43949501 on Build GRCh38
rs1017480815 Chr.X: 43949206-43949206 on Build GRCh38
rs1044052942 Chr.X: 43949313-43949313 on Build GRCh38
rs931358877 Chr.X: 43949274-43949274 on Build GRCh38
rs1019457541 Chr.X: 43949474-43949474 on Build GRCh38
rs1011278143 Chr.X: 43949377-43949377 on Build GRCh38
rs188552133 Chr.X: 43949472-43949472 on Build GRCh38
rs1031299822 Chr.X: 43949365-43949365 on Build GRCh38
rs768345763 Chr.X: 43949217-43949217 on Build GRCh38
rs778496777 Chr.X: 43949277-43949277 on Build GRCh38
rs919945868 Chr.X: 43949258-43949258 on Build GRCh38
rs757619775 Chr.X: 43949571-43949571 on Build GRCh38
rs772082194 Chr.X: 43949283-43949283 on Build GRCh38
rs923081497 Chr.X: 43949253-43949253 on Build GRCh38
rs1006878005 Chr.X: 43949481-43949481 on Build GRCh38
rs1057329068 Chr.X: 43949365-43949365 on Build GRCh38
rs764625639 Chr.X: 43949498-43949498 on Build GRCh38
rs900452652 Chr.X: 43949533-43949533 on Build GRCh38
rs746832328 Chr.X: 43949356-43949356 on Build GRCh38
rs947039388 Chr.X: 43949297-43949297 on Build GRCh38
rs762239468 Chr.X: 43949451-43949451 on Build GRCh38
rs938895829 Chr.X: 43949319-43949319 on Build GRCh38
rs143629446 Chr.X: 43949169-43949169 on Build GRCh38
rs914303655 Chr.X: 43949292-43949292 on Build GRCh38
rs1030222239 Chr.X: 43949577-43949577 on Build GRCh38
rs776787976 Chr.X: 43949373-43949373 on Build GRCh38
rs200362031 Chr.X: 43949364-43949365 on Build GRCh38
rs765315655 Chr.X: 43949469-43949470 on Build GRCh38
rs759738711 Chr.X: 43949462-43949462 on Build GRCh38
rs868833919 Chr.X: 43949506-43949506 on Build GRCh38
rs999796727 Chr.X: 43949357-43949357 on Build GRCh38
rs773423440 Chr.X: 43949494-43949494 on Build GRCh38
rs752083237 Chr.X: 43949534-43949534 on Build GRCh38
rs763482992 Chr.X: 43949473-43949473 on Build GRCh38

View Full Product Details