Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 129911059-129911606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP14A
Entrez Gene ID
10813
Gene Name
UTP14A small subunit processome component
Gene Aliases
NYCO16, SDCCAG16, dJ537K23.3
UniGene
Hs.458598
Gene Chromosome Location
Chr.X: 129906121-129929762 on Build GRCh38
Transcripts
NM_001166221.1 NM_006649.3 XM_017029237.1 XM_017029238.1

SNP Detail

SNP ID Chromosome Location
rs192003345 Chr.X: 129911439-129911439 on Build GRCh38
rs182664165 Chr.X: 129911299-129911299 on Build GRCh38
rs200802106 Chr.X: 129911174-129911174 on Build GRCh38
rs745463661 Chr.X: 129911183-129911183 on Build GRCh38
rs774639987 Chr.X: 129911388-129911389 on Build GRCh38
rs56384367 Chr.X: 129911524-129911524 on Build GRCh38
rs1004572347 Chr.X: 129911438-129911438 on Build GRCh38
rs935364552 Chr.X: 129911243-129911243 on Build GRCh38
rs1008350004 Chr.X: 129911447-129911447 on Build GRCh38
rs770567663 Chr.X: 129911207-129911207 on Build GRCh38
rs995613139 Chr.X: 129911535-129911535 on Build GRCh38
rs138075171 Chr.X: 129911311-129911311 on Build GRCh38
rs1027084962 Chr.X: 129911553-129911553 on Build GRCh38
rs946663477 Chr.X: 129911214-129911214 on Build GRCh38
rs190030006 Chr.X: 129911218-129911218 on Build GRCh38
rs758267938 Chr.X: 129911117-129911117 on Build GRCh38
rs990940663 Chr.X: 129911499-129911499 on Build GRCh38
rs960715779 Chr.X: 129911187-129911187 on Build GRCh38
rs1049758625 Chr.X: 129911321-129911321 on Build GRCh38
rs752215679 Chr.X: 129911450-129911450 on Build GRCh38
rs369325912 Chr.X: 129911191-129911191 on Build GRCh38
rs959658910 Chr.X: 129911554-129911554 on Build GRCh38
rs149103112 Chr.X: 129911500-129911500 on Build GRCh38
rs759433618 Chr.X: 129911543-129911543 on Build GRCh38
rs932531545 Chr.X: 129911310-129911310 on Build GRCh38
rs746967917 Chr.X: 129911148-129911148 on Build GRCh38
rs748336798 Chr.X: 129911186-129911190 on Build GRCh38
rs995793109 Chr.X: 129911322-129911322 on Build GRCh38
rs1054399060 Chr.X: 129911277-129911277 on Build GRCh38
rs767168626 Chr.X: 129911320-129911320 on Build GRCh38
rs761826697 Chr.X: 129911481-129911481 on Build GRCh38
rs187748709 Chr.X: 129911381-129911381 on Build GRCh38
rs769542185 Chr.X: 129911189-129911189 on Build GRCh38
rs891250601 Chr.X: 129911300-129911300 on Build GRCh38
rs777810656 Chr.X: 129911143-129911143 on Build GRCh38
rs927725469 Chr.X: 129911095-129911095 on Build GRCh38
rs897355471 Chr.X: 129911511-129911511 on Build GRCh38
rs934468423 Chr.X: 129911244-129911244 on Build GRCh38
rs770670116 Chr.X: 129911172-129911172 on Build GRCh38
rs746399193 Chr.X: 129911231-129911231 on Build GRCh38
rs780967482 Chr.X: 129911179-129911179 on Build GRCh38
rs775695531 Chr.X: 129911306-129911306 on Build GRCh38
rs920163358 Chr.X: 129911146-129911146 on Build GRCh38
rs923961579 Chr.X: 129911222-129911222 on Build GRCh38
rs765265673 Chr.X: 129911571-129911573 on Build GRCh38
rs951593807 Chr.X: 129911428-129911428 on Build GRCh38

View Full Product Details