Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202301086-202301590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGR6
Entrez Gene ID
59352
Gene Name
leucine rich repeat containing G protein-coupled receptor 6
Gene Aliases
GPCR, VTS20631
UniGene
Hs.497402
Gene Chromosome Location
Chr.1: 202193990-202319761 on Build GRCh38
Transcripts
NM_001017403.1 NM_001017404.1 NM_021636.2 XM_005245404.3 XM_011509838.2 XM_011509839.2 XM_011509840.2 XM_011509841.2 XM_011509842.2 XM_011509843.2 XM_011509844.2 XM_011509846.2 XM_017001996.1 XM_017001997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3481662 c.740C>T p.S247L Chr.1: 202301202-202301202 on Build GRCh38
6997800 c.702-4C>A p.? Chr.1: 202301160-202301160 on Build GRCh38
6700781 c.480G>A p.S160S Chr.1: 202301203-202301203 on Build GRCh38
5049619 c.473G>A p.G158E Chr.1: 202301196-202301196 on Build GRCh38
6700779 c.897G>A p.S299S Chr.1: 202301203-202301203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747208810 Chr.1: 202301273-202301273 on Build GRCh38
rs952181247 Chr.1: 202301509-202301509 on Build GRCh38
rs532531013 Chr.1: 202301455-202301455 on Build GRCh38
rs991909319 Chr.1: 202301402-202301402 on Build GRCh38
rs1033421397 Chr.1: 202301394-202301394 on Build GRCh38
rs1033452644 Chr.1: 202301135-202301135 on Build GRCh38
rs767233720 Chr.1: 202301203-202301203 on Build GRCh38
rs769069124 Chr.1: 202301149-202301149 on Build GRCh38
rs1002067067 Chr.1: 202301367-202301367 on Build GRCh38
rs916195112 Chr.1: 202301515-202301515 on Build GRCh38
rs891800667 Chr.1: 202301344-202301344 on Build GRCh38
rs1011235656 Chr.1: 202301423-202301423 on Build GRCh38
rs375015427 Chr.1: 202301131-202301131 on Build GRCh38
rs1009458426 Chr.1: 202301346-202301346 on Build GRCh38
rs756926299 Chr.1: 202301120-202301120 on Build GRCh38
rs1053011785 Chr.1: 202301335-202301335 on Build GRCh38
rs369363878 Chr.1: 202301392-202301392 on Build GRCh38
rs903667424 Chr.1: 202301359-202301359 on Build GRCh38
rs530137879 Chr.1: 202301405-202301405 on Build GRCh38
rs750014816 Chr.1: 202301229-202301229 on Build GRCh38
rs1008047428 Chr.1: 202301443-202301443 on Build GRCh38
rs761470187 Chr.1: 202301202-202301202 on Build GRCh38
rs755672763 Chr.1: 202301241-202301241 on Build GRCh38
rs762967039 Chr.1: 202301175-202301175 on Build GRCh38
rs771190946 Chr.1: 202301282-202301282 on Build GRCh38
rs981667425 Chr.1: 202301547-202301547 on Build GRCh38
rs960482901 Chr.1: 202301396-202301396 on Build GRCh38
rs561107007 Chr.1: 202301404-202301404 on Build GRCh38
rs984790044 Chr.1: 202301512-202301512 on Build GRCh38
rs993995314 Chr.1: 202301468-202301468 on Build GRCh38
rs968299811 Chr.1: 202301112-202301112 on Build GRCh38
rs760867154 Chr.1: 202301166-202301166 on Build GRCh38
rs1028699006 Chr.1: 202301428-202301428 on Build GRCh38
rs1019422076 Chr.1: 202301448-202301448 on Build GRCh38
rs570001486 Chr.1: 202301440-202301440 on Build GRCh38
rs757648365 Chr.1: 202301271-202301271 on Build GRCh38
rs907341634 Chr.1: 202301356-202301356 on Build GRCh38
rs111450303 Chr.1: 202301294-202301294 on Build GRCh38
rs970312264 Chr.1: 202301535-202301535 on Build GRCh38
rs909486233 Chr.1: 202301437-202301437 on Build GRCh38
rs901972478 Chr.1: 202301322-202301322 on Build GRCh38
rs986596296 Chr.1: 202301146-202301146 on Build GRCh38
rs748566693 Chr.1: 202301261-202301261 on Build GRCh38
rs775614276 Chr.1: 202301174-202301174 on Build GRCh38
rs190134164 Chr.1: 202301160-202301160 on Build GRCh38
rs763858097 Chr.1: 202301180-202301180 on Build GRCh38
rs780581243 Chr.1: 202301134-202301134 on Build GRCh38
rs371853249 Chr.1: 202301162-202301162 on Build GRCh38
rs909351497 Chr.1: 202301210-202301210 on Build GRCh38
rs781338281 Chr.1: 202301331-202301331 on Build GRCh38
rs948889422 Chr.1: 202301318-202301318 on Build GRCh38
rs552940487 Chr.1: 202301479-202301479 on Build GRCh38
rs758648270 Chr.1: 202301270-202301270 on Build GRCh38
rs929534261 Chr.1: 202301188-202301188 on Build GRCh38
rs755481197 Chr.1: 202301259-202301259 on Build GRCh38
rs764363529 Chr.1: 202301132-202301132 on Build GRCh38
rs779331350 Chr.1: 202301151-202301151 on Build GRCh38
rs985479292 Chr.1: 202301291-202301291 on Build GRCh38
rs771101043 Chr.1: 202301170-202301170 on Build GRCh38
rs550205377 Chr.1: 202301408-202301408 on Build GRCh38
rs779527512 Chr.1: 202301245-202301245 on Build GRCh38
rs889684012 Chr.1: 202301431-202301431 on Build GRCh38
rs751225109 Chr.1: 202301114-202301114 on Build GRCh38
rs369047348 Chr.1: 202301254-202301254 on Build GRCh38
rs185185831 Chr.1: 202301136-202301136 on Build GRCh38
rs527516922 Chr.1: 202301312-202301312 on Build GRCh38
rs748393899 Chr.1: 202301155-202301155 on Build GRCh38
rs1011911535 Chr.1: 202301115-202301115 on Build GRCh38
rs1025944554 Chr.1: 202301407-202301407 on Build GRCh38
rs566393483 Chr.1: 202301542-202301542 on Build GRCh38
rs1052835499 Chr.1: 202301395-202301395 on Build GRCh38
rs202195510 Chr.1: 202301260-202301260 on Build GRCh38

View Full Product Details