Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209706626-209707126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD11B1
Entrez Gene ID
3290
Gene Name
hydroxysteroid 11-beta dehydrogenase 1
Gene Aliases
11-DH, 11-beta-HSD1, CORTRD2, HDL, HSD11, HSD11B, HSD11L, SDR26C1
UniGene
Hs.195040
Gene Chromosome Location
Chr.1: 209686180-209734950 on Build GRCh38
Transcripts
NM_001206741.1 NM_005525.3 NM_181755.2 NR_134509.1 NR_134510.1

Gene Symbol
LOC101930114
Entrez Gene ID
101930114
Gene Name
uncharacterized LOC101930114
Gene Aliases
-
UniGene
Hs.663934
Gene Chromosome Location
Chr.1: 209661359-209742562 on Build GRCh38
Transcripts
NM_001206741.1 NM_005525.3 NM_181755.2 NR_134509.1 NR_134510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5379528 c.316G>A p.A106T Chr.1: 209706805-209706805 on Build GRCh38
6835421 c.365C>A p.T122N Chr.1: 209706976-209706976 on Build GRCh38
5379529 c.320G>A p.G107E Chr.1: 209706809-209706809 on Build GRCh38
6901216 c.437G>A p.S146N Chr.1: 209707048-209707048 on Build GRCh38
3482892 c.274A>C p.T92P Chr.1: 209706763-209706763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758528944 Chr.1: 209706675-209706675 on Build GRCh38
rs770803034 Chr.1: 209707048-209707048 on Build GRCh38
rs769636594 Chr.1: 209706905-209706905 on Build GRCh38
rs772708921 Chr.1: 209707033-209707033 on Build GRCh38
rs757812747 Chr.1: 209706794-209706794 on Build GRCh38
rs779069856 Chr.1: 209706658-209706658 on Build GRCh38
rs557818199 Chr.1: 209706867-209706867 on Build GRCh38
rs387907168 Chr.1: 209707020-209707020 on Build GRCh38
rs761827471 Chr.1: 209706754-209706754 on Build GRCh38
rs780563325 Chr.1: 209706814-209706814 on Build GRCh38
rs142849016 Chr.1: 209706813-209706813 on Build GRCh38
rs200402963 Chr.1: 209706784-209706784 on Build GRCh38
rs369189519 Chr.1: 209707055-209707055 on Build GRCh38
rs375266448 Chr.1: 209706820-209706820 on Build GRCh38
rs139132286 Chr.1: 209707013-209707013 on Build GRCh38
rs576055469 Chr.1: 209706869-209706869 on Build GRCh38
rs997730423 Chr.1: 209706811-209706811 on Build GRCh38
rs746055273 Chr.1: 209706867-209706868 on Build GRCh38
rs772361610 Chr.1: 209706870-209706871 on Build GRCh38
rs756712282 Chr.1: 209707093-209707093 on Build GRCh38
rs745448889 Chr.1: 209707049-209707049 on Build GRCh38
rs767312206 Chr.1: 209706768-209706768 on Build GRCh38
rs574132787 Chr.1: 209707003-209707003 on Build GRCh38
rs541250922 Chr.1: 209707085-209707085 on Build GRCh38
rs761600033 Chr.1: 209706900-209706901 on Build GRCh38
rs780579721 Chr.1: 209706981-209706981 on Build GRCh38
rs371896771 Chr.1: 209707102-209707102 on Build GRCh38
rs762684208 Chr.1: 209706750-209706750 on Build GRCh38
rs918443806 Chr.1: 209706789-209706789 on Build GRCh38
rs770250927 Chr.1: 209706745-209706745 on Build GRCh38
rs777651281 Chr.1: 209706653-209706653 on Build GRCh38
rs781320092 Chr.1: 209706961-209706961 on Build GRCh38
rs773131369 Chr.1: 209707072-209707077 on Build GRCh38
rs746592856 Chr.1: 209706783-209706783 on Build GRCh38
rs766162814 Chr.1: 209706902-209706902 on Build GRCh38
rs774493277 Chr.1: 209707072-209707072 on Build GRCh38
rs12086634 Chr.1: 209706914-209706914 on Build GRCh38
rs777821799 Chr.1: 209706692-209706692 on Build GRCh38
rs747817801 Chr.1: 209707009-209707009 on Build GRCh38
rs372161445 Chr.1: 209706751-209706751 on Build GRCh38
rs565994081 Chr.1: 209706657-209706657 on Build GRCh38
rs773078969 Chr.1: 209706868-209706868 on Build GRCh38
rs376208100 Chr.1: 209707021-209707021 on Build GRCh38
rs754712904 Chr.1: 209707000-209707000 on Build GRCh38
rs371864479 Chr.1: 209706922-209706922 on Build GRCh38
rs763184496 Chr.1: 209706915-209706915 on Build GRCh38
rs1007465522 Chr.1: 209706866-209706866 on Build GRCh38
rs147231644 Chr.1: 209706713-209706713 on Build GRCh38
rs768366285 Chr.1: 209706825-209706825 on Build GRCh38
rs751543983 Chr.1: 209706793-209706793 on Build GRCh38
rs200360270 Chr.1: 209706993-209706993 on Build GRCh38
rs922227830 Chr.1: 209706703-209706703 on Build GRCh38
rs72215241 Chr.1: 209706871-209706872 on Build GRCh38
rs45487298 Chr.1: 209706873-209706874 on Build GRCh38
rs746362951 Chr.1: 209706809-209706809 on Build GRCh38
rs887769190 Chr.1: 209706776-209706776 on Build GRCh38
rs748344337 Chr.1: 209706662-209706662 on Build GRCh38
rs773938487 Chr.1: 209706829-209706829 on Build GRCh38
rs779086937 Chr.1: 209706863-209706864 on Build GRCh38
rs578197035 Chr.1: 209707001-209707001 on Build GRCh38
rs773933366 Chr.1: 209706753-209706753 on Build GRCh38
rs775334005 Chr.1: 209706747-209706747 on Build GRCh38
rs760430284 Chr.1: 209706778-209706778 on Build GRCh38
rs765057092 Chr.1: 209707065-209707065 on Build GRCh38
rs769440424 Chr.1: 209707053-209707053 on Build GRCh38
rs750265281 Chr.1: 209706775-209706775 on Build GRCh38
rs759366094 Chr.1: 209707064-209707064 on Build GRCh38
rs767501937 Chr.1: 209707074-209707074 on Build GRCh38
rs770582268 Chr.1: 209706857-209706857 on Build GRCh38
rs200635241 Chr.1: 209706972-209706972 on Build GRCh38
rs569365044 Chr.1: 209706701-209706701 on Build GRCh38
rs760654300 Chr.1: 209706892-209706892 on Build GRCh38
rs746176745 Chr.1: 209706737-209706737 on Build GRCh38
rs894035182 Chr.1: 209706834-209706834 on Build GRCh38
rs756622561 Chr.1: 209706979-209706979 on Build GRCh38
rs761192044 Chr.1: 209706845-209706845 on Build GRCh38
rs549535293 Chr.1: 209706697-209706697 on Build GRCh38
rs781588623 Chr.1: 209706799-209706799 on Build GRCh38
rs777840125 Chr.1: 209706790-209706790 on Build GRCh38
rs757328952 Chr.1: 209706952-209706952 on Build GRCh38
rs746957909 Chr.1: 209707046-209707046 on Build GRCh38
rs368758549 Chr.1: 209706723-209706723 on Build GRCh38

View Full Product Details