Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244850452-244850979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPU
Entrez Gene ID
3192
Gene Name
heterogeneous nuclear ribonucleoprotein U
Gene Aliases
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U
UniGene
Hs.106212
Gene Chromosome Location
Chr.1: 244842123-244864720 on Build GRCh38
Transcripts
NM_004501.3 NM_031844.2 XM_017001115.1 XM_017001116.1 XM_017001117.1

SNP Detail

SNP ID Chromosome Location
rs899577344 Chr.1: 244850751-244850757 on Build GRCh38
rs572017176 Chr.1: 244850555-244850555 on Build GRCh38
rs779373510 Chr.1: 244850701-244850701 on Build GRCh38
rs908131410 Chr.1: 244850938-244850938 on Build GRCh38
rs570491531 Chr.1: 244850920-244850920 on Build GRCh38
rs142264012 Chr.1: 244850928-244850928 on Build GRCh38
rs957066376 Chr.1: 244850767-244850767 on Build GRCh38
rs1045876651 Chr.1: 244850955-244850955 on Build GRCh38
rs897832467 Chr.1: 244850867-244850867 on Build GRCh38
rs979751208 Chr.1: 244850482-244850482 on Build GRCh38
rs563017056 Chr.1: 244850731-244850731 on Build GRCh38
rs994794314 Chr.1: 244850908-244850908 on Build GRCh38
rs1054247635 Chr.1: 244850744-244850744 on Build GRCh38
rs78903859 Chr.1: 244850864-244850864 on Build GRCh38
rs961990884 Chr.1: 244850506-244850506 on Build GRCh38
rs771724245 Chr.1: 244850796-244850796 on Build GRCh38
rs886587370 Chr.1: 244850633-244850633 on Build GRCh38
rs1020230643 Chr.1: 244850789-244850789 on Build GRCh38
rs964798991 Chr.1: 244850888-244850888 on Build GRCh38
rs1495946 Chr.1: 244850505-244850505 on Build GRCh38
rs1045427784 Chr.1: 244850682-244850682 on Build GRCh38
rs932199477 Chr.1: 244850780-244850781 on Build GRCh38
rs1010565112 Chr.1: 244850779-244850779 on Build GRCh38
rs187647282 Chr.1: 244850786-244850786 on Build GRCh38
rs192186633 Chr.1: 244850836-244850836 on Build GRCh38
rs1048015654 Chr.1: 244850611-244850611 on Build GRCh38
rs933865112 Chr.1: 244850515-244850515 on Build GRCh38
rs576802086 Chr.1: 244850720-244850720 on Build GRCh38
rs987949582 Chr.1: 244850529-244850529 on Build GRCh38
rs1027655864 Chr.1: 244850545-244850545 on Build GRCh38
rs548106828 Chr.1: 244850879-244850879 on Build GRCh38
rs890607217 Chr.1: 244850826-244850827 on Build GRCh38
rs991722578 Chr.1: 244850590-244850590 on Build GRCh38
rs977340326 Chr.1: 244850699-244850699 on Build GRCh38
rs996747927 Chr.1: 244850478-244850478 on Build GRCh38
rs890690517 Chr.1: 244850483-244850483 on Build GRCh38
rs899381913 Chr.1: 244850604-244850604 on Build GRCh38
rs974859618 Chr.1: 244850907-244850907 on Build GRCh38
rs200921383 Chr.1: 244850484-244850484 on Build GRCh38
rs113603630 Chr.1: 244850822-244850822 on Build GRCh38
rs1024080506 Chr.1: 244850479-244850479 on Build GRCh38
rs969682819 Chr.1: 244850480-244850480 on Build GRCh38
rs1051016009 Chr.1: 244850792-244850792 on Build GRCh38
rs957286091 Chr.1: 244850485-244850485 on Build GRCh38
rs545926214 Chr.1: 244850723-244850723 on Build GRCh38
rs35952469 Chr.1: 244850483-244850484 on Build GRCh38
rs796209846 Chr.1: 244850484-244850485 on Build GRCh38
rs745559022 Chr.1: 244850741-244850741 on Build GRCh38
rs913048175 Chr.1: 244850513-244850513 on Build GRCh38
rs930885074 Chr.1: 244850610-244850610 on Build GRCh38
rs540917842 Chr.1: 244850561-244850561 on Build GRCh38
rs758465529 Chr.1: 244850600-244850601 on Build GRCh38
rs1039056693 Chr.1: 244850602-244850602 on Build GRCh38
rs150871109 Chr.1: 244850952-244850952 on Build GRCh38
rs1048432074 Chr.1: 244850933-244850933 on Build GRCh38
rs749743527 Chr.1: 244850560-244850560 on Build GRCh38
rs548769177 Chr.1: 244850818-244850818 on Build GRCh38
rs745783795 Chr.1: 244850871-244850871 on Build GRCh38
rs935617211 Chr.1: 244850728-244850728 on Build GRCh38
rs1014417180 Chr.1: 244850477-244850477 on Build GRCh38
rs920593092 Chr.1: 244850915-244850915 on Build GRCh38
rs1001114629 Chr.1: 244850717-244850717 on Build GRCh38
rs560158144 Chr.1: 244850641-244850641 on Build GRCh38

View Full Product Details