Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13329495-13329991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPHS1
Entrez Gene ID
22929
Gene Name
selenophosphate synthetase 1
Gene Aliases
SELD, SPS, SPS1
UniGene
Hs.124027
Gene Chromosome Location
Chr.10: 13317428-13348298 on Build GRCh38
Transcripts
NM_001195602.1 NM_001195604.1 NM_012247.4 XM_017015942.1 XM_017015943.1 XM_017015944.1 XM_017015945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4422261 c.651+7A>T p.? Chr.10: 13329691-13329691 on Build GRCh38
4877712 c.586G>A p.V196M Chr.10: 13329763-13329763 on Build GRCh38
4927192 c.602A>G p.K201R Chr.10: 13329747-13329747 on Build GRCh38
916276 c.585C>T p.D195D Chr.10: 13329764-13329764 on Build GRCh38
2023111 c.603A>G p.K201K Chr.10: 13329746-13329746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776288969 Chr.10: 13329673-13329673 on Build GRCh38
rs200522802 Chr.10: 13329669-13329669 on Build GRCh38
rs61851591 Chr.10: 13329855-13329855 on Build GRCh38
rs1049009259 Chr.10: 13329535-13329538 on Build GRCh38
rs753560698 Chr.10: 13329927-13329927 on Build GRCh38
rs116599653 Chr.10: 13329580-13329580 on Build GRCh38
rs749796060 Chr.10: 13329796-13329796 on Build GRCh38
rs768713790 Chr.10: 13329650-13329650 on Build GRCh38
rs916575332 Chr.10: 13329957-13329957 on Build GRCh38
rs1046386389 Chr.10: 13329965-13329965 on Build GRCh38
rs187638640 Chr.10: 13329558-13329558 on Build GRCh38
rs754602419 Chr.10: 13329705-13329705 on Build GRCh38
rs1044761274 Chr.10: 13329924-13329924 on Build GRCh38
rs1008644375 Chr.10: 13329875-13329875 on Build GRCh38
rs147813777 Chr.10: 13329776-13329776 on Build GRCh38
rs34648460 Chr.10: 13329596-13329597 on Build GRCh38
rs374648215 Chr.10: 13329656-13329656 on Build GRCh38
rs991842897 Chr.10: 13329956-13329956 on Build GRCh38
rs190513759 Chr.10: 13329838-13329838 on Build GRCh38
rs957289865 Chr.10: 13329909-13329909 on Build GRCh38
rs1028641096 Chr.10: 13329864-13329864 on Build GRCh38
rs564340233 Chr.10: 13329907-13329907 on Build GRCh38
rs1014666824 Chr.10: 13329595-13329595 on Build GRCh38
rs1053766820 Chr.10: 13329791-13329791 on Build GRCh38
rs537861561 Chr.10: 13329660-13329660 on Build GRCh38
rs761758566 Chr.10: 13329809-13329809 on Build GRCh38
rs955853836 Chr.10: 13329622-13329622 on Build GRCh38
rs995515875 Chr.10: 13329861-13329861 on Build GRCh38
rs767585908 Chr.10: 13329655-13329655 on Build GRCh38
rs763075901 Chr.10: 13329649-13329649 on Build GRCh38
rs779979593 Chr.10: 13329867-13329867 on Build GRCh38
rs1015056607 Chr.10: 13329848-13329848 on Build GRCh38
rs1029089778 Chr.10: 13329572-13329572 on Build GRCh38
rs940160099 Chr.10: 13329874-13329874 on Build GRCh38
rs770128438 Chr.10: 13329780-13329780 on Build GRCh38
rs780601846 Chr.10: 13329794-13329794 on Build GRCh38
rs1044403230 Chr.10: 13329598-13329598 on Build GRCh38
rs767094906 Chr.10: 13329704-13329704 on Build GRCh38
rs746273837 Chr.10: 13329779-13329779 on Build GRCh38
rs1000130285 Chr.10: 13329605-13329605 on Build GRCh38
rs953133572 Chr.10: 13329575-13329575 on Build GRCh38
rs940065264 Chr.10: 13329562-13329562 on Build GRCh38
rs967127533 Chr.10: 13329883-13329883 on Build GRCh38
rs778490277 Chr.10: 13329717-13329717 on Build GRCh38
rs753814691 Chr.10: 13329690-13329690 on Build GRCh38
rs759313613 Chr.10: 13329792-13329796 on Build GRCh38
rs774224958 Chr.10: 13329651-13329651 on Build GRCh38
rs376521416 Chr.10: 13329839-13329839 on Build GRCh38
rs948091287 Chr.10: 13329964-13329964 on Build GRCh38
rs945901928 Chr.10: 13329614-13329614 on Build GRCh38
rs573291979 Chr.10: 13329583-13329583 on Build GRCh38
rs1032783449 Chr.10: 13329616-13329616 on Build GRCh38
rs752093154 Chr.10: 13329754-13329754 on Build GRCh38
rs750322927 Chr.10: 13329810-13329810 on Build GRCh38
rs950169914 Chr.10: 13329914-13329914 on Build GRCh38
rs773282243 Chr.10: 13329824-13329824 on Build GRCh38
rs1018642519 Chr.10: 13329734-13329734 on Build GRCh38
rs766469081 Chr.10: 13329680-13329680 on Build GRCh38
rs905739427 Chr.10: 13329603-13329603 on Build GRCh38
rs774511053 Chr.10: 13329807-13329807 on Build GRCh38
rs977222976 Chr.10: 13329889-13329889 on Build GRCh38
rs772376975 Chr.10: 13329823-13329823 on Build GRCh38
rs906708651 Chr.10: 13329829-13329829 on Build GRCh38
rs761208413 Chr.10: 13329653-13329653 on Build GRCh38
rs1018665248 Chr.10: 13329876-13329876 on Build GRCh38
rs369269469 Chr.10: 13329805-13329805 on Build GRCh38
rs144400182 Chr.10: 13329553-13329553 on Build GRCh38

View Full Product Details