Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13347810-13348350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPHS1
Entrez Gene ID
22929
Gene Name
selenophosphate synthetase 1
Gene Aliases
SELD, SPS, SPS1
UniGene
Hs.124027
Gene Chromosome Location
Chr.10: 13317428-13348298 on Build GRCh38
Transcripts
NM_001195602.1 NM_001195604.1 NM_012247.4 XM_017015944.1

SNP Detail

SNP ID Chromosome Location
rs960342210 Chr.10: 13348073-13348073 on Build GRCh38
rs868491937 Chr.10: 13347849-13347849 on Build GRCh38
rs1049431566 Chr.10: 13348314-13348314 on Build GRCh38
rs1034868708 Chr.10: 13348203-13348203 on Build GRCh38
rs960415904 Chr.10: 13348206-13348206 on Build GRCh38
rs577531809 Chr.10: 13348328-13348328 on Build GRCh38
rs1053120661 Chr.10: 13348151-13348151 on Build GRCh38
rs994778454 Chr.10: 13347987-13347987 on Build GRCh38
rs950534733 Chr.10: 13348028-13348028 on Build GRCh38
rs1039120349 Chr.10: 13348291-13348291 on Build GRCh38
rs950301197 Chr.10: 13347972-13347972 on Build GRCh38
rs979415403 Chr.10: 13348217-13348217 on Build GRCh38
rs1043586846 Chr.10: 13347914-13347914 on Build GRCh38
rs1027867806 Chr.10: 13347988-13347988 on Build GRCh38
rs970678702 Chr.10: 13348113-13348118 on Build GRCh38
rs934704552 Chr.10: 13347943-13347943 on Build GRCh38
rs921925458 Chr.10: 13348306-13348306 on Build GRCh38
rs926110821 Chr.10: 13348121-13348121 on Build GRCh38
rs1023680594 Chr.10: 13348212-13348218 on Build GRCh38
rs886259650 Chr.10: 13348191-13348191 on Build GRCh38
rs573710907 Chr.10: 13348312-13348312 on Build GRCh38
rs912078051 Chr.10: 13348236-13348236 on Build GRCh38
rs1038934182 Chr.10: 13347978-13347978 on Build GRCh38
rs572185148 Chr.10: 13348113-13348115 on Build GRCh38
rs939032105 Chr.10: 13347847-13347847 on Build GRCh38
rs993066236 Chr.10: 13347852-13347852 on Build GRCh38
rs988918174 Chr.10: 13348232-13348232 on Build GRCh38
rs917478401 Chr.10: 13347917-13347917 on Build GRCh38
rs977046134 Chr.10: 13348220-13348220 on Build GRCh38
rs956191093 Chr.10: 13348221-13348221 on Build GRCh38
rs904695737 Chr.10: 13347933-13347933 on Build GRCh38
rs944813050 Chr.10: 13348169-13348169 on Build GRCh38
rs990867177 Chr.10: 13348097-13348097 on Build GRCh38
rs1053168970 Chr.10: 13347947-13347947 on Build GRCh38
rs1049013031 Chr.10: 13348188-13348188 on Build GRCh38
rs1004632971 Chr.10: 13348040-13348040 on Build GRCh38
rs1034795067 Chr.10: 13348043-13348043 on Build GRCh38
rs926238538 Chr.10: 13348219-13348219 on Build GRCh38
rs886331826 Chr.10: 13348321-13348321 on Build GRCh38
rs934863065 Chr.10: 13348141-13348142 on Build GRCh38
rs979037633 Chr.10: 13348113-13348121 on Build GRCh38
rs890695233 Chr.10: 13348177-13348201 on Build GRCh38
rs900396159 Chr.10: 13347979-13347979 on Build GRCh38
rs1004742299 Chr.10: 13348200-13348200 on Build GRCh38
rs916171988 Chr.10: 13347867-13347867 on Build GRCh38
rs900509512 Chr.10: 13348181-13348181 on Build GRCh38
rs890449838 Chr.10: 13347977-13347977 on Build GRCh38
rs944753847 Chr.10: 13348242-13348242 on Build GRCh38
rs967777114 Chr.10: 13348216-13348216 on Build GRCh38
rs866165475 Chr.10: 13348107-13348107 on Build GRCh38
rs912002304 Chr.10: 13348165-13348165 on Build GRCh38
rs995125079 Chr.10: 13348183-13348183 on Build GRCh38
rs949016773 Chr.10: 13347868-13347868 on Build GRCh38

View Full Product Details