Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24983659-24984173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENKUR
Entrez Gene ID
219670
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
C10orf63, CFAP106
UniGene
Hs.534486
Gene Chromosome Location
Chr.10: 24981979-25062279 on Build GRCh38
Transcripts
NM_001270383.1 NM_145010.3 NR_072992.1 NR_072993.1 XM_006717389.1 XM_006717390.1 XM_011519348.1 XM_017015830.1 XM_017016665.1

Gene Symbol
THNSL1
Entrez Gene ID
79896
Gene Name
threonine synthase like 1
Gene Aliases
TSH1
UniGene
Hs.645274
Gene Chromosome Location
Chr.10: 24952319-25026664 on Build GRCh38
Transcripts
NM_001270383.1 NM_145010.3 NR_072992.1 NR_072993.1 XM_006717389.1 XM_006717390.1 XM_011519348.1 XM_017015830.1 XM_017016665.1

SNP Detail

SNP ID Chromosome Location
rs116056901 Chr.10: 24983720-24983720 on Build GRCh38
rs977957516 Chr.10: 24983885-24983885 on Build GRCh38
rs893185895 Chr.10: 24983848-24983848 on Build GRCh38
rs934681701 Chr.10: 24983687-24983687 on Build GRCh38
rs571301061 Chr.10: 24983722-24983723 on Build GRCh38
rs769977492 Chr.10: 24983969-24983969 on Build GRCh38
rs773490803 Chr.10: 24984125-24984125 on Build GRCh38
rs1026966147 Chr.10: 24983926-24983926 on Build GRCh38
rs986667651 Chr.10: 24984064-24984064 on Build GRCh38
rs959150557 Chr.10: 24983790-24983790 on Build GRCh38
rs184470144 Chr.10: 24983993-24983993 on Build GRCh38
rs552204892 Chr.10: 24983948-24983948 on Build GRCh38
rs934584343 Chr.10: 24984090-24984090 on Build GRCh38
rs997666055 Chr.10: 24984111-24984111 on Build GRCh38
rs893211365 Chr.10: 24983693-24983693 on Build GRCh38
rs955365059 Chr.10: 24984062-24984062 on Build GRCh38
rs1054457622 Chr.10: 24984048-24984048 on Build GRCh38
rs917716873 Chr.10: 24983881-24983881 on Build GRCh38
rs138661411 Chr.10: 24984098-24984098 on Build GRCh38
rs549790255 Chr.10: 24983745-24983745 on Build GRCh38
rs950964211 Chr.10: 24983953-24983953 on Build GRCh38
rs984461270 Chr.10: 24983761-24983761 on Build GRCh38
rs190630512 Chr.10: 24984095-24984095 on Build GRCh38
rs990980038 Chr.10: 24983823-24983823 on Build GRCh38
rs537719591 Chr.10: 24984055-24984055 on Build GRCh38
rs934636802 Chr.10: 24983758-24983758 on Build GRCh38
rs144893250 Chr.10: 24983883-24983883 on Build GRCh38
rs551120888 Chr.10: 24983709-24983709 on Build GRCh38
rs1016378014 Chr.10: 24983778-24983778 on Build GRCh38
rs758586830 Chr.10: 24984031-24984031 on Build GRCh38
rs907358762 Chr.10: 24983895-24983895 on Build GRCh38
rs112970617 Chr.10: 24984058-24984058 on Build GRCh38
rs180840520 Chr.10: 24983755-24983755 on Build GRCh38
rs571246623 Chr.10: 24983710-24983710 on Build GRCh38
rs923844872 Chr.10: 24983981-24983981 on Build GRCh38
rs924501087 Chr.10: 24984082-24984082 on Build GRCh38
rs528277041 Chr.10: 24983705-24983705 on Build GRCh38
rs901987981 Chr.10: 24984100-24984100 on Build GRCh38
rs1003073403 Chr.10: 24983920-24983920 on Build GRCh38
rs975559225 Chr.10: 24984047-24984047 on Build GRCh38
rs1054891197 Chr.10: 24983692-24983692 on Build GRCh38
rs1026513675 Chr.10: 24984128-24984128 on Build GRCh38
rs962409426 Chr.10: 24983999-24983999 on Build GRCh38
rs566121529 Chr.10: 24983934-24983934 on Build GRCh38
rs578261997 Chr.10: 24984052-24984052 on Build GRCh38
rs555603285 Chr.10: 24983910-24983910 on Build GRCh38
rs1054396413 Chr.10: 24983759-24983759 on Build GRCh38
rs760342072 Chr.10: 24983976-24983976 on Build GRCh38
rs1051438465 Chr.10: 24983753-24983753 on Build GRCh38
rs79969885 Chr.10: 24983702-24983702 on Build GRCh38

View Full Product Details