Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35638952-35639442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD8
Entrez Gene ID
8325
Gene Name
frizzled class receptor 8
Gene Aliases
FZ-8, hFZ8
UniGene
Hs.302634
Gene Chromosome Location
Chr.10: 35638249-35641434 on Build GRCh38
Transcripts
NM_031866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369008 c.2026C>T p.R676W Chr.10: 35639404-35639404 on Build GRCh38
6041400 c.2021C>A p.T674K Chr.10: 35639409-35639409 on Build GRCh38
6668205 c.2050T>C p.S684P Chr.10: 35639380-35639380 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750174519 Chr.10: 35639352-35639352 on Build GRCh38
rs1043549482 Chr.10: 35639016-35639016 on Build GRCh38
rs771336456 Chr.10: 35639394-35639394 on Build GRCh38
rs902374503 Chr.10: 35639023-35639023 on Build GRCh38
rs962468828 Chr.10: 35639097-35639097 on Build GRCh38
rs780407889 Chr.10: 35639365-35639365 on Build GRCh38
rs937491640 Chr.10: 35639330-35639330 on Build GRCh38
rs770358804 Chr.10: 35639308-35639308 on Build GRCh38
rs749640685 Chr.10: 35639388-35639388 on Build GRCh38
rs528379270 Chr.10: 35639310-35639310 on Build GRCh38
rs753420118 Chr.10: 35639333-35639333 on Build GRCh38
rs923794836 Chr.10: 35639215-35639215 on Build GRCh38
rs1044731853 Chr.10: 35639272-35639272 on Build GRCh38
rs185790987 Chr.10: 35639080-35639080 on Build GRCh38
rs1019206708 Chr.10: 35639135-35639135 on Build GRCh38
rs999442088 Chr.10: 35639050-35639050 on Build GRCh38
rs1046308772 Chr.10: 35639093-35639093 on Build GRCh38
rs765233006 Chr.10: 35639350-35639350 on Build GRCh38
rs625858 Chr.10: 35639296-35639296 on Build GRCh38
rs563454421 Chr.10: 35639115-35639115 on Build GRCh38
rs947413569 Chr.10: 35639382-35639382 on Build GRCh38
rs896416252 Chr.10: 35639256-35639256 on Build GRCh38
rs532319218 Chr.10: 35639116-35639116 on Build GRCh38
rs1056347771 Chr.10: 35639239-35639239 on Build GRCh38
rs773394659 Chr.10: 35639309-35639309 on Build GRCh38
rs974488656 Chr.10: 35638975-35638975 on Build GRCh38
rs912139669 Chr.10: 35639136-35639136 on Build GRCh38
rs764271622 Chr.10: 35639306-35639329 on Build GRCh38
rs748639103 Chr.10: 35639306-35639306 on Build GRCh38
rs758332611 Chr.10: 35639362-35639362 on Build GRCh38
rs903153944 Chr.10: 35639387-35639387 on Build GRCh38
rs768913254 Chr.10: 35639294-35639294 on Build GRCh38
rs747280151 Chr.10: 35639372-35639372 on Build GRCh38
rs983684029 Chr.10: 35639127-35639127 on Build GRCh38
rs965881000 Chr.10: 35639145-35639145 on Build GRCh38
rs111703785 Chr.10: 35639091-35639091 on Build GRCh38
rs773777833 Chr.10: 35639386-35639386 on Build GRCh38
rs770135266 Chr.10: 35639384-35639384 on Build GRCh38
rs755051376 Chr.10: 35639374-35639374 on Build GRCh38
rs775182871 Chr.10: 35639314-35639314 on Build GRCh38
rs1023359085 Chr.10: 35639092-35639092 on Build GRCh38
rs548163640 Chr.10: 35639319-35639319 on Build GRCh38
rs946529264 Chr.10: 35639015-35639015 on Build GRCh38
rs745966124 Chr.10: 35639122-35639122 on Build GRCh38
rs373749029 Chr.10: 35639235-35639235 on Build GRCh38
rs1055768361 Chr.10: 35639099-35639099 on Build GRCh38
rs1011524666 Chr.10: 35639112-35639112 on Build GRCh38
rs41276110 Chr.10: 35639182-35639182 on Build GRCh38
rs946789674 Chr.10: 35639189-35639189 on Build GRCh38
rs1044233398 Chr.10: 35639201-35639201 on Build GRCh38
rs965990779 Chr.10: 35639138-35639138 on Build GRCh38
rs1039062506 Chr.10: 35639003-35639003 on Build GRCh38
rs974734497 Chr.10: 35639180-35639180 on Build GRCh38
rs763828550 Chr.10: 35639406-35639406 on Build GRCh38
rs775092983 Chr.10: 35639396-35639396 on Build GRCh38
rs922034163 Chr.10: 35639011-35639011 on Build GRCh38
rs757092983 Chr.10: 35639338-35639338 on Build GRCh38
rs748276505 Chr.10: 35639383-35639383 on Build GRCh38
rs935163293 Chr.10: 35639232-35639232 on Build GRCh38
rs1056823052 Chr.10: 35639059-35639059 on Build GRCh38
rs921613794 Chr.10: 35638996-35638996 on Build GRCh38
rs896114064 Chr.10: 35639069-35639069 on Build GRCh38
rs114001774 Chr.10: 35639104-35639104 on Build GRCh38
rs901679379 Chr.10: 35639422-35639422 on Build GRCh38
rs1011893967 Chr.10: 35639257-35639257 on Build GRCh38
rs760281664 Chr.10: 35639397-35639397 on Build GRCh38
rs951723836 Chr.10: 35639269-35639269 on Build GRCh38
rs937442687 Chr.10: 35639099-35639099 on Build GRCh38
rs775945052 Chr.10: 35639423-35639423 on Build GRCh38
rs568002712 Chr.10: 35639375-35639375 on Build GRCh38

View Full Product Details