Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91941940-91942487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
UniGene
Hs.500526
Gene Chromosome Location
Chr.10: 91923769-92030998 on Build GRCh38
Transcripts
NM_003972.2 XM_011540326.1 XM_011540329.2 XM_017016877.1 XM_017016878.1 XM_017016879.1 XM_017016880.1 XM_017016881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4484770 c.284C>T p.S95L Chr.10: 91942452-91942452 on Build GRCh38
1969437 c.304A>C p.N102H Chr.10: 91942472-91942472 on Build GRCh38
5440821 c.265G>A p.E89K Chr.10: 91942433-91942433 on Build GRCh38
7001379 c.254-2_254-1ins11 p.? Chr.10: 91942420-91942421 on Build GRCh38
279213 c.299G>A p.R100Q Chr.10: 91942467-91942467 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs868529651 Chr.10: 91942298-91942298 on Build GRCh38
rs183148676 Chr.10: 91942357-91942357 on Build GRCh38
rs572525363 Chr.10: 91941972-91941972 on Build GRCh38
rs559336098 Chr.10: 91941967-91941967 on Build GRCh38
rs567136049 Chr.10: 91942276-91942276 on Build GRCh38
rs561478074 Chr.10: 91942093-91942093 on Build GRCh38
rs199538823 Chr.10: 91942442-91942442 on Build GRCh38
rs985745567 Chr.10: 91942011-91942011 on Build GRCh38
rs749331695 Chr.10: 91942387-91942387 on Build GRCh38
rs765617555 Chr.10: 91942424-91942424 on Build GRCh38
rs1019813743 Chr.10: 91942361-91942362 on Build GRCh38
rs746713075 Chr.10: 91942305-91942310 on Build GRCh38
rs551707449 Chr.10: 91942138-91942138 on Build GRCh38
rs775373054 Chr.10: 91942436-91942436 on Build GRCh38
rs950040547 Chr.10: 91942433-91942433 on Build GRCh38
rs989072595 Chr.10: 91942371-91942371 on Build GRCh38
rs12569736 Chr.10: 91942216-91942216 on Build GRCh38
rs936016991 Chr.10: 91942084-91942084 on Build GRCh38
rs113869491 Chr.10: 91942299-91942304 on Build GRCh38
rs771547619 Chr.10: 91942298-91942303 on Build GRCh38
rs398114637 Chr.10: 91942295-91942296 on Build GRCh38
rs528432975 Chr.10: 91941973-91941973 on Build GRCh38
rs922789647 Chr.10: 91941981-91941981 on Build GRCh38
rs777835824 Chr.10: 91942382-91942382 on Build GRCh38
rs879074023 Chr.10: 91942299-91942302 on Build GRCh38
rs766999909 Chr.10: 91942134-91942134 on Build GRCh38
rs764051948 Chr.10: 91942450-91942450 on Build GRCh38
rs1015044492 Chr.10: 91942224-91942224 on Build GRCh38
rs1020921732 Chr.10: 91942060-91942073 on Build GRCh38
rs765783198 Chr.10: 91942299-91942310 on Build GRCh38
rs954238738 Chr.10: 91941995-91941995 on Build GRCh38
rs868633654 Chr.10: 91942431-91942431 on Build GRCh38
rs58443276 Chr.10: 91942326-91942331 on Build GRCh38
rs985995809 Chr.10: 91942340-91942340 on Build GRCh38
rs71025373 Chr.10: 91942318-91942319 on Build GRCh38
rs547296249 Chr.10: 91942268-91942268 on Build GRCh38
rs533320247 Chr.10: 91942255-91942255 on Build GRCh38
rs746045054 Chr.10: 91942412-91942412 on Build GRCh38
rs761384650 Chr.10: 91942111-91942111 on Build GRCh38
rs148405175 Chr.10: 91942453-91942453 on Build GRCh38
rs1011701879 Chr.10: 91942332-91942332 on Build GRCh38
rs71484276 Chr.10: 91942331-91942332 on Build GRCh38
rs550936601 Chr.10: 91942209-91942209 on Build GRCh38
rs776416061 Chr.10: 91942310-91942311 on Build GRCh38
rs1001294683 Chr.10: 91942194-91942194 on Build GRCh38
rs965684370 Chr.10: 91942250-91942250 on Build GRCh38
rs372700430 Chr.10: 91942462-91942462 on Build GRCh38
rs774701108 Chr.10: 91942408-91942408 on Build GRCh38
rs146552699 Chr.10: 91942355-91942355 on Build GRCh38
rs1045773486 Chr.10: 91942145-91942145 on Build GRCh38
rs765550232 Chr.10: 91941992-91941992 on Build GRCh38
rs1048989740 Chr.10: 91942202-91942202 on Build GRCh38
rs67087052 Chr.10: 91942285-91942286 on Build GRCh38
rs772039529 Chr.10: 91942413-91942413 on Build GRCh38
rs61669989 Chr.10: 91942324-91942331 on Build GRCh38
rs927358272 Chr.10: 91942460-91942460 on Build GRCh38
rs1028720363 Chr.10: 91942285-91942285 on Build GRCh38
rs776917610 Chr.10: 91942296-91942307 on Build GRCh38
rs374249746 Chr.10: 91942388-91942388 on Build GRCh38
rs1011737650 Chr.10: 91941974-91941974 on Build GRCh38
rs954213985 Chr.10: 91942324-91942324 on Build GRCh38
rs201728312 Chr.10: 91942304-91942304 on Build GRCh38
rs773985626 Chr.10: 91942043-91942043 on Build GRCh38
rs1053492620 Chr.10: 91942089-91942089 on Build GRCh38
rs938313554 Chr.10: 91942286-91942288 on Build GRCh38
rs905733291 Chr.10: 91942185-91942185 on Build GRCh38
rs972740539 Chr.10: 91942064-91942064 on Build GRCh38
rs142847767 Chr.10: 91942164-91942164 on Build GRCh38
rs541388322 Chr.10: 91942012-91942012 on Build GRCh38
rs986079201 Chr.10: 91942001-91942001 on Build GRCh38

View Full Product Details