Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103277058-103277554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INA
Entrez Gene ID
9118
Gene Name
internexin neuronal intermediate filament protein alpha
Gene Aliases
NEF5, NF-66, TXBP-1
UniGene
Hs.500916
Gene Chromosome Location
Chr.10: 103277163-103290351 on Build GRCh38
Transcripts
NM_032727.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4614101 c.29_30insCTC p.S14_Y15insS Chr.10: 103277240-103277241 on Build GRCh38
4591937 c.76T>C p.S26P Chr.10: 103277287-103277287 on Build GRCh38
7182668 c.234C>T p.S78S Chr.10: 103277445-103277445 on Build GRCh38
6349168 c.131C>A p.S44Y Chr.10: 103277342-103277342 on Build GRCh38
683496 c.330G>A p.E110E Chr.10: 103277541-103277541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781371956 Chr.10: 103277179-103277179 on Build GRCh38
rs748045007 Chr.10: 103277358-103277372 on Build GRCh38
rs754148970 Chr.10: 103277426-103277426 on Build GRCh38
rs201476129 Chr.10: 103277442-103277442 on Build GRCh38
rs761768626 Chr.10: 103277414-103277414 on Build GRCh38
rs761309952 Chr.10: 103277198-103277198 on Build GRCh38
rs373787135 Chr.10: 103277316-103277316 on Build GRCh38
rs949512541 Chr.10: 103277121-103277121 on Build GRCh38
rs1017858812 Chr.10: 103277477-103277477 on Build GRCh38
rs993408128 Chr.10: 103277159-103277159 on Build GRCh38
rs1017490165 Chr.10: 103277145-103277145 on Build GRCh38
rs867625007 Chr.10: 103277090-103277090 on Build GRCh38
rs763225246 Chr.10: 103277331-103277331 on Build GRCh38
rs762048241 Chr.10: 103277467-103277467 on Build GRCh38
rs754749718 Chr.10: 103277241-103277243 on Build GRCh38
rs994747200 Chr.10: 103277512-103277512 on Build GRCh38
rs746234662 Chr.10: 103277185-103277185 on Build GRCh38
rs770210578 Chr.10: 103277262-103277262 on Build GRCh38
rs770511692 Chr.10: 103277375-103277375 on Build GRCh38
rs199785374 Chr.10: 103277216-103277216 on Build GRCh38
rs765170021 Chr.10: 103277326-103277326 on Build GRCh38
rs760759982 Chr.10: 103277482-103277482 on Build GRCh38
rs753003968 Chr.10: 103277222-103277222 on Build GRCh38
rs536679475 Chr.10: 103277235-103277235 on Build GRCh38
rs76287008 Chr.10: 103277150-103277150 on Build GRCh38
rs529183084 Chr.10: 103277465-103277465 on Build GRCh38
rs1017410186 Chr.10: 103277200-103277200 on Build GRCh38
rs769879980 Chr.10: 103277394-103277394 on Build GRCh38
rs780198749 Chr.10: 103277463-103277463 on Build GRCh38
rs760483767 Chr.10: 103277206-103277206 on Build GRCh38
rs754439685 Chr.10: 103277502-103277502 on Build GRCh38
rs780806405 Chr.10: 103277451-103277451 on Build GRCh38
rs780348356 Chr.10: 103277359-103277359 on Build GRCh38
rs547316582 Chr.10: 103277083-103277083 on Build GRCh38
rs777201376 Chr.10: 103277409-103277410 on Build GRCh38
rs773179927 Chr.10: 103277201-103277201 on Build GRCh38
rs778507741 Chr.10: 103277279-103277279 on Build GRCh38
rs775444664 Chr.10: 103277330-103277330 on Build GRCh38
rs773213037 Chr.10: 103277480-103277480 on Build GRCh38
rs200553495 Chr.10: 103277342-103277342 on Build GRCh38
rs778308103 Chr.10: 103277172-103277172 on Build GRCh38
rs890313738 Chr.10: 103277177-103277177 on Build GRCh38
rs765584342 Chr.10: 103277508-103277508 on Build GRCh38
rs771781429 Chr.10: 103277292-103277292 on Build GRCh38
rs980097300 Chr.10: 103277398-103277398 on Build GRCh38
rs980512655 Chr.10: 103277178-103277178 on Build GRCh38
rs542471919 Chr.10: 103277346-103277346 on Build GRCh38
rs370048294 Chr.10: 103277233-103277233 on Build GRCh38
rs898863226 Chr.10: 103277490-103277490 on Build GRCh38
rs746490997 Chr.10: 103277368-103277368 on Build GRCh38
rs767635416 Chr.10: 103277418-103277418 on Build GRCh38
rs747848985 Chr.10: 103277288-103277288 on Build GRCh38
rs149171677 Chr.10: 103277462-103277462 on Build GRCh38
rs772560394 Chr.10: 103277294-103277294 on Build GRCh38
rs756450190 Chr.10: 103277344-103277344 on Build GRCh38
rs1029936087 Chr.10: 103277124-103277124 on Build GRCh38
rs565756954 Chr.10: 103277523-103277523 on Build GRCh38
rs890210175 Chr.10: 103277282-103277282 on Build GRCh38
rs745374711 Chr.10: 103277457-103277457 on Build GRCh38
rs747515070 Chr.10: 103277175-103277175 on Build GRCh38
rs554077468 Chr.10: 103277357-103277357 on Build GRCh38
rs948712787 Chr.10: 103277173-103277173 on Build GRCh38
rs772313683 Chr.10: 103277478-103277478 on Build GRCh38
rs1026167399 Chr.10: 103277155-103277155 on Build GRCh38
rs992666702 Chr.10: 103277385-103277385 on Build GRCh38
rs1063454 Chr.10: 103277400-103277400 on Build GRCh38
rs189225017 Chr.10: 103277430-103277430 on Build GRCh38
rs757638294 Chr.10: 103277227-103277227 on Build GRCh38
rs963630167 Chr.10: 103277149-103277149 on Build GRCh38
rs776772343 Chr.10: 103277320-103277320 on Build GRCh38
rs2234974 Chr.10: 103277199-103277199 on Build GRCh38
rs759421583 Chr.10: 103277213-103277213 on Build GRCh38
rs569324079 Chr.10: 103277220-103277220 on Build GRCh38
rs201647490 Chr.10: 103277290-103277290 on Build GRCh38
rs771640176 Chr.10: 103277388-103277389 on Build GRCh38
rs746537046 Chr.10: 103277315-103277315 on Build GRCh38
rs766862746 Chr.10: 103277499-103277499 on Build GRCh38
rs766879145 Chr.10: 103277424-103277424 on Build GRCh38
rs372908458 Chr.10: 103277388-103277388 on Build GRCh38
rs1051210597 Chr.10: 103277431-103277431 on Build GRCh38
rs539160986 Chr.10: 103277136-103277136 on Build GRCh38
rs1032408678 Chr.10: 103277496-103277496 on Build GRCh38
rs751535812 Chr.10: 103277334-103277334 on Build GRCh38
rs377641394 Chr.10: 103277226-103277226 on Build GRCh38
rs142360479 Chr.10: 103277466-103277466 on Build GRCh38
rs755431963 Chr.10: 103277427-103277427 on Build GRCh38
rs775356655 Chr.10: 103277395-103277395 on Build GRCh38
rs752807908 Chr.10: 103277171-103277171 on Build GRCh38
rs750968378 Chr.10: 103277343-103277343 on Build GRCh38
rs779285630 Chr.10: 103277167-103277167 on Build GRCh38
rs572573223 Chr.10: 103277362-103277362 on Build GRCh38
rs531818807 Chr.10: 103277411-103277411 on Build GRCh38
rs756576893 Chr.10: 103277422-103277422 on Build GRCh38
rs764379458 Chr.10: 103277332-103277332 on Build GRCh38
rs757924398 Chr.10: 103277364-103277364 on Build GRCh38
rs1063455 Chr.10: 103277485-103277485 on Build GRCh38
rs554942308 Chr.10: 103277236-103277236 on Build GRCh38
rs945125965 Chr.10: 103277250-103277250 on Build GRCh38
rs764208250 Chr.10: 103277410-103277410 on Build GRCh38
rs752601985 Chr.10: 103277176-103277176 on Build GRCh38
rs376203081 Chr.10: 103277337-103277337 on Build GRCh38
rs914110724 Chr.10: 103277110-103277110 on Build GRCh38
rs759677028 Chr.10: 103277240-103277241 on Build GRCh38
rs753032949 Chr.10: 103277510-103277510 on Build GRCh38
rs1051495557 Chr.10: 103277252-103277252 on Build GRCh38
rs370418792 Chr.10: 103277423-103277423 on Build GRCh38
rs958016892 Chr.10: 103277295-103277295 on Build GRCh38
rs756880231 Chr.10: 103277444-103277444 on Build GRCh38
rs749939557 Chr.10: 103277530-103277530 on Build GRCh38
rs753513355 Chr.10: 103277210-103277210 on Build GRCh38
rs892186399 Chr.10: 103277103-103277103 on Build GRCh38
rs565827144 Chr.10: 103277132-103277132 on Build GRCh38
rs768238921 Chr.10: 103277197-103277197 on Build GRCh38
rs756900391 Chr.10: 103277521-103277521 on Build GRCh38
rs1009301929 Chr.10: 103277104-103277104 on Build GRCh38
rs759370417 Chr.10: 103277322-103277322 on Build GRCh38
rs1007881360 Chr.10: 103277139-103277139 on Build GRCh38
rs755214403 Chr.10: 103277166-103277166 on Build GRCh38
rs950758638 Chr.10: 103277163-103277163 on Build GRCh38
rs750525367 Chr.10: 103277421-103277421 on Build GRCh38
rs758527412 Chr.10: 103277225-103277225 on Build GRCh38
rs202015210 Chr.10: 103277376-103277376 on Build GRCh38
rs376053740 Chr.10: 103277397-103277397 on Build GRCh38

View Full Product Details