Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123162493-123162991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB3
Entrez Gene ID
9184
Gene Name
BUB3, mitotic checkpoint protein
Gene Aliases
BUB3L, hBUB3
UniGene
Hs.418533
Gene Chromosome Location
Chr.10: 123154244-123165370 on Build GRCh38
Transcripts
NM_001007793.2 NM_004725.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
915761 c.941G>A p.R314H Chr.10: 123162798-123162798 on Build GRCh38
1321482 c.971+3G>A p.? Chr.10: 123162831-123162831 on Build GRCh38
915759 c.821G>T p.R274L Chr.10: 123162678-123162678 on Build GRCh38
4969269 c.940C>T p.R314C Chr.10: 123162797-123162797 on Build GRCh38
915757 c.820C>T p.R274W Chr.10: 123162677-123162677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7920684 Chr.10: 123162854-123162854 on Build GRCh38
rs762530204 Chr.10: 123162706-123162706 on Build GRCh38
rs745672644 Chr.10: 123162761-123162761 on Build GRCh38
rs201610216 Chr.10: 123162723-123162723 on Build GRCh38
rs979739422 Chr.10: 123162942-123162942 on Build GRCh38
rs34457395 Chr.10: 123162856-123162857 on Build GRCh38
rs755147507 Chr.10: 123162742-123162742 on Build GRCh38
rs916221744 Chr.10: 123162557-123162557 on Build GRCh38
rs769341898 Chr.10: 123162542-123162542 on Build GRCh38
rs146673157 Chr.10: 123162873-123162873 on Build GRCh38
rs770812400 Chr.10: 123162686-123162686 on Build GRCh38
rs1004863018 Chr.10: 123162677-123162677 on Build GRCh38
rs925971852 Chr.10: 123162944-123162944 on Build GRCh38
rs935957888 Chr.10: 123162954-123162954 on Build GRCh38
rs762303389 Chr.10: 123162593-123162593 on Build GRCh38
rs751945794 Chr.10: 123162588-123162588 on Build GRCh38
rs188414049 Chr.10: 123162599-123162599 on Build GRCh38
rs764676872 Chr.10: 123162581-123162581 on Build GRCh38
rs991859630 Chr.10: 123162900-123162900 on Build GRCh38
rs781106915 Chr.10: 123162688-123162688 on Build GRCh38
rs760984169 Chr.10: 123162907-123162907 on Build GRCh38
rs542101472 Chr.10: 123162772-123162772 on Build GRCh38
rs777836898 Chr.10: 123162673-123162673 on Build GRCh38
rs898150775 Chr.10: 123162732-123162732 on Build GRCh38
rs762320809 Chr.10: 123162578-123162578 on Build GRCh38
rs751559876 Chr.10: 123162746-123162746 on Build GRCh38
rs775172244 Chr.10: 123162567-123162567 on Build GRCh38
rs747654646 Chr.10: 123162802-123162802 on Build GRCh38
rs200887768 Chr.10: 123162565-123162565 on Build GRCh38
rs916356143 Chr.10: 123162925-123162925 on Build GRCh38
rs1038033353 Chr.10: 123162931-123162931 on Build GRCh38
rs780205740 Chr.10: 123162642-123162642 on Build GRCh38
rs112586035 Chr.10: 123162617-123162617 on Build GRCh38
rs947722283 Chr.10: 123162926-123162926 on Build GRCh38
rs754952400 Chr.10: 123162670-123162670 on Build GRCh38
rs994006181 Chr.10: 123162693-123162693 on Build GRCh38
rs887330972 Chr.10: 123162664-123162664 on Build GRCh38
rs750692465 Chr.10: 123162602-123162602 on Build GRCh38
rs199889334 Chr.10: 123162627-123162627 on Build GRCh38
rs761029872 Chr.10: 123162855-123162855 on Build GRCh38
rs866193666 Chr.10: 123162659-123162659 on Build GRCh38
rs1046405277 Chr.10: 123162663-123162663 on Build GRCh38
rs757176984 Chr.10: 123162747-123162747 on Build GRCh38
rs755794503 Chr.10: 123162857-123162857 on Build GRCh38
rs552637983 Chr.10: 123162967-123162967 on Build GRCh38
rs776963503 Chr.10: 123162864-123162864 on Build GRCh38
rs576902773 Chr.10: 123162652-123162652 on Build GRCh38
rs140277834 Chr.10: 123162758-123162758 on Build GRCh38
rs919608964 Chr.10: 123162965-123162965 on Build GRCh38
rs753933712 Chr.10: 123162662-123162662 on Build GRCh38
rs769592647 Chr.10: 123162700-123162700 on Build GRCh38
rs746909888 Chr.10: 123162685-123162685 on Build GRCh38
rs750814689 Chr.10: 123162738-123162738 on Build GRCh38
rs202197920 Chr.10: 123162709-123162709 on Build GRCh38
rs745619504 Chr.10: 123162694-123162694 on Build GRCh38
rs960250950 Chr.10: 123162886-123162886 on Build GRCh38
rs1802255 Chr.10: 123162731-123162731 on Build GRCh38
rs146645506 Chr.10: 123162768-123162768 on Build GRCh38
rs766719690 Chr.10: 123162860-123162860 on Build GRCh38
rs769647696 Chr.10: 123162766-123162766 on Build GRCh38
rs775227343 Chr.10: 123162702-123162702 on Build GRCh38
rs868083882 Chr.10: 123162644-123162644 on Build GRCh38
rs201222585 Chr.10: 123162715-123162715 on Build GRCh38
rs151202304 Chr.10: 123162726-123162726 on Build GRCh38
rs762924864 Chr.10: 123162787-123162787 on Build GRCh38
rs767908023 Chr.10: 123162737-123162737 on Build GRCh38
rs372686671 Chr.10: 123162739-123162739 on Build GRCh38
rs756296981 Chr.10: 123162635-123162635 on Build GRCh38
rs772586328 Chr.10: 123162813-123162813 on Build GRCh38
rs778839112 Chr.10: 123162745-123162745 on Build GRCh38
rs1011607443 Chr.10: 123162788-123162788 on Build GRCh38
rs199696433 Chr.10: 123162811-123162811 on Build GRCh38
rs183700875 Chr.10: 123162518-123162518 on Build GRCh38
rs769473400 Chr.10: 123162566-123162566 on Build GRCh38
rs759649716 Chr.10: 123162867-123162867 on Build GRCh38

View Full Product Details