Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125730172-125730709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDRF1
Entrez Gene ID
26098
Gene Name
erythroid differentiation regulatory factor 1
Gene Aliases
C10orf137
UniGene
Hs.468688
Gene Chromosome Location
Chr.10: 125719515-125764143 on Build GRCh38
Transcripts
NM_001202438.1 NM_015608.2 NR_110857.1 NR_110858.1 NR_110859.1 XM_011539622.1 XM_017016066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613847 c.1079G>A p.C360Y Chr.10: 125730350-125730350 on Build GRCh38
1492177 c.935A>T p.N312I Chr.10: 125730308-125730308 on Build GRCh38
1492178 c.930A>T p.P310P Chr.10: 125730303-125730303 on Build GRCh38
4012480 c.1080C>T p.C360C Chr.10: 125730351-125730351 on Build GRCh38
5794790 c.1017-3T>C p.? Chr.10: 125730285-125730285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919530513 Chr.10: 125730553-125730553 on Build GRCh38
rs981207207 Chr.10: 125730231-125730231 on Build GRCh38
rs753983724 Chr.10: 125730363-125730363 on Build GRCh38
rs7896701 Chr.10: 125730575-125730575 on Build GRCh38
rs774979933 Chr.10: 125730226-125730226 on Build GRCh38
rs992711101 Chr.10: 125730357-125730357 on Build GRCh38
rs6597725 Chr.10: 125730668-125730668 on Build GRCh38
rs561245323 Chr.10: 125730680-125730680 on Build GRCh38
rs767994005 Chr.10: 125730261-125730261 on Build GRCh38
rs1039383150 Chr.10: 125730492-125730492 on Build GRCh38
rs374511897 Chr.10: 125730440-125730440 on Build GRCh38
rs756692130 Chr.10: 125730232-125730235 on Build GRCh38
rs188234716 Chr.10: 125730485-125730485 on Build GRCh38
rs1002839148 Chr.10: 125730472-125730472 on Build GRCh38
rs751493979 Chr.10: 125730414-125730414 on Build GRCh38
rs568975186 Chr.10: 125730425-125730426 on Build GRCh38
rs534409384 Chr.10: 125730205-125730205 on Build GRCh38
rs112992746 Chr.10: 125730381-125730381 on Build GRCh38
rs574798277 Chr.10: 125730591-125730591 on Build GRCh38
rs775359851 Chr.10: 125730587-125730587 on Build GRCh38
rs554398034 Chr.10: 125730672-125730674 on Build GRCh38
rs909653200 Chr.10: 125730473-125730473 on Build GRCh38
rs373293309 Chr.10: 125730582-125730582 on Build GRCh38
rs934251060 Chr.10: 125730542-125730542 on Build GRCh38
rs754989284 Chr.10: 125730387-125730387 on Build GRCh38
rs558774674 Chr.10: 125730353-125730353 on Build GRCh38
rs972700813 Chr.10: 125730541-125730541 on Build GRCh38
rs537983377 Chr.10: 125730574-125730574 on Build GRCh38
rs777743988 Chr.10: 125730412-125730412 on Build GRCh38
rs903156317 Chr.10: 125730465-125730465 on Build GRCh38
rs757118280 Chr.10: 125730436-125730436 on Build GRCh38
rs762331061 Chr.10: 125730251-125730251 on Build GRCh38
rs918427853 Chr.10: 125730360-125730360 on Build GRCh38
rs750751043 Chr.10: 125730268-125730268 on Build GRCh38
rs929776777 Chr.10: 125730415-125730415 on Build GRCh38
rs370679928 Chr.10: 125730429-125730430 on Build GRCh38
rs1022446697 Chr.10: 125730630-125730630 on Build GRCh38
rs914367932 Chr.10: 125730639-125730639 on Build GRCh38
rs958780930 Chr.10: 125730479-125730479 on Build GRCh38
rs764705333 Chr.10: 125730225-125730225 on Build GRCh38
rs900839182 Chr.10: 125730525-125730525 on Build GRCh38
rs1002286995 Chr.10: 125730681-125730681 on Build GRCh38
rs536320298 Chr.10: 125730483-125730484 on Build GRCh38
rs1011891726 Chr.10: 125730356-125730356 on Build GRCh38
rs942417417 Chr.10: 125730620-125730620 on Build GRCh38

View Full Product Details