Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62839031-62839528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR74
Entrez Gene ID
54663
Gene Name
WD repeat domain 74
Gene Aliases
-
UniGene
Hs.654620
Gene Chromosome Location
Chr.11: 62832905-62841809 on Build GRCh38
Transcripts
NM_001307977.1 NM_018093.3 XM_005274055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6822184 c.216C>A p.T72T Chr.11: 62839191-62839191 on Build GRCh38
6133995 c.140C>T p.A47V Chr.11: 62839353-62839353 on Build GRCh38
3986310 c.60G>C p.L20F Chr.11: 62839511-62839511 on Build GRCh38
6069824 c.279C>T p.L93L Chr.11: 62839128-62839128 on Build GRCh38
6822178 c.93G>A p.A31A Chr.11: 62839400-62839400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761501075 Chr.11: 62839242-62839242 on Build GRCh38
rs747434607 Chr.11: 62839088-62839094 on Build GRCh38
rs768820241 Chr.11: 62839203-62839203 on Build GRCh38
rs761398726 Chr.11: 62839462-62839462 on Build GRCh38
rs771377225 Chr.11: 62839217-62839217 on Build GRCh38
rs779145574 Chr.11: 62839385-62839385 on Build GRCh38
rs752364338 Chr.11: 62839142-62839142 on Build GRCh38
rs767849189 Chr.11: 62839325-62839325 on Build GRCh38
rs915506236 Chr.11: 62839115-62839115 on Build GRCh38
rs761711065 Chr.11: 62839328-62839328 on Build GRCh38
rs758316576 Chr.11: 62839150-62839150 on Build GRCh38
rs771719739 Chr.11: 62839467-62839467 on Build GRCh38
rs900934378 Chr.11: 62839281-62839281 on Build GRCh38
rs772893432 Chr.11: 62839479-62839479 on Build GRCh38
rs965722772 Chr.11: 62839453-62839453 on Build GRCh38
rs764002716 Chr.11: 62839151-62839151 on Build GRCh38
rs765005824 Chr.11: 62839138-62839138 on Build GRCh38
rs774143691 Chr.11: 62839326-62839326 on Build GRCh38
rs748450325 Chr.11: 62839212-62839212 on Build GRCh38
rs750187611 Chr.11: 62839332-62839332 on Build GRCh38
rs752597115 Chr.11: 62839069-62839069 on Build GRCh38
rs539200289 Chr.11: 62839291-62839291 on Build GRCh38
rs752782215 Chr.11: 62839500-62839500 on Build GRCh38
rs780134398 Chr.11: 62839272-62839272 on Build GRCh38
rs926413236 Chr.11: 62839165-62839165 on Build GRCh38
rs752838865 Chr.11: 62839387-62839387 on Build GRCh38
rs745871770 Chr.11: 62839082-62839082 on Build GRCh38
rs759125069 Chr.11: 62839219-62839219 on Build GRCh38
rs770070302 Chr.11: 62839083-62839083 on Build GRCh38
rs199550021 Chr.11: 62839235-62839235 on Build GRCh38
rs556883805 Chr.11: 62839207-62839207 on Build GRCh38
rs567273843 Chr.11: 62839152-62839152 on Build GRCh38
rs750824540 Chr.11: 62839369-62839369 on Build GRCh38
rs764320206 Chr.11: 62839506-62839506 on Build GRCh38
rs758596641 Chr.11: 62839401-62839401 on Build GRCh38
rs780935672 Chr.11: 62839164-62839164 on Build GRCh38
rs931263325 Chr.11: 62839194-62839194 on Build GRCh38
rs763773336 Chr.11: 62839237-62839237 on Build GRCh38
rs768253603 Chr.11: 62839459-62839459 on Build GRCh38
rs774105076 Chr.11: 62839460-62839460 on Build GRCh38
rs369248508 Chr.11: 62839073-62839073 on Build GRCh38
rs760565116 Chr.11: 62839347-62839347 on Build GRCh38
rs781761716 Chr.11: 62839441-62839441 on Build GRCh38
rs200644559 Chr.11: 62839480-62839480 on Build GRCh38
rs200389987 Chr.11: 62839070-62839070 on Build GRCh38
rs536299172 Chr.11: 62839206-62839206 on Build GRCh38
rs777009922 Chr.11: 62839282-62839282 on Build GRCh38
rs750300129 Chr.11: 62839264-62839264 on Build GRCh38
rs778391878 Chr.11: 62839302-62839302 on Build GRCh38
rs74789950 Chr.11: 62839435-62839435 on Build GRCh38
rs541177302 Chr.11: 62839377-62839377 on Build GRCh38
rs758552593 Chr.11: 62839505-62839505 on Build GRCh38
rs780297098 Chr.11: 62839195-62839195 on Build GRCh38
rs1049898577 Chr.11: 62839310-62839310 on Build GRCh38
rs866772092 Chr.11: 62839183-62839183 on Build GRCh38
rs749493618 Chr.11: 62839201-62839201 on Build GRCh38
rs370434980 Chr.11: 62839364-62839364 on Build GRCh38
rs747332581 Chr.11: 62839430-62839430 on Build GRCh38
rs748874864 Chr.11: 62839442-62839442 on Build GRCh38
rs772690331 Chr.11: 62839218-62839218 on Build GRCh38
rs776515689 Chr.11: 62839487-62839487 on Build GRCh38
rs767197459 Chr.11: 62839330-62839330 on Build GRCh38
rs376677404 Chr.11: 62839180-62839180 on Build GRCh38
rs150613372 Chr.11: 62839492-62839492 on Build GRCh38
rs1055033034 Chr.11: 62839464-62839464 on Build GRCh38
rs374514023 Chr.11: 62839186-62839186 on Build GRCh38
rs111846457 Chr.11: 62839334-62839334 on Build GRCh38
rs906496851 Chr.11: 62839285-62839285 on Build GRCh38
rs547374694 Chr.11: 62839132-62839132 on Build GRCh38
rs377645483 Chr.11: 62839128-62839128 on Build GRCh38
rs746920557 Chr.11: 62839071-62839071 on Build GRCh38
rs371820946 Chr.11: 62839484-62839484 on Build GRCh38
rs369430523 Chr.11: 62839136-62839136 on Build GRCh38
rs571299195 Chr.11: 62839072-62839072 on Build GRCh38
rs919009472 Chr.11: 62839204-62839204 on Build GRCh38
rs767560991 Chr.11: 62839257-62839257 on Build GRCh38
rs778029633 Chr.11: 62839415-62839415 on Build GRCh38
rs759766180 Chr.11: 62839426-62839426 on Build GRCh38
rs767841763 Chr.11: 62839129-62839129 on Build GRCh38
rs764631164 Chr.11: 62839220-62839220 on Build GRCh38
rs756271271 Chr.11: 62839268-62839268 on Build GRCh38
rs201289915 Chr.11: 62839190-62839190 on Build GRCh38
rs753922886 Chr.11: 62839065-62839066 on Build GRCh38
rs368356605 Chr.11: 62839445-62839445 on Build GRCh38
rs756147059 Chr.11: 62839188-62839188 on Build GRCh38
rs60799389 Chr.11: 62839308-62839308 on Build GRCh38
rs376886585 Chr.11: 62839232-62839232 on Build GRCh38
rs749722385 Chr.11: 62839114-62839114 on Build GRCh38
rs775145640 Chr.11: 62839312-62839312 on Build GRCh38
rs751415769 Chr.11: 62839240-62839240 on Build GRCh38
rs775564727 Chr.11: 62839103-62839103 on Build GRCh38
rs939715505 Chr.11: 62839263-62839263 on Build GRCh38
rs911529348 Chr.11: 62839156-62839156 on Build GRCh38
rs201618759 Chr.11: 62839295-62839295 on Build GRCh38
rs755203173 Chr.11: 62839289-62839289 on Build GRCh38
rs762494919 Chr.11: 62839324-62839324 on Build GRCh38
rs201550993 Chr.11: 62839122-62839122 on Build GRCh38
rs766243027 Chr.11: 62839353-62839353 on Build GRCh38
rs753845556 Chr.11: 62839274-62839274 on Build GRCh38
rs989657300 Chr.11: 62839091-62839091 on Build GRCh38
rs535814738 Chr.11: 62839373-62839373 on Build GRCh38
rs866881471 Chr.11: 62839344-62839344 on Build GRCh38
rs773492636 Chr.11: 62839066-62839072 on Build GRCh38
rs765321626 Chr.11: 62839498-62839498 on Build GRCh38
rs774890860 Chr.11: 62839124-62839124 on Build GRCh38

View Full Product Details