Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 116845145-116845651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOA1-AS
Entrez Gene ID
104326055
Gene Name
APOA1 antisense RNA
Gene Aliases
-
UniGene
Hs.627813
Gene Chromosome Location
Chr.11: 116836117-116855729 on Build GRCh38
Transcripts
NR_126362.1 NM_001281748.1 NM_001281749.1 NM_025164.4 XM_005271481.2 XM_005271482.4 XM_005271484.3 XM_005271485.3 XM_011542722.1 XM_011542723.2 XM_011542724.2 XM_011542725.2 XM_017017424.1 XM_017017425.1 XM_017017426.1 XM_017017427.1

Gene Symbol
SIK3
Entrez Gene ID
23387
Gene Name
SIK family kinase 3
Gene Aliases
L19, QSK, SIK-3
UniGene
Hs.167451
Gene Chromosome Location
Chr.11: 116843402-117098453 on Build GRCh38
Transcripts
NR_126362.1 NM_001281748.1 NM_001281749.1 NM_025164.4 XM_005271481.2 XM_005271482.4 XM_005271484.3 XM_005271485.3 XM_011542722.1 XM_011542723.2 XM_011542724.2 XM_011542725.2 XM_017017424.1 XM_017017425.1 XM_017017426.1 XM_017017427.1

SNP Detail

SNP ID Chromosome Location
rs1034576271 Chr.11: 116845485-116845485 on Build GRCh38
rs961157807 Chr.11: 116845265-116845265 on Build GRCh38
rs758296711 Chr.11: 116845341-116845341 on Build GRCh38
rs961147926 Chr.11: 116845554-116845554 on Build GRCh38
rs774718154 Chr.11: 116845481-116845481 on Build GRCh38
rs973020306 Chr.11: 116845269-116845269 on Build GRCh38
rs985752768 Chr.11: 116845604-116845604 on Build GRCh38
rs11420452 Chr.11: 116845290-116845291 on Build GRCh38
rs909645349 Chr.11: 116845253-116845253 on Build GRCh38
rs1046348875 Chr.11: 116845431-116845431 on Build GRCh38
rs911752345 Chr.11: 116845274-116845274 on Build GRCh38
rs561593375 Chr.11: 116845325-116845325 on Build GRCh38
rs907414075 Chr.11: 116845386-116845386 on Build GRCh38
rs757377562 Chr.11: 116845287-116845288 on Build GRCh38
rs533039203 Chr.11: 116845571-116845571 on Build GRCh38
rs191753633 Chr.11: 116845364-116845364 on Build GRCh38
rs1049733305 Chr.11: 116845281-116845281 on Build GRCh38
rs565845402 Chr.11: 116845601-116845601 on Build GRCh38
rs1016639322 Chr.11: 116845523-116845523 on Build GRCh38
rs563725578 Chr.11: 116845493-116845493 on Build GRCh38
rs552432726 Chr.11: 116845582-116845582 on Build GRCh38
rs766647238 Chr.11: 116845478-116845478 on Build GRCh38
rs907430911 Chr.11: 116845453-116845453 on Build GRCh38
rs1054503856 Chr.11: 116845305-116845305 on Build GRCh38
rs1001654198 Chr.11: 116845466-116845466 on Build GRCh38
rs780942362 Chr.11: 116845234-116845234 on Build GRCh38
rs541691918 Chr.11: 116845235-116845235 on Build GRCh38
rs762242553 Chr.11: 116845498-116845498 on Build GRCh38
rs552513504 Chr.11: 116845300-116845300 on Build GRCh38
rs1010395370 Chr.11: 116845400-116845400 on Build GRCh38
rs933930013 Chr.11: 116845274-116845274 on Build GRCh38
rs543847728 Chr.11: 116845425-116845425 on Build GRCh38
rs922539325 Chr.11: 116845273-116845273 on Build GRCh38
rs376207934 Chr.11: 116845195-116845195 on Build GRCh38
rs1056453075 Chr.11: 116845486-116845487 on Build GRCh38
rs773517907 Chr.11: 116845572-116845572 on Build GRCh38
rs959047518 Chr.11: 116845218-116845218 on Build GRCh38
rs184002013 Chr.11: 116845499-116845499 on Build GRCh38
rs946157699 Chr.11: 116845369-116845369 on Build GRCh38
rs769407120 Chr.11: 116845371-116845371 on Build GRCh38
rs746808435 Chr.11: 116845590-116845591 on Build GRCh38
rs913322934 Chr.11: 116845335-116845335 on Build GRCh38
rs972579248 Chr.11: 116845555-116845555 on Build GRCh38
rs770655244 Chr.11: 116845181-116845181 on Build GRCh38
rs911202737 Chr.11: 116845290-116845290 on Build GRCh38

View Full Product Details