Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31667635-31668166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFBKMT
Entrez Gene ID
254013
Gene Name
electron transfer flavoprotein beta subunit lysine methyltransferase
Gene Aliases
C12orf72, ETFB-KMT, METTL20
UniGene
Hs.740628
Gene Chromosome Location
Chr.12: 31647160-31669082 on Build GRCh38
Transcripts
NM_001135863.1 NM_001135864.1 NM_173802.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1299320 c.694C>T p.H232Y Chr.12: 31667895-31667895 on Build GRCh38
938760 c.561C>T p.G187G Chr.12: 31667762-31667762 on Build GRCh38
4041407 c.639A>T p.R213R Chr.12: 31667840-31667840 on Build GRCh38
938761 c.562G>A p.D188N Chr.12: 31667763-31667763 on Build GRCh38
4041408 c.683G>A p.S228N Chr.12: 31667884-31667884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774637167 Chr.12: 31667678-31667678 on Build GRCh38
rs187315527 Chr.12: 31668050-31668050 on Build GRCh38
rs759869819 Chr.12: 31667792-31667792 on Build GRCh38
rs771774667 Chr.12: 31667911-31667911 on Build GRCh38
rs768921654 Chr.12: 31667749-31667749 on Build GRCh38
rs11051523 Chr.12: 31667784-31667784 on Build GRCh38
rs746577989 Chr.12: 31667809-31667810 on Build GRCh38
rs748689531 Chr.12: 31667684-31667684 on Build GRCh38
rs375673238 Chr.12: 31668043-31668043 on Build GRCh38
rs138981620 Chr.12: 31667968-31667968 on Build GRCh38
rs746884192 Chr.12: 31668038-31668038 on Build GRCh38
rs765595766 Chr.12: 31667803-31667803 on Build GRCh38
rs137969510 Chr.12: 31667973-31667974 on Build GRCh38
rs773662236 Chr.12: 31667783-31667783 on Build GRCh38
rs75288003 Chr.12: 31667977-31667977 on Build GRCh38
rs371064181 Chr.12: 31668025-31668025 on Build GRCh38
rs374501015 Chr.12: 31667830-31667830 on Build GRCh38
rs753766062 Chr.12: 31667955-31667955 on Build GRCh38
rs754466105 Chr.12: 31667831-31667831 on Build GRCh38
rs11833865 Chr.12: 31668082-31668082 on Build GRCh38
rs765824714 Chr.12: 31667825-31667825 on Build GRCh38
rs773750432 Chr.12: 31667715-31667715 on Build GRCh38
rs143179970 Chr.12: 31667763-31667763 on Build GRCh38
rs890913282 Chr.12: 31667730-31667730 on Build GRCh38
rs781778594 Chr.12: 31667990-31667990 on Build GRCh38
rs1046751185 Chr.12: 31667693-31667693 on Build GRCh38
rs771429036 Chr.12: 31667734-31667738 on Build GRCh38
rs770711072 Chr.12: 31668042-31668042 on Build GRCh38
rs746370920 Chr.12: 31667999-31667999 on Build GRCh38
rs779627322 Chr.12: 31667672-31667672 on Build GRCh38
rs1014467646 Chr.12: 31668068-31668068 on Build GRCh38
rs115438422 Chr.12: 31667922-31667922 on Build GRCh38
rs140131797 Chr.12: 31667910-31667910 on Build GRCh38
rs775920698 Chr.12: 31667906-31667906 on Build GRCh38
rs191590289 Chr.12: 31667782-31667782 on Build GRCh38
rs866822826 Chr.12: 31667812-31667812 on Build GRCh38
rs748565034 Chr.12: 31667777-31667777 on Build GRCh38
rs140221965 Chr.12: 31667838-31667838 on Build GRCh38
rs780223212 Chr.12: 31668014-31668014 on Build GRCh38
rs1031979845 Chr.12: 31668033-31668033 on Build GRCh38
rs145210983 Chr.12: 31667960-31667960 on Build GRCh38
rs970105672 Chr.12: 31668130-31668131 on Build GRCh38
rs769009890 Chr.12: 31667674-31667674 on Build GRCh38
rs532044112 Chr.12: 31668019-31668019 on Build GRCh38
rs572835303 Chr.12: 31667764-31667764 on Build GRCh38
rs200960305 Chr.12: 31667747-31667747 on Build GRCh38
rs761756915 Chr.12: 31667817-31667817 on Build GRCh38
rs200404212 Chr.12: 31667762-31667762 on Build GRCh38
rs766391839 Chr.12: 31667949-31667949 on Build GRCh38
rs201977098 Chr.12: 31667871-31667871 on Build GRCh38
rs1021288103 Chr.12: 31667971-31667971 on Build GRCh38
rs149097385 Chr.12: 31667716-31667716 on Build GRCh38
rs776587047 Chr.12: 31667949-31667950 on Build GRCh38
rs375682347 Chr.12: 31667748-31667748 on Build GRCh38
rs768556154 Chr.12: 31667882-31667883 on Build GRCh38
rs766477603 Chr.12: 31667719-31667719 on Build GRCh38
rs764004720 Chr.12: 31667737-31667737 on Build GRCh38
rs774988598 Chr.12: 31667749-31667751 on Build GRCh38
rs772611330 Chr.12: 31667711-31667711 on Build GRCh38
rs750329151 Chr.12: 31667820-31667820 on Build GRCh38
rs757539947 Chr.12: 31667989-31667989 on Build GRCh38
rs767336172 Chr.12: 31667818-31667818 on Build GRCh38
rs370377082 Chr.12: 31667736-31667736 on Build GRCh38
rs1030470545 Chr.12: 31667687-31667687 on Build GRCh38
rs151164668 Chr.12: 31667862-31667862 on Build GRCh38
rs183659283 Chr.12: 31667867-31667867 on Build GRCh38
rs759116519 Chr.12: 31667957-31667957 on Build GRCh38
rs959908851 Chr.12: 31668055-31668055 on Build GRCh38
rs767401938 Chr.12: 31667742-31667742 on Build GRCh38
rs776723811 Chr.12: 31667790-31667790 on Build GRCh38
rs371569520 Chr.12: 31667962-31667962 on Build GRCh38
rs760557243 Chr.12: 31667948-31667948 on Build GRCh38
rs372518966 Chr.12: 31667863-31667863 on Build GRCh38
rs200465923 Chr.12: 31668013-31668013 on Build GRCh38
rs374343664 Chr.12: 31667741-31667741 on Build GRCh38
rs745993697 Chr.12: 31667889-31667889 on Build GRCh38
rs562157491 Chr.12: 31667925-31667925 on Build GRCh38
rs777964462 Chr.12: 31668121-31668121 on Build GRCh38
rs575191407 Chr.12: 31667809-31667809 on Build GRCh38
rs748963162 Chr.12: 31667673-31667673 on Build GRCh38
rs769979813 Chr.12: 31667904-31667904 on Build GRCh38
rs752345267 Chr.12: 31667839-31667839 on Build GRCh38
rs140923820 Chr.12: 31667823-31667823 on Build GRCh38

View Full Product Details