Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120738719-120739222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADS
Entrez Gene ID
35
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
Gene Aliases
ACAD3, SCAD
UniGene
Hs.507076
Gene Chromosome Location
Chr.12: 120725738-120740008 on Build GRCh38
Transcripts
NM_000017.3 NM_001302554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1639055 c.1067C>T p.A356V Chr.12: 120739177-120739177 on Build GRCh38
6852894 c.974G>A p.R325Q Chr.12: 120738860-120738860 on Build GRCh38
7296219 c.1029G>A p.K343K Chr.12: 120738915-120738915 on Build GRCh38
4995088 c.982A>T p.T328S Chr.12: 120738868-120738868 on Build GRCh38
7235654 c.1066G>A p.A356T Chr.12: 120739176-120739176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555045670 Chr.12: 120739056-120739056 on Build GRCh38
rs370161104 Chr.12: 120738935-120738935 on Build GRCh38
rs796051906 Chr.12: 120738874-120738876 on Build GRCh38
rs777823861 Chr.12: 120738758-120738758 on Build GRCh38
rs979058757 Chr.12: 120738978-120738979 on Build GRCh38
rs377559470 Chr.12: 120739178-120739178 on Build GRCh38
rs757880686 Chr.12: 120738822-120738822 on Build GRCh38
rs756508815 Chr.12: 120738924-120738924 on Build GRCh38
rs57852948 Chr.12: 120739000-120739001 on Build GRCh38
rs931944863 Chr.12: 120738968-120738968 on Build GRCh38
rs745757125 Chr.12: 120738940-120738942 on Build GRCh38
rs753547944 Chr.12: 120738809-120738809 on Build GRCh38
rs894829104 Chr.12: 120739034-120739034 on Build GRCh38
rs768426621 Chr.12: 120738958-120738958 on Build GRCh38
rs121908006 Chr.12: 120738859-120738859 on Build GRCh38
rs757766697 Chr.12: 120739106-120739106 on Build GRCh38
rs375767053 Chr.12: 120738925-120738925 on Build GRCh38
rs751692085 Chr.12: 120738784-120738786 on Build GRCh38
rs374802717 Chr.12: 120738800-120738800 on Build GRCh38
rs1049568378 Chr.12: 120739055-120739055 on Build GRCh38
rs145594828 Chr.12: 120739163-120739163 on Build GRCh38
rs781172419 Chr.12: 120739162-120739162 on Build GRCh38
rs781742944 Chr.12: 120738825-120738825 on Build GRCh38
rs932226538 Chr.12: 120738782-120738782 on Build GRCh38
rs765011205 Chr.12: 120738965-120738965 on Build GRCh38
rs35503787 Chr.12: 120738813-120738814 on Build GRCh38
rs753258340 Chr.12: 120738915-120738915 on Build GRCh38
rs487915 Chr.12: 120738746-120738746 on Build GRCh38
rs900784894 Chr.12: 120738779-120738779 on Build GRCh38
rs796051903 Chr.12: 120739149-120739149 on Build GRCh38
rs387906950 Chr.12: 120739141-120739141 on Build GRCh38
rs762451430 Chr.12: 120739092-120739092 on Build GRCh38
rs113168595 Chr.12: 120738770-120738770 on Build GRCh38
rs770439736 Chr.12: 120739174-120739174 on Build GRCh38
rs759011410 Chr.12: 120739181-120739181 on Build GRCh38
rs907938067 Chr.12: 120738747-120738747 on Build GRCh38
rs750672257 Chr.12: 120739127-120739127 on Build GRCh38
rs140372512 Chr.12: 120739099-120739099 on Build GRCh38
rs202078273 Chr.12: 120739164-120739164 on Build GRCh38
rs563943837 Chr.12: 120738933-120738933 on Build GRCh38
rs766506561 Chr.12: 120738777-120738778 on Build GRCh38
rs566185314 Chr.12: 120738976-120738976 on Build GRCh38
rs545598561 Chr.12: 120739143-120739143 on Build GRCh38
rs757049257 Chr.12: 120738923-120738924 on Build GRCh38
rs750045046 Chr.12: 120738902-120738902 on Build GRCh38
rs987468270 Chr.12: 120739170-120739170 on Build GRCh38
rs767769414 Chr.12: 120738898-120738900 on Build GRCh38
rs752558816 Chr.12: 120738797-120738797 on Build GRCh38
rs758484911 Chr.12: 120739129-120739129 on Build GRCh38
rs754594081 Chr.12: 120738810-120738810 on Build GRCh38
rs946248018 Chr.12: 120738991-120738991 on Build GRCh38
rs777939247 Chr.12: 120738844-120738844 on Build GRCh38
rs746266217 Chr.12: 120738984-120738984 on Build GRCh38
rs371199821 Chr.12: 120738802-120738802 on Build GRCh38
rs754459133 Chr.12: 120739182-120739182 on Build GRCh38
rs754369412 Chr.12: 120739105-120739105 on Build GRCh38
rs932525260 Chr.12: 120738860-120738860 on Build GRCh38
rs568010677 Chr.12: 120738879-120738879 on Build GRCh38
rs534848765 Chr.12: 120739005-120739005 on Build GRCh38
rs764917783 Chr.12: 120738899-120738899 on Build GRCh38
rs570328023 Chr.12: 120738953-120738953 on Build GRCh38
rs922358310 Chr.12: 120739029-120739029 on Build GRCh38
rs752914704 Chr.12: 120738922-120738922 on Build GRCh38
rs749523619 Chr.12: 120738849-120738849 on Build GRCh38
rs371468373 Chr.12: 120738983-120738983 on Build GRCh38
rs397744635 Chr.12: 120739004-120739005 on Build GRCh38
rs778604166 Chr.12: 120738929-120738937 on Build GRCh38
rs764750642 Chr.12: 120739101-120739101 on Build GRCh38
rs771241888 Chr.12: 120738857-120738857 on Build GRCh38
rs757992559 Chr.12: 120738905-120738905 on Build GRCh38
rs904655362 Chr.12: 120739002-120739002 on Build GRCh38
rs768748973 Chr.12: 120739160-120739160 on Build GRCh38
rs796051904 Chr.12: 120739167-120739167 on Build GRCh38
rs779408255 Chr.12: 120739120-120739120 on Build GRCh38
rs755177866 Chr.12: 120738807-120738807 on Build GRCh38
rs528266975 Chr.12: 120739188-120739188 on Build GRCh38
rs190464662 Chr.12: 120739076-120739076 on Build GRCh38
rs756848037 Chr.12: 120738813-120738813 on Build GRCh38
rs377672631 Chr.12: 120738960-120738960 on Build GRCh38
rs748594281 Chr.12: 120738828-120738828 on Build GRCh38
rs970614348 Chr.12: 120739082-120739082 on Build GRCh38
rs767442503 Chr.12: 120738778-120738778 on Build GRCh38
rs140853839 Chr.12: 120738874-120738874 on Build GRCh38
rs200346345 Chr.12: 120738897-120738897 on Build GRCh38
rs28941773 Chr.12: 120739168-120739168 on Build GRCh38
rs749885391 Chr.12: 120738815-120738815 on Build GRCh38
rs778423843 Chr.12: 120738814-120738814 on Build GRCh38
rs761474976 Chr.12: 120738880-120738880 on Build GRCh38
rs775376445 Chr.12: 120738956-120738956 on Build GRCh38
rs541587321 Chr.12: 120739194-120739194 on Build GRCh38
rs765914571 Chr.12: 120739098-120739098 on Build GRCh38
rs942118123 Chr.12: 120738959-120738959 on Build GRCh38
rs1057517155 Chr.12: 120739141-120739141 on Build GRCh38
rs1011856639 Chr.12: 120739036-120739036 on Build GRCh38
rs1037337896 Chr.12: 120738925-120738925 on Build GRCh38
rs1000626538 Chr.12: 120739004-120739004 on Build GRCh38
rs2071266 Chr.12: 120738967-120738967 on Build GRCh38
rs1057516639 Chr.12: 120738916-120738916 on Build GRCh38
rs3915 Chr.12: 120738876-120738876 on Build GRCh38
rs765789568 Chr.12: 120738911-120738911 on Build GRCh38
rs768733898 Chr.12: 120739176-120739176 on Build GRCh38
rs199633532 Chr.12: 120738875-120738875 on Build GRCh38
rs942049307 Chr.12: 120738769-120738769 on Build GRCh38
rs34289241 Chr.12: 120739131-120739132 on Build GRCh38
rs537027219 Chr.12: 120738934-120738934 on Build GRCh38
rs756555604 Chr.12: 120738838-120738838 on Build GRCh38
rs759579281 Chr.12: 120738771-120738771 on Build GRCh38
rs1024820595 Chr.12: 120738926-120738926 on Build GRCh38
rs697853 Chr.12: 120739175-120739175 on Build GRCh38
rs12315099 Chr.12: 120738951-120738951 on Build GRCh38
rs1041882591 Chr.12: 120738995-120738995 on Build GRCh38

View Full Product Details