Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121778088-121778627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOF
Entrez Gene ID
54509
Gene Name
ras homolog family member F, filopodia associated
Gene Aliases
ARHF, RIF
UniGene
Hs.707579
Gene Chromosome Location
Chr.12: 121777754-121793688 on Build GRCh38
Transcripts
NM_019034.2 NM_001080825.2 XM_005253843.4 XM_017018853.1 XM_017018854.1

Gene Symbol
TMEM120B
Entrez Gene ID
144404
Gene Name
transmembrane protein 120B
Gene Aliases
-
UniGene
Hs.644504
Gene Chromosome Location
Chr.12: 121712752-121782070 on Build GRCh38
Transcripts
NM_019034.2 NM_001080825.2 XM_005253843.4 XM_017018853.1 XM_017018854.1

SNP Detail

SNP ID Chromosome Location
rs113401351 Chr.12: 121778324-121778324 on Build GRCh38
rs990855913 Chr.12: 121778257-121778257 on Build GRCh38
rs1045460628 Chr.12: 121778508-121778508 on Build GRCh38
rs1021101433 Chr.12: 121778205-121778205 on Build GRCh38
rs1002368170 Chr.12: 121778248-121778248 on Build GRCh38
rs902675915 Chr.12: 121778223-121778223 on Build GRCh38
rs1033871335 Chr.12: 121778249-121778249 on Build GRCh38
rs983219580 Chr.12: 121778556-121778556 on Build GRCh38
rs973678305 Chr.12: 121778604-121778604 on Build GRCh38
rs958350326 Chr.12: 121778250-121778250 on Build GRCh38
rs1047837994 Chr.12: 121778188-121778188 on Build GRCh38
rs959070616 Chr.12: 121778247-121778247 on Build GRCh38
rs930436171 Chr.12: 121778175-121778175 on Build GRCh38
rs775248318 Chr.12: 121778586-121778587 on Build GRCh38
rs560432744 Chr.12: 121778190-121778190 on Build GRCh38
rs544454849 Chr.12: 121778119-121778120 on Build GRCh38
rs891245835 Chr.12: 121778181-121778181 on Build GRCh38
rs79867896 Chr.12: 121778513-121778513 on Build GRCh38
rs185128602 Chr.12: 121778320-121778320 on Build GRCh38
rs938272630 Chr.12: 121778532-121778532 on Build GRCh38
rs149023564 Chr.12: 121778289-121778289 on Build GRCh38
rs527567712 Chr.12: 121778224-121778224 on Build GRCh38
rs1021004504 Chr.12: 121778587-121778587 on Build GRCh38
rs141972494 Chr.12: 121778429-121778429 on Build GRCh38
rs911813937 Chr.12: 121778127-121778127 on Build GRCh38
rs1001401152 Chr.12: 121778428-121778428 on Build GRCh38
rs895420084 Chr.12: 121778469-121778469 on Build GRCh38
rs949690742 Chr.12: 121778451-121778455 on Build GRCh38
rs983188213 Chr.12: 121778276-121778276 on Build GRCh38
rs558451578 Chr.12: 121778573-121778573 on Build GRCh38
rs931911104 Chr.12: 121778310-121778310 on Build GRCh38
rs74478146 Chr.12: 121778577-121778577 on Build GRCh38
rs3782337 Chr.12: 121778136-121778136 on Build GRCh38
rs930486613 Chr.12: 121778381-121778381 on Build GRCh38
rs973348542 Chr.12: 121778293-121778293 on Build GRCh38
rs970354840 Chr.12: 121778554-121778554 on Build GRCh38
rs570500854 Chr.12: 121778261-121778261 on Build GRCh38
rs181045560 Chr.12: 121778124-121778124 on Build GRCh38
rs541628325 Chr.12: 121778174-121778174 on Build GRCh38
rs932606521 Chr.12: 121778153-121778153 on Build GRCh38
rs190913228 Chr.12: 121778479-121778479 on Build GRCh38
rs1011493153 Chr.12: 121778585-121778585 on Build GRCh38
rs183997704 Chr.12: 121778578-121778578 on Build GRCh38
rs1052202 Chr.12: 121778130-121778130 on Build GRCh38
rs1032601822 Chr.12: 121778465-121778465 on Build GRCh38
rs780366965 Chr.12: 121778523-121778523 on Build GRCh38
rs1010961417 Chr.12: 121778258-121778258 on Build GRCh38
rs566412089 Chr.12: 121778427-121778427 on Build GRCh38
rs369932352 Chr.12: 121778279-121778279 on Build GRCh38
rs920339510 Chr.12: 121778137-121778137 on Build GRCh38
rs143838180 Chr.12: 121778315-121778315 on Build GRCh38
rs571768935 Chr.12: 121778311-121778311 on Build GRCh38
rs35474958 Chr.12: 121778451-121778452 on Build GRCh38
rs769929962 Chr.12: 121778195-121778195 on Build GRCh38
rs906762575 Chr.12: 121778526-121778526 on Build GRCh38
rs906816255 Chr.12: 121778246-121778246 on Build GRCh38
rs543824359 Chr.12: 121778263-121778263 on Build GRCh38
rs1042424324 Chr.12: 121778346-121778346 on Build GRCh38
rs920471250 Chr.12: 121778375-121778375 on Build GRCh38
rs893949421 Chr.12: 121778585-121778586 on Build GRCh38
rs61954262 Chr.12: 121778448-121778448 on Build GRCh38
rs758815050 Chr.12: 121778366-121778366 on Build GRCh38
rs966697703 Chr.12: 121778282-121778282 on Build GRCh38
rs886077844 Chr.12: 121778193-121778193 on Build GRCh38
rs774012085 Chr.12: 121778120-121778120 on Build GRCh38
rs751651464 Chr.12: 121778450-121778451 on Build GRCh38
rs986296702 Chr.12: 121778483-121778483 on Build GRCh38
rs57344511 Chr.12: 121778471-121778472 on Build GRCh38
rs137894421 Chr.12: 121778235-121778235 on Build GRCh38
rs921811176 Chr.12: 121778308-121778308 on Build GRCh38
rs767016349 Chr.12: 121778142-121778142 on Build GRCh38

View Full Product Details