Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 114324079-114324601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAMP1
Entrez Gene ID
283489
Gene Name
chromosome alignment maintaining phosphoprotein 1
Gene Aliases
C13orf8, CAMP, CHAMP, MRD40, ZNF828
UniGene
Hs.7542
Gene Chromosome Location
Chr.13: 114314490-114327328 on Build GRCh38
Transcripts
NM_001164144.1 NM_001164145.1 NM_032436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793159 c.601C>T p.P201S Chr.13: 114324443-114324443 on Build GRCh38
117458 c.265G>C p.D89H Chr.13: 114324107-114324107 on Build GRCh38
4920253 c.327T>C p.P109P Chr.13: 114324169-114324169 on Build GRCh38
3753606 c.702G>A p.P234P Chr.13: 114324544-114324544 on Build GRCh38
3793158 c.554C>T p.P185L Chr.13: 114324396-114324396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369063490 Chr.13: 114324237-114324237 on Build GRCh38
rs782626360 Chr.13: 114324514-114324514 on Build GRCh38
rs782452899 Chr.13: 114324267-114324267 on Build GRCh38
rs35054271 Chr.13: 114324118-114324118 on Build GRCh38
rs781942855 Chr.13: 114324557-114324557 on Build GRCh38
rs782242424 Chr.13: 114324148-114324148 on Build GRCh38
rs372767810 Chr.13: 114324562-114324562 on Build GRCh38
rs928746655 Chr.13: 114324523-114324523 on Build GRCh38
rs143111151 Chr.13: 114324105-114324105 on Build GRCh38
rs782133261 Chr.13: 114324347-114324347 on Build GRCh38
rs782063204 Chr.13: 114324467-114324467 on Build GRCh38
rs781916452 Chr.13: 114324225-114324225 on Build GRCh38
rs781979890 Chr.13: 114324179-114324179 on Build GRCh38
rs782652629 Chr.13: 114324125-114324125 on Build GRCh38
rs781891460 Chr.13: 114324254-114324254 on Build GRCh38
rs376019121 Chr.13: 114324470-114324470 on Build GRCh38
rs148053905 Chr.13: 114324485-114324485 on Build GRCh38
rs782782411 Chr.13: 114324287-114324287 on Build GRCh38
rs782223907 Chr.13: 114324543-114324543 on Build GRCh38
rs782332427 Chr.13: 114324471-114324471 on Build GRCh38
rs782103470 Chr.13: 114324477-114324477 on Build GRCh38
rs782138377 Chr.13: 114324223-114324223 on Build GRCh38
rs189158463 Chr.13: 114324358-114324358 on Build GRCh38
rs141932275 Chr.13: 114324386-114324386 on Build GRCh38
rs782662372 Chr.13: 114324159-114324159 on Build GRCh38
rs782123443 Chr.13: 114324252-114324252 on Build GRCh38
rs782729324 Chr.13: 114324382-114324382 on Build GRCh38
rs139973531 Chr.13: 114324500-114324500 on Build GRCh38
rs782368765 Chr.13: 114324192-114324192 on Build GRCh38
rs782659992 Chr.13: 114324414-114324414 on Build GRCh38
rs782724274 Chr.13: 114324224-114324224 on Build GRCh38
rs898434882 Chr.13: 114324255-114324255 on Build GRCh38
rs372508540 Chr.13: 114324443-114324443 on Build GRCh38
rs782179967 Chr.13: 114324534-114324534 on Build GRCh38
rs782505873 Chr.13: 114324248-114324248 on Build GRCh38
rs781918135 Chr.13: 114324465-114324465 on Build GRCh38
rs782717643 Chr.13: 114324494-114324494 on Build GRCh38
rs573294954 Chr.13: 114324229-114324229 on Build GRCh38
rs201497347 Chr.13: 114324187-114324187 on Build GRCh38
rs200240271 Chr.13: 114324379-114324379 on Build GRCh38
rs782813507 Chr.13: 114324481-114324481 on Build GRCh38
rs199714471 Chr.13: 114324551-114324551 on Build GRCh38
rs781857456 Chr.13: 114324579-114324579 on Build GRCh38
rs781825160 Chr.13: 114324499-114324499 on Build GRCh38
rs782404252 Chr.13: 114324288-114324288 on Build GRCh38
rs782748396 Chr.13: 114324566-114324566 on Build GRCh38
rs782352206 Chr.13: 114324325-114324325 on Build GRCh38
rs781876316 Chr.13: 114324241-114324241 on Build GRCh38
rs863225077 Chr.13: 114324384-114324385 on Build GRCh38
rs782547315 Chr.13: 114324412-114324412 on Build GRCh38
rs782812955 Chr.13: 114324253-114324253 on Build GRCh38
rs782718781 Chr.13: 114324376-114324376 on Build GRCh38
rs782038403 Chr.13: 114324367-114324367 on Build GRCh38
rs781805407 Chr.13: 114324418-114324418 on Build GRCh38
rs782469663 Chr.13: 114324107-114324107 on Build GRCh38
rs957871537 Chr.13: 114324437-114324437 on Build GRCh38
rs782278722 Chr.13: 114324455-114324455 on Build GRCh38
rs144012078 Chr.13: 114324519-114324519 on Build GRCh38
rs540964802 Chr.13: 114324232-114324232 on Build GRCh38
rs781848204 Chr.13: 114324384-114324384 on Build GRCh38
rs568507339 Chr.13: 114324474-114324474 on Build GRCh38
rs782730794 Chr.13: 114324236-114324236 on Build GRCh38
rs1007989539 Chr.13: 114324326-114324326 on Build GRCh38
rs953865034 Chr.13: 114324313-114324313 on Build GRCh38
rs148563449 Chr.13: 114324336-114324336 on Build GRCh38
rs924961662 Chr.13: 114324395-114324395 on Build GRCh38
rs541872327 Chr.13: 114324153-114324153 on Build GRCh38
rs782623550 Chr.13: 114324440-114324440 on Build GRCh38
rs781911171 Chr.13: 114324486-114324486 on Build GRCh38
rs782028595 Chr.13: 114324213-114324213 on Build GRCh38
rs782232019 Chr.13: 114324269-114324269 on Build GRCh38
rs559362276 Chr.13: 114324276-114324276 on Build GRCh38
rs782153954 Chr.13: 114324371-114324371 on Build GRCh38
rs3813131 Chr.13: 114324544-114324544 on Build GRCh38
rs782625369 Chr.13: 114324274-114324274 on Build GRCh38
rs782417189 Chr.13: 114324457-114324457 on Build GRCh38
rs144710092 Chr.13: 114324157-114324157 on Build GRCh38
rs201875013 Chr.13: 114324517-114324517 on Build GRCh38
rs531791050 Chr.13: 114324381-114324381 on Build GRCh38
rs782597168 Chr.13: 114324540-114324540 on Build GRCh38
rs574736824 Chr.13: 114324348-114324348 on Build GRCh38
rs782583966 Chr.13: 114324268-114324268 on Build GRCh38
rs141043775 Chr.13: 114324487-114324487 on Build GRCh38
rs371219971 Chr.13: 114324300-114324300 on Build GRCh38
rs782626191 Chr.13: 114324449-114324449 on Build GRCh38
rs782689029 Chr.13: 114324532-114324532 on Build GRCh38
rs782436384 Chr.13: 114324436-114324436 on Build GRCh38
rs797044961 Chr.13: 114324477-114324477 on Build GRCh38

View Full Product Details