Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40459498-40459989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAHD1
Entrez Gene ID
22893
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
UniGene
Hs.22109
Gene Chromosome Location
Chr.15: 40439721-40468242 on Build GRCh38
Transcripts
NM_001301132.1 NM_014952.4 NR_125372.1 XM_011521366.2 XM_011521367.2 XM_011521368.2 XM_011521369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6323225 c.1060C>T p.P354S Chr.15: 40459524-40459524 on Build GRCh38
5849077 c.1156G>A p.E386K Chr.15: 40459620-40459620 on Build GRCh38
470569 c.1062G>A p.P354P Chr.15: 40459526-40459526 on Build GRCh38
6277341 c.1253C>T p.A418V Chr.15: 40459717-40459717 on Build GRCh38
4054367 c.1182G>A p.S394S Chr.15: 40459646-40459646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012344794 Chr.15: 40459624-40459624 on Build GRCh38
rs769196707 Chr.15: 40459918-40459918 on Build GRCh38
rs746510563 Chr.15: 40459777-40459777 on Build GRCh38
rs202063791 Chr.15: 40459550-40459550 on Build GRCh38
rs1043834764 Chr.15: 40459724-40459724 on Build GRCh38
rs755937706 Chr.15: 40459852-40459852 on Build GRCh38
rs367799058 Chr.15: 40459768-40459768 on Build GRCh38
rs780418645 Chr.15: 40459557-40459557 on Build GRCh38
rs942838199 Chr.15: 40459761-40459761 on Build GRCh38
rs138759345 Chr.15: 40459821-40459821 on Build GRCh38
rs563593493 Chr.15: 40459759-40459759 on Build GRCh38
rs144573633 Chr.15: 40459787-40459787 on Build GRCh38
rs755421064 Chr.15: 40459656-40459656 on Build GRCh38
rs759683131 Chr.15: 40459800-40459800 on Build GRCh38
rs371754615 Chr.15: 40459774-40459774 on Build GRCh38
rs766957915 Chr.15: 40459646-40459646 on Build GRCh38
rs768404480 Chr.15: 40459636-40459636 on Build GRCh38
rs765294338 Chr.15: 40459547-40459547 on Build GRCh38
rs766527016 Chr.15: 40459733-40459733 on Build GRCh38
rs150048194 Chr.15: 40459523-40459523 on Build GRCh38
rs902321004 Chr.15: 40459739-40459739 on Build GRCh38
rs143488235 Chr.15: 40459692-40459692 on Build GRCh38
rs770348013 Chr.15: 40459585-40459585 on Build GRCh38
rs374994019 Chr.15: 40459651-40459651 on Build GRCh38
rs140818617 Chr.15: 40459716-40459716 on Build GRCh38
rs775813863 Chr.15: 40459810-40459810 on Build GRCh38
rs779244674 Chr.15: 40459881-40459881 on Build GRCh38
rs369024568 Chr.15: 40459847-40459847 on Build GRCh38
rs775425103 Chr.15: 40459533-40459533 on Build GRCh38
rs764299958 Chr.15: 40459814-40459814 on Build GRCh38
rs763114810 Chr.15: 40459730-40459730 on Build GRCh38
rs770092994 Chr.15: 40459721-40459721 on Build GRCh38
rs766019498 Chr.15: 40459829-40459829 on Build GRCh38
rs760488359 Chr.15: 40459632-40459632 on Build GRCh38
rs148148082 Chr.15: 40459883-40459883 on Build GRCh38
rs761030589 Chr.15: 40459911-40459911 on Build GRCh38
rs756063015 Chr.15: 40459948-40459948 on Build GRCh38
rs200967955 Chr.15: 40459742-40459742 on Build GRCh38
rs1017020536 Chr.15: 40459619-40459619 on Build GRCh38
rs924047026 Chr.15: 40459955-40459955 on Build GRCh38
rs773282258 Chr.15: 40459725-40459725 on Build GRCh38
rs774628328 Chr.15: 40459798-40459798 on Build GRCh38
rs774998635 Chr.15: 40459736-40459736 on Build GRCh38
rs146446117 Chr.15: 40459698-40459698 on Build GRCh38
rs955828643 Chr.15: 40459706-40459706 on Build GRCh38
rs755371821 Chr.15: 40459567-40459567 on Build GRCh38
rs771344428 Chr.15: 40459868-40459868 on Build GRCh38
rs764353618 Chr.15: 40459537-40459537 on Build GRCh38
rs372997308 Chr.15: 40459848-40459848 on Build GRCh38
rs776069267 Chr.15: 40459906-40459906 on Build GRCh38
rs745435563 Chr.15: 40459697-40459697 on Build GRCh38
rs201303207 Chr.15: 40459812-40459812 on Build GRCh38
rs754014467 Chr.15: 40459542-40459542 on Build GRCh38
rs775387776 Chr.15: 40459622-40459622 on Build GRCh38
rs7172685 Chr.15: 40459804-40459804 on Build GRCh38
rs3743144 Chr.15: 40459935-40459935 on Build GRCh38
rs371861712 Chr.15: 40459639-40459639 on Build GRCh38
rs746521906 Chr.15: 40459815-40459815 on Build GRCh38
rs778169435 Chr.15: 40459602-40459602 on Build GRCh38
rs760059662 Chr.15: 40459748-40459748 on Build GRCh38
rs1056335833 Chr.15: 40459964-40459964 on Build GRCh38
rs532982645 Chr.15: 40459720-40459720 on Build GRCh38
rs200116152 Chr.15: 40459587-40459587 on Build GRCh38
rs200344667 Chr.15: 40459773-40459773 on Build GRCh38
rs182865888 Chr.15: 40459946-40459946 on Build GRCh38
rs144778374 Chr.15: 40459667-40459667 on Build GRCh38
rs984252393 Chr.15: 40459603-40459603 on Build GRCh38
rs777535263 Chr.15: 40459522-40459522 on Build GRCh38
rs1056769623 Chr.15: 40459750-40459750 on Build GRCh38
rs750107761 Chr.15: 40459820-40459820 on Build GRCh38
rs749811114 Chr.15: 40459756-40459756 on Build GRCh38
rs749400262 Chr.15: 40459792-40459792 on Build GRCh38
rs752532598 Chr.15: 40459849-40459849 on Build GRCh38
rs751160913 Chr.15: 40459941-40459941 on Build GRCh38
rs777373100 Chr.15: 40459858-40459858 on Build GRCh38
rs779523753 Chr.15: 40459782-40459782 on Build GRCh38
rs773513658 Chr.15: 40459790-40459790 on Build GRCh38
rs537044689 Chr.15: 40459532-40459532 on Build GRCh38
rs867849397 Chr.15: 40459566-40459566 on Build GRCh38
rs748905255 Chr.15: 40459576-40459576 on Build GRCh38
rs35802092 Chr.15: 40459558-40459558 on Build GRCh38
rs568426696 Chr.15: 40459957-40459957 on Build GRCh38
rs1047371529 Chr.15: 40459545-40459545 on Build GRCh38
rs141845947 Chr.15: 40459866-40459866 on Build GRCh38
rs756414945 Chr.15: 40459769-40459769 on Build GRCh38
rs776843244 Chr.15: 40459637-40459637 on Build GRCh38
rs768122732 Chr.15: 40459758-40459758 on Build GRCh38
rs145339527 Chr.15: 40459526-40459526 on Build GRCh38
rs764968544 Chr.15: 40459770-40459770 on Build GRCh38
rs750580950 Chr.15: 40459645-40459645 on Build GRCh38
rs771144157 Chr.15: 40459796-40459796 on Build GRCh38
rs774183424 Chr.15: 40459531-40459531 on Build GRCh38
rs746882646 Chr.15: 40459710-40459710 on Build GRCh38
rs935171387 Chr.15: 40459744-40459744 on Build GRCh38
rs772089802 Chr.15: 40459607-40459607 on Build GRCh38
rs767922787 Chr.15: 40459661-40459661 on Build GRCh38
rs886082428 Chr.15: 40459830-40459830 on Build GRCh38
rs74927458 Chr.15: 40459934-40459934 on Build GRCh38
rs140142679 Chr.15: 40459520-40459520 on Build GRCh38
rs961531645 Chr.15: 40459631-40459631 on Build GRCh38
rs202103992 Chr.15: 40459690-40459690 on Build GRCh38
rs753177633 Chr.15: 40459764-40459764 on Build GRCh38
rs557265103 Chr.15: 40459534-40459534 on Build GRCh38
rs772239640 Chr.15: 40459885-40459885 on Build GRCh38
rs972930077 Chr.15: 40459689-40459689 on Build GRCh38
rs761912886 Chr.15: 40459546-40459546 on Build GRCh38
rs199500209 Chr.15: 40459788-40459788 on Build GRCh38
rs766866689 Chr.15: 40459943-40459943 on Build GRCh38
rs1034959264 Chr.15: 40459525-40459525 on Build GRCh38
rs777590311 Chr.15: 40459572-40459572 on Build GRCh38
rs549165983 Chr.15: 40459775-40459775 on Build GRCh38

View Full Product Details