Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24157419-24157913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNLR1
Entrez Gene ID
163702
Gene Name
interferon lambda receptor 1
Gene Aliases
CRF2/12, IFNLR, IL-28R1, IL28RA, LICR2
UniGene
Hs.221375
Gene Chromosome Location
Chr.1: 24154156-24187275 on Build GRCh38
Transcripts
NM_170743.3 NM_173064.2 NM_173065.2 XM_006710394.3 XM_017000479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243797 c.1029C>A p.Y343* Chr.1: 24157664-24157664 on Build GRCh38
2051417 c.1012G>A p.V338I Chr.1: 24157681-24157681 on Build GRCh38
906704 c.1011C>T p.G337G Chr.1: 24157682-24157682 on Build GRCh38
2051414 c.1126C>T p.P376S Chr.1: 24157567-24157567 on Build GRCh38
6242148 c.929C>A p.A310D Chr.1: 24157764-24157764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779756118 Chr.1: 24157523-24157523 on Build GRCh38
rs760334052 Chr.1: 24157606-24157606 on Build GRCh38
rs779422742 Chr.1: 24157716-24157716 on Build GRCh38
rs746977640 Chr.1: 24157834-24157834 on Build GRCh38
rs74399859 Chr.1: 24157730-24157730 on Build GRCh38
rs770831697 Chr.1: 24157590-24157590 on Build GRCh38
rs116722436 Chr.1: 24157705-24157705 on Build GRCh38
rs760801237 Chr.1: 24157885-24157885 on Build GRCh38
rs760421826 Chr.1: 24157532-24157532 on Build GRCh38
rs984226142 Chr.1: 24157443-24157443 on Build GRCh38
rs749138239 Chr.1: 24157574-24157574 on Build GRCh38
rs777541683 Chr.1: 24157573-24157573 on Build GRCh38
rs755178790 Chr.1: 24157653-24157653 on Build GRCh38
rs141153963 Chr.1: 24157823-24157823 on Build GRCh38
rs773437363 Chr.1: 24157849-24157849 on Build GRCh38
rs781439385 Chr.1: 24157668-24157668 on Build GRCh38
rs143084792 Chr.1: 24157619-24157619 on Build GRCh38
rs71575792 Chr.1: 24157805-24157805 on Build GRCh38
rs767166431 Chr.1: 24157518-24157518 on Build GRCh38
rs540698164 Chr.1: 24157878-24157878 on Build GRCh38
rs746570821 Chr.1: 24157717-24157717 on Build GRCh38
rs1021721452 Chr.1: 24157549-24157549 on Build GRCh38
rs778755478 Chr.1: 24157593-24157593 on Build GRCh38
rs368149277 Chr.1: 24157852-24157852 on Build GRCh38
rs113018552 Chr.1: 24157484-24157484 on Build GRCh38
rs780083752 Chr.1: 24157830-24157830 on Build GRCh38
rs764869999 Chr.1: 24157797-24157797 on Build GRCh38
rs895467233 Chr.1: 24157616-24157616 on Build GRCh38
rs1057242155 Chr.1: 24157614-24157614 on Build GRCh38
rs370153471 Chr.1: 24157521-24157521 on Build GRCh38
rs997672602 Chr.1: 24157448-24157448 on Build GRCh38
rs747735842 Chr.1: 24157839-24157839 on Build GRCh38
rs773897609 Chr.1: 24157503-24157503 on Build GRCh38
rs368532574 Chr.1: 24157718-24157718 on Build GRCh38
rs1053378478 Chr.1: 24157796-24157796 on Build GRCh38
rs763529301 Chr.1: 24157624-24157624 on Build GRCh38
rs150863617 Chr.1: 24157725-24157725 on Build GRCh38
rs543005854 Chr.1: 24157781-24157781 on Build GRCh38
rs763367931 Chr.1: 24157764-24157764 on Build GRCh38
rs138679926 Chr.1: 24157474-24157474 on Build GRCh38
rs766969987 Chr.1: 24157768-24157768 on Build GRCh38
rs568899756 Chr.1: 24157547-24157547 on Build GRCh38
rs770738010 Chr.1: 24157501-24157503 on Build GRCh38
rs771310364 Chr.1: 24157858-24157858 on Build GRCh38
rs376141935 Chr.1: 24157603-24157603 on Build GRCh38
rs762398991 Chr.1: 24157554-24157554 on Build GRCh38
rs144531654 Chr.1: 24157808-24157808 on Build GRCh38
rs200381326 Chr.1: 24157782-24157782 on Build GRCh38
rs141659592 Chr.1: 24157609-24157609 on Build GRCh38
rs565033215 Chr.1: 24157440-24157440 on Build GRCh38
rs754256886 Chr.1: 24157551-24157551 on Build GRCh38
rs774236929 Chr.1: 24157518-24157519 on Build GRCh38
rs749158730 Chr.1: 24157478-24157478 on Build GRCh38
rs935452669 Chr.1: 24157510-24157510 on Build GRCh38
rs751730039 Chr.1: 24157649-24157649 on Build GRCh38
rs4298678 Chr.1: 24157613-24157613 on Build GRCh38
rs868705219 Chr.1: 24157576-24157576 on Build GRCh38
rs780206434 Chr.1: 24157566-24157566 on Build GRCh38
rs201737608 Chr.1: 24157669-24157669 on Build GRCh38
rs977164858 Chr.1: 24157460-24157460 on Build GRCh38
rs773933736 Chr.1: 24157795-24157795 on Build GRCh38
rs375114618 Chr.1: 24157848-24157848 on Build GRCh38
rs769529500 Chr.1: 24157472-24157472 on Build GRCh38
rs778788413 Chr.1: 24157690-24157690 on Build GRCh38
rs570292636 Chr.1: 24157747-24157747 on Build GRCh38
rs775075076 Chr.1: 24157605-24157605 on Build GRCh38
rs765896629 Chr.1: 24157557-24157557 on Build GRCh38
rs767194890 Chr.1: 24157441-24157441 on Build GRCh38
rs749709626 Chr.1: 24157850-24157850 on Build GRCh38
rs964289970 Chr.1: 24157456-24157456 on Build GRCh38
rs771431074 Chr.1: 24157596-24157596 on Build GRCh38
rs80148543 Chr.1: 24157681-24157681 on Build GRCh38
rs775547470 Chr.1: 24157879-24157879 on Build GRCh38
rs1052512012 Chr.1: 24157526-24157526 on Build GRCh38
rs764432341 Chr.1: 24157890-24157890 on Build GRCh38
rs201444954 Chr.1: 24157875-24157875 on Build GRCh38
rs267598459 Chr.1: 24157567-24157567 on Build GRCh38
rs113490731 Chr.1: 24157760-24157760 on Build GRCh38
rs758373936 Chr.1: 24157562-24157562 on Build GRCh38
rs745848085 Chr.1: 24157595-24157595 on Build GRCh38
rs145001756 Chr.1: 24157779-24157779 on Build GRCh38
rs774798920 Chr.1: 24157862-24157862 on Build GRCh38
rs532465282 Chr.1: 24157445-24157445 on Build GRCh38
rs201566947 Chr.1: 24157741-24157741 on Build GRCh38
rs918384348 Chr.1: 24157664-24157664 on Build GRCh38
rs757005624 Chr.1: 24157686-24157686 on Build GRCh38
rs776388185 Chr.1: 24157729-24157729 on Build GRCh38
rs72648587 Chr.1: 24157618-24157618 on Build GRCh38
rs766304881 Chr.1: 24157645-24157645 on Build GRCh38
rs573297615 Chr.1: 24157835-24157835 on Build GRCh38
rs866174990 Chr.1: 24157636-24157636 on Build GRCh38
rs978627338 Chr.1: 24157863-24157863 on Build GRCh38
rs772933526 Chr.1: 24157561-24157561 on Build GRCh38
rs1012045768 Chr.1: 24157516-24157516 on Build GRCh38
rs1040762399 Chr.1: 24157750-24157750 on Build GRCh38
rs575592471 Chr.1: 24157778-24157778 on Build GRCh38
rs759017398 Chr.1: 24157506-24157506 on Build GRCh38
rs200332015 Chr.1: 24157480-24157480 on Build GRCh38

View Full Product Details