Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67207751-67208253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC29
Entrez Gene ID
26231
Gene Name
leucine rich repeat containing 29
Gene Aliases
FBL9, FBXL9
UniGene
Hs.461000
Gene Chromosome Location
Chr.16: 67207139-67227062 on Build GRCh38
Transcripts
NM_001004055.1 NM_012163.2 XM_017023126.1 XM_017023127.1 XM_017023128.1 XM_017023129.1 XM_017023130.1 XM_017023131.1 XM_017023132.1 XM_017023133.1 XM_017023134.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1256804 c.446G>A p.S149N Chr.16: 67207930-67207930 on Build GRCh38
4543755 c.343G>A p.D115N Chr.16: 67208033-67208033 on Build GRCh38
6703735 c.482G>A p.S161N Chr.16: 67207894-67207894 on Build GRCh38
7375576 c.494T>C p.L165P Chr.16: 67207882-67207882 on Build GRCh38
236480 c.502C>A p.Q168K Chr.16: 67207874-67207874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201737046 Chr.16: 67207893-67207893 on Build GRCh38
rs768313340 Chr.16: 67207911-67207911 on Build GRCh38
rs774725737 Chr.16: 67207826-67207826 on Build GRCh38
rs768175886 Chr.16: 67207835-67207835 on Build GRCh38
rs967929874 Chr.16: 67208035-67208035 on Build GRCh38
rs34461431 Chr.16: 67207981-67207981 on Build GRCh38
rs759506271 Chr.16: 67207969-67207969 on Build GRCh38
rs772569052 Chr.16: 67208037-67208037 on Build GRCh38
rs775075598 Chr.16: 67207806-67207806 on Build GRCh38
rs776982797 Chr.16: 67208087-67208087 on Build GRCh38
rs755168119 Chr.16: 67207852-67207852 on Build GRCh38
rs113434123 Chr.16: 67207963-67207963 on Build GRCh38
rs776721799 Chr.16: 67207964-67207964 on Build GRCh38
rs377193332 Chr.16: 67207859-67207859 on Build GRCh38
rs56234712 Chr.16: 67207851-67207851 on Build GRCh38
rs779561425 Chr.16: 67207902-67207902 on Build GRCh38
rs920140475 Chr.16: 67207885-67207885 on Build GRCh38
rs759942001 Chr.16: 67208112-67208112 on Build GRCh38
rs369292006 Chr.16: 67208078-67208078 on Build GRCh38
rs748918853 Chr.16: 67207904-67207904 on Build GRCh38
rs199784030 Chr.16: 67207991-67207991 on Build GRCh38
rs774283271 Chr.16: 67207816-67207816 on Build GRCh38
rs745432563 Chr.16: 67207793-67207793 on Build GRCh38
rs74849615 Chr.16: 67207882-67207882 on Build GRCh38
rs766475374 Chr.16: 67207841-67207841 on Build GRCh38
rs992688752 Chr.16: 67208141-67208141 on Build GRCh38
rs765817654 Chr.16: 67208215-67208215 on Build GRCh38
rs750182247 Chr.16: 67208137-67208137 on Build GRCh38
rs777556830 Chr.16: 67207785-67207785 on Build GRCh38
rs201641535 Chr.16: 67208005-67208005 on Build GRCh38
rs777071757 Chr.16: 67208094-67208094 on Build GRCh38
rs765228230 Chr.16: 67208105-67208105 on Build GRCh38
rs1000788178 Chr.16: 67208221-67208221 on Build GRCh38
rs948240813 Chr.16: 67207857-67207857 on Build GRCh38
rs868675768 Chr.16: 67208182-67208182 on Build GRCh38
rs750376508 Chr.16: 67207998-67207998 on Build GRCh38
rs779399678 Chr.16: 67207791-67207791 on Build GRCh38
rs781164252 Chr.16: 67207890-67207890 on Build GRCh38
rs1056106714 Chr.16: 67208150-67208150 on Build GRCh38
rs918120945 Chr.16: 67207854-67207854 on Build GRCh38
rs754420158 Chr.16: 67208022-67208022 on Build GRCh38
rs138009426 Chr.16: 67208084-67208084 on Build GRCh38
rs779459404 Chr.16: 67208009-67208009 on Build GRCh38
rs200531835 Chr.16: 67207933-67207933 on Build GRCh38
rs569126198 Chr.16: 67207959-67207959 on Build GRCh38
rs751397817 Chr.16: 67207868-67207868 on Build GRCh38
rs896316001 Chr.16: 67207865-67207865 on Build GRCh38
rs773933020 Chr.16: 67207900-67207900 on Build GRCh38
rs563212209 Chr.16: 67208177-67208177 on Build GRCh38
rs760895682 Chr.16: 67207836-67207836 on Build GRCh38
rs956729772 Chr.16: 67208139-67208139 on Build GRCh38
rs746959997 Chr.16: 67207788-67207788 on Build GRCh38
rs768918919 Chr.16: 67207808-67207808 on Build GRCh38
rs147035656 Chr.16: 67207976-67207976 on Build GRCh38
rs537308243 Chr.16: 67207927-67207927 on Build GRCh38
rs150382034 Chr.16: 67208028-67208028 on Build GRCh38
rs185256293 Chr.16: 67207807-67207807 on Build GRCh38
rs945187901 Chr.16: 67208176-67208176 on Build GRCh38
rs34595673 Chr.16: 67208027-67208027 on Build GRCh38
rs767324895 Chr.16: 67208128-67208128 on Build GRCh38
rs931566991 Chr.16: 67208096-67208096 on Build GRCh38
rs1009446196 Chr.16: 67207879-67207879 on Build GRCh38
rs769284183 Chr.16: 67207794-67207794 on Build GRCh38
rs190512815 Chr.16: 67208125-67208125 on Build GRCh38
rs766338312 Chr.16: 67207782-67207782 on Build GRCh38
rs748866934 Chr.16: 67208016-67208016 on Build GRCh38
rs577048292 Chr.16: 67208097-67208097 on Build GRCh38
rs760984061 Chr.16: 67207836-67207837 on Build GRCh38
rs896325920 Chr.16: 67208164-67208164 on Build GRCh38
rs775698364 Chr.16: 67208107-67208107 on Build GRCh38
rs536661378 Chr.16: 67207982-67207982 on Build GRCh38
rs904968088 Chr.16: 67207840-67207840 on Build GRCh38
rs758194014 Chr.16: 67207783-67207783 on Build GRCh38
rs979403201 Chr.16: 67207820-67207820 on Build GRCh38
rs772546944 Chr.16: 67207942-67207942 on Build GRCh38
rs969964802 Chr.16: 67208197-67208197 on Build GRCh38
rs773651702 Chr.16: 67208049-67208049 on Build GRCh38
rs372190285 Chr.16: 67207792-67207792 on Build GRCh38

View Full Product Details