Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 146807998-146807492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYDIN
Entrez Gene ID
54768
Gene Name
HYDIN, axonemal central pair apparatus protein
Gene Aliases
CILD5, HYDIN1, HYDIN2, PPP1R31
UniGene
Hs.351914
Gene Chromosome Location
Chr.16: 70802084-71230722 on Build GRCh38
Transcripts
NM_001270974.2 XM_006721206.2 XM_011523146.2 XM_011523147.1 XM_011523148.1 XM_011523151.1 XM_011523152.1 XM_011523155.2 XM_017023346.1 XM_017023347.1 XM_017023348.1 NR_103556.1

Gene Symbol
HYDIN2
Entrez Gene ID
100288805
Gene Name
HYDIN2, axonemal central pair apparatus protein (pseudogene)
Gene Aliases
HYDINP1
UniGene
Hs.648712
Gene Chromosome Location
Chr.1: 146547489-146822034 on Build GRCh38
Transcripts
NM_001270974.2 XM_006721206.2 XM_011523146.2 XM_011523147.1 XM_011523148.1 XM_011523151.1 XM_011523152.1 XM_011523155.2 XM_017023346.1 XM_017023347.1 XM_017023348.1 NR_103556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6482156 c.1140G>T p.T380T Chr.16: 70907419-70907419 on Build GRCh38
6301127 c.8308T>C p.Y2770H Chr.16: 70907433-70907433 on Build GRCh38
6482155 c.8322G>T p.T2774T Chr.16: 70907419-70907419 on Build GRCh38
6028161 c.7904_8369+366del1810 p.? Chr.16: 70907006-70908815 on Build GRCh38
6028162 c.722_1187+366del1810 p.? Chr.16: 70907006-70908815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541076248 Chr.16: 70907679-70907679 on Build GRCh38
rs754127824 Chr.16: 70907431-70907431 on Build GRCh38
rs202224164 Chr.16: 70907415-70907415 on Build GRCh38
rs200410378 Chr.16: 70907459-70907459 on Build GRCh38
rs1049626933 Chr.16: 70907635-70907635 on Build GRCh38
rs1000146353 Chr.16: 70907683-70907683 on Build GRCh38
rs535935975 Chr.16: 70907365-70907365 on Build GRCh38
rs558420833 Chr.16: 70907585-70907585 on Build GRCh38
rs200420917 Chr.16: 70907368-70907368 on Build GRCh38
rs540050976 Chr.16: 70907581-70907581 on Build GRCh38
rs368950142 Chr.16: 70907413-70907413 on Build GRCh38
rs1031327471 Chr.16: 70907705-70907705 on Build GRCh38
rs532929283 Chr.16: 70907744-70907744 on Build GRCh38
rs576787960 Chr.16: 70907604-70907604 on Build GRCh38
rs543808041 Chr.16: 70907607-70907607 on Build GRCh38
rs755427774 Chr.16: 70907440-70907440 on Build GRCh38
rs113656574 Chr.16: 70907461-70907461 on Build GRCh38
rs1798485 Chr.16: 70907717-70907717 on Build GRCh38
rs779556830 Chr.16: 70907446-70907446 on Build GRCh38
rs566246383 Chr.16: 70907546-70907546 on Build GRCh38
rs980348652 Chr.16: 70907334-70907334 on Build GRCh38
rs573563795 Chr.16: 70907669-70907669 on Build GRCh38
rs760766538 Chr.16: 70907419-70907419 on Build GRCh38
rs917401554 Chr.16: 70907322-70907322 on Build GRCh38
rs796226719 Chr.16: 70907675-70907676 on Build GRCh38
rs1013987622 Chr.16: 70907754-70907754 on Build GRCh38
rs868463319 Chr.16: 70907481-70907481 on Build GRCh38
rs750534052 Chr.16: 70907395-70907395 on Build GRCh38
rs376087083 Chr.16: 70907387-70907387 on Build GRCh38
rs201896117 Chr.16: 70907475-70907475 on Build GRCh38
rs1798486 Chr.16: 70907620-70907620 on Build GRCh38
rs555332195 Chr.16: 70907648-70907648 on Build GRCh38
rs867874027 Chr.16: 70907598-70907598 on Build GRCh38
rs376675911 Chr.16: 70907684-70907685 on Build GRCh38
rs944775442 Chr.16: 70907460-70907460 on Build GRCh38
rs904435903 Chr.16: 70907658-70907658 on Build GRCh38
rs900977248 Chr.16: 70907489-70907489 on Build GRCh38
rs916994889 Chr.16: 70907498-70907498 on Build GRCh38
rs200593817 Chr.16: 70907474-70907474 on Build GRCh38
rs191001028 Chr.16: 70907710-70907710 on Build GRCh38
rs970646747 Chr.16: 70907323-70907323 on Build GRCh38
rs766663360 Chr.16: 70907426-70907426 on Build GRCh38
rs971228211 Chr.16: 70907591-70907591 on Build GRCh38

View Full Product Details