Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10303108-10303595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH13
Entrez Gene ID
8735
Gene Name
myosin heavy chain 13
Gene Aliases
MyHC-IIL, MyHC-eo
UniGene
Hs.711142
Gene Chromosome Location
Chr.17: 10300866-10373005 on Build GRCh38
Transcripts
NM_003802.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6145702 c.5554A>T p.K1852* Chr.17: 10303411-10303411 on Build GRCh38
6145701 c.5554A>T p.K1852* Chr.17: 10303411-10303411 on Build GRCh38
3937229 c.5473G>A p.E1825K Chr.17: 10303492-10303492 on Build GRCh38
2924283 c.5608G>A p.D1870N Chr.17: 10303255-10303255 on Build GRCh38
3402563 c.5641T>C p.S1881P Chr.17: 10303222-10303222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761531272 Chr.17: 10303281-10303281 on Build GRCh38
rs180735007 Chr.17: 10303161-10303161 on Build GRCh38
rs767538459 Chr.17: 10303489-10303489 on Build GRCh38
rs770725242 Chr.17: 10303546-10303546 on Build GRCh38
rs372183463 Chr.17: 10303457-10303457 on Build GRCh38
rs749886416 Chr.17: 10303197-10303197 on Build GRCh38
rs772107063 Chr.17: 10303322-10303322 on Build GRCh38
rs780603557 Chr.17: 10303337-10303337 on Build GRCh38
rs759176017 Chr.17: 10303494-10303495 on Build GRCh38
rs370780677 Chr.17: 10303190-10303190 on Build GRCh38
rs931055650 Chr.17: 10303569-10303569 on Build GRCh38
rs900650148 Chr.17: 10303554-10303554 on Build GRCh38
rs773938731 Chr.17: 10303469-10303469 on Build GRCh38
rs774892955 Chr.17: 10303172-10303172 on Build GRCh38
rs767313943 Chr.17: 10303395-10303395 on Build GRCh38
rs776804584 Chr.17: 10303280-10303280 on Build GRCh38
rs770281302 Chr.17: 10303452-10303452 on Build GRCh38
rs770388142 Chr.17: 10303155-10303158 on Build GRCh38
rs761201873 Chr.17: 10303290-10303290 on Build GRCh38
rs373608771 Chr.17: 10303217-10303217 on Build GRCh38
rs781531789 Chr.17: 10303425-10303425 on Build GRCh38
rs1028288241 Chr.17: 10303564-10303564 on Build GRCh38
rs918328798 Chr.17: 10303315-10303315 on Build GRCh38
rs775259767 Chr.17: 10303491-10303491 on Build GRCh38
rs558521017 Chr.17: 10303535-10303535 on Build GRCh38
rs772496865 Chr.17: 10303175-10303175 on Build GRCh38
rs140895457 Chr.17: 10303495-10303495 on Build GRCh38
rs996374133 Chr.17: 10303555-10303555 on Build GRCh38
rs953660690 Chr.17: 10303453-10303453 on Build GRCh38
rs748590797 Chr.17: 10303440-10303440 on Build GRCh38
rs770638948 Chr.17: 10303460-10303460 on Build GRCh38
rs758917055 Chr.17: 10303481-10303481 on Build GRCh38
rs773900297 Chr.17: 10303298-10303299 on Build GRCh38
rs751513177 Chr.17: 10303299-10303299 on Build GRCh38
rs953614598 Chr.17: 10303240-10303240 on Build GRCh38
rs771036994 Chr.17: 10303165-10303165 on Build GRCh38
rs748958560 Chr.17: 10303458-10303458 on Build GRCh38
rs765976474 Chr.17: 10303389-10303389 on Build GRCh38
rs780094985 Chr.17: 10303210-10303210 on Build GRCh38
rs781052633 Chr.17: 10303306-10303306 on Build GRCh38
rs749369845 Chr.17: 10303158-10303158 on Build GRCh38
rs755881748 Chr.17: 10303399-10303399 on Build GRCh38
rs528285170 Chr.17: 10303459-10303459 on Build GRCh38
rs895759074 Chr.17: 10303393-10303393 on Build GRCh38
rs759520191 Chr.17: 10303394-10303394 on Build GRCh38
rs559233974 Chr.17: 10303420-10303420 on Build GRCh38
rs1019107240 Chr.17: 10303550-10303557 on Build GRCh38
rs912177822 Chr.17: 10303285-10303285 on Build GRCh38
rs3744550 Chr.17: 10303278-10303278 on Build GRCh38
rs367784389 Chr.17: 10303256-10303256 on Build GRCh38
rs745996968 Chr.17: 10303321-10303321 on Build GRCh38
rs773464497 Chr.17: 10303147-10303147 on Build GRCh38
rs144305118 Chr.17: 10303308-10303308 on Build GRCh38
rs764819259 Chr.17: 10303196-10303196 on Build GRCh38
rs760697599 Chr.17: 10303405-10303405 on Build GRCh38
rs1035671121 Chr.17: 10303139-10303139 on Build GRCh38
rs867111476 Chr.17: 10303309-10303309 on Build GRCh38
rs372688195 Chr.17: 10303275-10303275 on Build GRCh38
rs750637766 Chr.17: 10303406-10303406 on Build GRCh38
rs766462483 Chr.17: 10303507-10303507 on Build GRCh38
rs781782326 Chr.17: 10303519-10303519 on Build GRCh38
rs1048188012 Chr.17: 10303364-10303364 on Build GRCh38
rs778284222 Chr.17: 10303539-10303539 on Build GRCh38
rs769623186 Chr.17: 10303367-10303367 on Build GRCh38
rs758559636 Chr.17: 10303408-10303408 on Build GRCh38
rs760044832 Chr.17: 10303174-10303174 on Build GRCh38
rs777085794 Chr.17: 10303357-10303357 on Build GRCh38
rs530814088 Chr.17: 10303234-10303234 on Build GRCh38
rs763700519 Chr.17: 10303184-10303184 on Build GRCh38
rs554038619 Chr.17: 10303353-10303353 on Build GRCh38
rs543090373 Chr.17: 10303499-10303499 on Build GRCh38
rs772590454 Chr.17: 10303261-10303261 on Build GRCh38
rs779057316 Chr.17: 10303318-10303318 on Build GRCh38
rs1001862159 Chr.17: 10303130-10303130 on Build GRCh38
rs754985281 Chr.17: 10303514-10303514 on Build GRCh38
rs751711488 Chr.17: 10303512-10303512 on Build GRCh38
rs200438057 Chr.17: 10303391-10303391 on Build GRCh38
rs754784056 Chr.17: 10303301-10303301 on Build GRCh38
rs947978108 Chr.17: 10303418-10303418 on Build GRCh38
rs767224849 Chr.17: 10303532-10303534 on Build GRCh38
rs369185144 Chr.17: 10303424-10303424 on Build GRCh38
rs201316135 Chr.17: 10303340-10303340 on Build GRCh38
rs754772846 Chr.17: 10303219-10303219 on Build GRCh38
rs749628632 Chr.17: 10303544-10303544 on Build GRCh38
rs960040117 Chr.17: 10303142-10303142 on Build GRCh38
rs374874203 Chr.17: 10303494-10303494 on Build GRCh38
rs772972370 Chr.17: 10303381-10303381 on Build GRCh38
rs769053503 Chr.17: 10303279-10303279 on Build GRCh38
rs35367939 Chr.17: 10303139-10303140 on Build GRCh38
rs748413974 Chr.17: 10303359-10303359 on Build GRCh38
rs372224078 Chr.17: 10303286-10303286 on Build GRCh38
rs978942656 Chr.17: 10303189-10303189 on Build GRCh38
rs770998019 Chr.17: 10303250-10303250 on Build GRCh38
rs745908111 Chr.17: 10303258-10303258 on Build GRCh38
rs749510137 Chr.17: 10303246-10303246 on Build GRCh38
rs775862240 Chr.17: 10303176-10303176 on Build GRCh38
rs1057058668 Chr.17: 10303392-10303392 on Build GRCh38
rs752555483 Chr.17: 10303398-10303398 on Build GRCh38
rs150001403 Chr.17: 10303177-10303177 on Build GRCh38
rs1008010920 Chr.17: 10303545-10303545 on Build GRCh38
rs753497489 Chr.17: 10303185-10303185 on Build GRCh38
rs905709907 Chr.17: 10303369-10303369 on Build GRCh38
rs748654105 Chr.17: 10303528-10303528 on Build GRCh38
rs758441670 Chr.17: 10303198-10303198 on Build GRCh38
rs184401376 Chr.17: 10303388-10303388 on Build GRCh38

View Full Product Details