Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38256294-38256747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R14A
Entrez Gene ID
94274
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14A
Gene Aliases
CPI-17, CPI17, PPP1INL
UniGene
Hs.631569
Gene Chromosome Location
Chr.19: 38251237-38256591 on Build GRCh38
Transcripts
NM_001243947.1 NM_033256.2

SNP Detail

SNP ID Chromosome Location
rs933554886 Chr.19: 38256405-38256405 on Build GRCh38
rs980532074 Chr.19: 38256515-38256515 on Build GRCh38
rs955481740 Chr.19: 38256706-38256706 on Build GRCh38
rs911836299 Chr.19: 38256594-38256594 on Build GRCh38
rs534310219 Chr.19: 38256578-38256578 on Build GRCh38
rs1025997661 Chr.19: 38256415-38256415 on Build GRCh38
rs1039335513 Chr.19: 38256609-38256609 on Build GRCh38
rs898855547 Chr.19: 38256610-38256610 on Build GRCh38
rs994560180 Chr.19: 38256408-38256408 on Build GRCh38
rs555545714 Chr.19: 38256593-38256593 on Build GRCh38
rs1001805803 Chr.19: 38256670-38256670 on Build GRCh38
rs913353398 Chr.19: 38256685-38256685 on Build GRCh38
rs757355373 Chr.19: 38256344-38256344 on Build GRCh38
rs1047478921 Chr.19: 38256622-38256622 on Build GRCh38
rs898948837 Chr.19: 38256382-38256382 on Build GRCh38
rs970335080 Chr.19: 38256504-38256504 on Build GRCh38
rs940830471 Chr.19: 38256475-38256475 on Build GRCh38
rs78814973 Chr.19: 38256677-38256677 on Build GRCh38
rs566708397 Chr.19: 38256552-38256552 on Build GRCh38
rs572714205 Chr.19: 38256725-38256725 on Build GRCh38
rs987739275 Chr.19: 38256721-38256721 on Build GRCh38
rs867461769 Chr.19: 38256634-38256634 on Build GRCh38
rs950890485 Chr.19: 38256449-38256449 on Build GRCh38
rs945198362 Chr.19: 38256582-38256582 on Build GRCh38
rs138141663 Chr.19: 38256681-38256681 on Build GRCh38
rs973543968 Chr.19: 38256689-38256689 on Build GRCh38
rs958058261 Chr.19: 38256330-38256330 on Build GRCh38
rs574315699 Chr.19: 38256627-38256627 on Build GRCh38
rs7247241 Chr.19: 38256632-38256632 on Build GRCh38
rs551801396 Chr.19: 38256506-38256506 on Build GRCh38
rs999969588 Chr.19: 38256628-38256628 on Build GRCh38
rs572830131 Chr.19: 38256595-38256595 on Build GRCh38
rs895728751 Chr.19: 38256561-38256562 on Build GRCh38
rs908105368 Chr.19: 38256684-38256685 on Build GRCh38
rs970742399 Chr.19: 38256643-38256643 on Build GRCh38
rs560878936 Chr.19: 38256529-38256529 on Build GRCh38
rs974693757 Chr.19: 38256357-38256357 on Build GRCh38
rs765504162 Chr.19: 38256694-38256694 on Build GRCh38
rs7246386 Chr.19: 38256454-38256454 on Build GRCh38
rs988911355 Chr.19: 38256569-38256569 on Build GRCh38
rs943287809 Chr.19: 38256602-38256602 on Build GRCh38
rs988777030 Chr.19: 38256684-38256684 on Build GRCh38
rs925927987 Chr.19: 38256549-38256549 on Build GRCh38
rs751696283 Chr.19: 38256328-38256328 on Build GRCh38
rs8100395 Chr.19: 38256477-38256477 on Build GRCh38
rs922191913 Chr.19: 38256369-38256369 on Build GRCh38
rs962227834 Chr.19: 38256685-38256685 on Build GRCh38
rs764336163 Chr.19: 38256317-38256317 on Build GRCh38
rs996141572 Chr.19: 38256651-38256651 on Build GRCh38
rs564115753 Chr.19: 38256722-38256722 on Build GRCh38
rs913279877 Chr.19: 38256575-38256575 on Build GRCh38
rs988048400 Chr.19: 38256419-38256419 on Build GRCh38
rs994425370 Chr.19: 38256617-38256617 on Build GRCh38
rs184977156 Chr.19: 38256629-38256629 on Build GRCh38
rs920354340 Chr.19: 38256705-38256705 on Build GRCh38
rs1037484516 Chr.19: 38256476-38256476 on Build GRCh38

View Full Product Details