Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54361208-54361725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf73
Entrez Gene ID
129852
Gene Name
chromosome 2 open reading frame 73
Gene Aliases
-
UniGene
Hs.732626
Gene Chromosome Location
Chr.2: 54329961-54383673 on Build GRCh38
Transcripts
NM_001100396.1 XM_011532510.2

SNP Detail

SNP ID Chromosome Location
rs560010250 Chr.2: 54361257-54361257 on Build GRCh38
rs779503598 Chr.2: 54361501-54361501 on Build GRCh38
rs191005648 Chr.2: 54361702-54361702 on Build GRCh38
rs543288477 Chr.2: 54361635-54361635 on Build GRCh38
rs993589205 Chr.2: 54361438-54361438 on Build GRCh38
rs573283718 Chr.2: 54361484-54361484 on Build GRCh38
rs1016416137 Chr.2: 54361416-54361416 on Build GRCh38
rs892904692 Chr.2: 54361264-54361264 on Build GRCh38
rs12987146 Chr.2: 54361375-54361375 on Build GRCh38
rs947158466 Chr.2: 54361682-54361682 on Build GRCh38
rs140946693 Chr.2: 54361627-54361627 on Build GRCh38
rs920129694 Chr.2: 54361597-54361597 on Build GRCh38
rs932901257 Chr.2: 54361371-54361372 on Build GRCh38
rs974231749 Chr.2: 54361592-54361592 on Build GRCh38
rs34682303 Chr.2: 54361332-54361332 on Build GRCh38
rs145675647 Chr.2: 54361262-54361262 on Build GRCh38
rs992178565 Chr.2: 54361542-54361542 on Build GRCh38
rs981334155 Chr.2: 54361243-54361243 on Build GRCh38
rs756835261 Chr.2: 54361371-54361371 on Build GRCh38
rs190816145 Chr.2: 54361435-54361435 on Build GRCh38
rs958600384 Chr.2: 54361602-54361602 on Build GRCh38
rs889347283 Chr.2: 54361406-54361406 on Build GRCh38
rs1022550823 Chr.2: 54361480-54361480 on Build GRCh38
rs772439610 Chr.2: 54361603-54361603 on Build GRCh38
rs1038957321 Chr.2: 54361316-54361316 on Build GRCh38
rs1041812939 Chr.2: 54361290-54361290 on Build GRCh38
rs960721506 Chr.2: 54361540-54361540 on Build GRCh38
rs79251363 Chr.2: 54361528-54361528 on Build GRCh38
rs1026785760 Chr.2: 54361478-54361478 on Build GRCh38
rs201398531 Chr.2: 54361598-54361599 on Build GRCh38
rs570991856 Chr.2: 54361265-54361265 on Build GRCh38
rs928516864 Chr.2: 54361250-54361250 on Build GRCh38
rs1020820188 Chr.2: 54361686-54361686 on Build GRCh38
rs991753316 Chr.2: 54361606-54361606 on Build GRCh38
rs1018624312 Chr.2: 54361451-54361451 on Build GRCh38
rs908595958 Chr.2: 54361574-54361574 on Build GRCh38
rs902843261 Chr.2: 54361331-54361331 on Build GRCh38
rs940180303 Chr.2: 54361312-54361312 on Build GRCh38
rs147294483 Chr.2: 54361320-54361320 on Build GRCh38
rs940110674 Chr.2: 54361576-54361576 on Build GRCh38
rs997150247 Chr.2: 54361364-54361364 on Build GRCh38
rs892221957 Chr.2: 54361455-54361455 on Build GRCh38
rs947136826 Chr.2: 54361267-54361267 on Build GRCh38
rs185061835 Chr.2: 54361648-54361648 on Build GRCh38
rs773429178 Chr.2: 54361680-54361680 on Build GRCh38
rs980717324 Chr.2: 54361526-54361526 on Build GRCh38
rs180997835 Chr.2: 54361487-54361487 on Build GRCh38
rs1008170490 Chr.2: 54361415-54361415 on Build GRCh38
rs898716151 Chr.2: 54361359-54361359 on Build GRCh38

View Full Product Details